Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 122698615-122699200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
UniGene
Hs.649459
Gene Chromosome Location
Chr.7: 122318424-122886759 on Build GRCh38
Transcripts
NM_001009571.3 NM_001167940.1 NM_017954.10 XM_005250695.4 XM_005250696.4 XM_005250697.4 XM_005250698.4 XM_005250699.4 XM_005250700.4 XM_005250701.4 XM_005250702.4 XM_005250703.4 XM_005250704.4 XM_005250705.4 XM_005250706.4 XM_005250707.4 XM_005250708.3 XM_011516695.2 XM_011516696.2 XM_011516697.2 XM_017012794.1 XM_017012795.1 XM_017012796.1 XM_017012797.1 XM_017012798.1 XM_017012799.1 NM_139175.1

Gene Symbol
RNF133
Entrez Gene ID
168433
Gene Name
ring finger protein 133
Gene Aliases
-
UniGene
Hs.126730
Gene Chromosome Location
Chr.7: 122697712-122699154 on Build GRCh38
Transcripts
NM_001009571.3 NM_001167940.1 NM_017954.10 XM_005250695.4 XM_005250696.4 XM_005250697.4 XM_005250698.4 XM_005250699.4 XM_005250700.4 XM_005250701.4 XM_005250702.4 XM_005250703.4 XM_005250704.4 XM_005250705.4 XM_005250706.4 XM_005250707.4 XM_005250708.3 XM_011516695.2 XM_011516696.2 XM_011516697.2 XM_017012794.1 XM_017012795.1 XM_017012796.1 XM_017012797.1 XM_017012798.1 XM_017012799.1 NM_139175.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4840385 c.194G>A p.S65N Chr.7: 122698725-122698725 on Build GRCh38
353473 c.274T>G p.F92V Chr.7: 122698645-122698645 on Build GRCh38
4801892 c.9A>G p.L3L Chr.7: 122698910-122698910 on Build GRCh38
4769428 c.276C>A p.F92L Chr.7: 122698643-122698643 on Build GRCh38
1084846 c.271A>G p.I91V Chr.7: 122698648-122698648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777566705 Chr.7: 122698648-122698648 on Build GRCh38
rs771800452 Chr.7: 122698949-122698949 on Build GRCh38
rs995521707 Chr.7: 122698658-122698658 on Build GRCh38
rs201538143 Chr.7: 122698932-122698932 on Build GRCh38
rs763907543 Chr.7: 122698813-122698813 on Build GRCh38
rs759042567 Chr.7: 122698896-122698896 on Build GRCh38
rs923410009 Chr.7: 122698856-122698856 on Build GRCh38
rs749589745 Chr.7: 122698777-122698777 on Build GRCh38
rs749580566 Chr.7: 122698692-122698692 on Build GRCh38
rs774637986 Chr.7: 122698821-122698821 on Build GRCh38
rs757605766 Chr.7: 122699060-122699060 on Build GRCh38
rs778412885 Chr.7: 122698906-122698906 on Build GRCh38
rs561872205 Chr.7: 122699145-122699145 on Build GRCh38
rs769267976 Chr.7: 122698694-122698694 on Build GRCh38
rs748492482 Chr.7: 122698886-122698886 on Build GRCh38
rs758401726 Chr.7: 122698643-122698643 on Build GRCh38
rs540428066 Chr.7: 122699095-122699095 on Build GRCh38
rs755306284 Chr.7: 122698772-122698772 on Build GRCh38
rs759128596 Chr.7: 122699046-122699046 on Build GRCh38
rs1020016581 Chr.7: 122699160-122699160 on Build GRCh38
rs775423221 Chr.7: 122698703-122698703 on Build GRCh38
rs755182334 Chr.7: 122698870-122698870 on Build GRCh38
rs769542252 Chr.7: 122698798-122698798 on Build GRCh38
rs771548782 Chr.7: 122698741-122698742 on Build GRCh38
rs1036840205 Chr.7: 122699022-122699022 on Build GRCh38
rs1025576306 Chr.7: 122698704-122698704 on Build GRCh38
rs747491181 Chr.7: 122698907-122698907 on Build GRCh38
rs1001661032 Chr.7: 122698991-122698991 on Build GRCh38
rs759285030 Chr.7: 122698700-122698700 on Build GRCh38
rs780513026 Chr.7: 122698690-122698690 on Build GRCh38
rs541990483 Chr.7: 122699012-122699012 on Build GRCh38
rs370779825 Chr.7: 122698955-122698955 on Build GRCh38
rs757142742 Chr.7: 122698737-122698737 on Build GRCh38
rs989004442 Chr.7: 122698862-122698862 on Build GRCh38
rs751695080 Chr.7: 122698650-122698650 on Build GRCh38
rs775016583 Chr.7: 122698801-122698801 on Build GRCh38
rs923387540 Chr.7: 122699170-122699170 on Build GRCh38
rs769189158 Chr.7: 122698915-122698915 on Build GRCh38
rs775016679 Chr.7: 122698696-122698696 on Build GRCh38
rs753933818 Chr.7: 122698962-122698962 on Build GRCh38
rs780129485 Chr.7: 122698763-122698763 on Build GRCh38
rs769856521 Chr.7: 122698680-122698680 on Build GRCh38
rs750476006 Chr.7: 122698751-122698751 on Build GRCh38
rs745712640 Chr.7: 122699025-122699025 on Build GRCh38
rs892082783 Chr.7: 122699179-122699179 on Build GRCh38
rs974868981 Chr.7: 122698822-122698822 on Build GRCh38
rs746207151 Chr.7: 122698783-122698787 on Build GRCh38
rs751500392 Chr.7: 122698734-122698734 on Build GRCh38
rs71574716 Chr.7: 122698916-122698916 on Build GRCh38
rs762345843 Chr.7: 122698697-122698697 on Build GRCh38
rs774079776 Chr.7: 122698815-122698815 on Build GRCh38
rs769063800 Chr.7: 122698789-122698789 on Build GRCh38
rs750580667 Chr.7: 122698837-122698837 on Build GRCh38
rs889862312 Chr.7: 122699098-122699098 on Build GRCh38
rs74327836 Chr.7: 122698876-122698876 on Build GRCh38
rs1025566256 Chr.7: 122699062-122699062 on Build GRCh38
rs909278252 Chr.7: 122698790-122698790 on Build GRCh38
rs756395852 Chr.7: 122698753-122698753 on Build GRCh38
rs762642789 Chr.7: 122698713-122698713 on Build GRCh38
rs376509948 Chr.7: 122698672-122698672 on Build GRCh38
rs757340494 Chr.7: 122698655-122698655 on Build GRCh38
rs979134878 Chr.7: 122698931-122698931 on Build GRCh38
rs1042725911 Chr.7: 122698981-122698981 on Build GRCh38
rs370998344 Chr.7: 122698702-122698702 on Build GRCh38
rs145490506 Chr.7: 122698716-122698716 on Build GRCh38
rs867580063 Chr.7: 122698877-122698877 on Build GRCh38
rs775950251 Chr.7: 122698832-122698835 on Build GRCh38
rs754042065 Chr.7: 122698866-122698866 on Build GRCh38
rs762599656 Chr.7: 122698806-122698806 on Build GRCh38
rs748688523 Chr.7: 122698779-122698779 on Build GRCh38
rs781180655 Chr.7: 122698657-122698657 on Build GRCh38
rs770600227 Chr.7: 122698698-122698698 on Build GRCh38
rs768928637 Chr.7: 122698918-122698919 on Build GRCh38
rs577024939 Chr.7: 122698765-122698765 on Build GRCh38
rs1035959668 Chr.7: 122698677-122698677 on Build GRCh38
rs759878094 Chr.7: 122698963-122698963 on Build GRCh38
rs770016299 Chr.7: 122698806-122698807 on Build GRCh38
rs766394959 Chr.7: 122698961-122698961 on Build GRCh38
rs140329176 Chr.7: 122698699-122698699 on Build GRCh38
rs773030210 Chr.7: 122698950-122698950 on Build GRCh38
rs992231940 Chr.7: 122698972-122698972 on Build GRCh38
rs756165355 Chr.7: 122698840-122698840 on Build GRCh38
rs984777819 Chr.7: 122699115-122699115 on Build GRCh38
rs766418174 Chr.7: 122698844-122698844 on Build GRCh38
rs1008269834 Chr.7: 122699124-122699124 on Build GRCh38
rs767408635 Chr.7: 122698833-122698833 on Build GRCh38
rs866697270 Chr.7: 122698805-122698806 on Build GRCh38
rs1049801704 Chr.7: 122699061-122699061 on Build GRCh38
rs776323822 Chr.7: 122698785-122698785 on Build GRCh38
rs975193360 Chr.7: 122699165-122699165 on Build GRCh38
rs781776136 Chr.7: 122698766-122698767 on Build GRCh38
rs373118781 Chr.7: 122698929-122698929 on Build GRCh38
rs917862869 Chr.7: 122698990-122698990 on Build GRCh38
rs895606646 Chr.7: 122699041-122699041 on Build GRCh38
rs767604596 Chr.7: 122698747-122698747 on Build GRCh38
rs749471581 Chr.7: 122698768-122698768 on Build GRCh38
rs761589626 Chr.7: 122698934-122698934 on Build GRCh38
rs933475583 Chr.7: 122699174-122699174 on Build GRCh38
rs939927306 Chr.7: 122698997-122698997 on Build GRCh38
rs1016244450 Chr.7: 122699149-122699149 on Build GRCh38
rs763030573 Chr.7: 122698885-122698886 on Build GRCh38
rs576187735 Chr.7: 122698994-122698994 on Build GRCh38
rs137930197 Chr.7: 122698784-122698784 on Build GRCh38

View Full Product Details