Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9585137-9585663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WEE1
Entrez Gene ID
7465
Gene Name
WEE1 G2 checkpoint kinase
Gene Aliases
WEE1A, WEE1hu
UniGene
Hs.249441
Gene Chromosome Location
Chr.11: 9573681-9589767 on Build GRCh38
Transcripts
NM_001143976.1 NM_003390.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6293048 c.1289-7T>C p.? Chr.11: 9585251-9585251 on Build GRCh38
3703776 c.1289-6T>C p.? Chr.11: 9585252-9585252 on Build GRCh38
4598157 c.1463T>A p.L488* Chr.11: 9585520-9585520 on Build GRCh38
6822470 c.1311C>A p.T437T Chr.11: 9585280-9585280 on Build GRCh38
5616557 c.1395G>C p.G465G Chr.11: 9585452-9585452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759729611 Chr.11: 9585546-9585546 on Build GRCh38
rs369028164 Chr.11: 9585338-9585338 on Build GRCh38
rs775081501 Chr.11: 9585356-9585356 on Build GRCh38
rs532283069 Chr.11: 9585481-9585481 on Build GRCh38
rs889237710 Chr.11: 9585164-9585164 on Build GRCh38
rs958450506 Chr.11: 9585322-9585322 on Build GRCh38
rs147537355 Chr.11: 9585269-9585269 on Build GRCh38
rs773071613 Chr.11: 9585511-9585511 on Build GRCh38
rs759076836 Chr.11: 9585218-9585218 on Build GRCh38
rs919913614 Chr.11: 9585634-9585638 on Build GRCh38
rs748563069 Chr.11: 9585435-9585435 on Build GRCh38
rs769549337 Chr.11: 9585504-9585504 on Build GRCh38
rs546618567 Chr.11: 9585620-9585624 on Build GRCh38
rs549439989 Chr.11: 9585281-9585281 on Build GRCh38
rs751660078 Chr.11: 9585410-9585410 on Build GRCh38
rs761820389 Chr.11: 9585379-9585379 on Build GRCh38
rs943452383 Chr.11: 9585516-9585516 on Build GRCh38
rs755011027 Chr.11: 9585411-9585411 on Build GRCh38
rs953871056 Chr.11: 9585559-9585559 on Build GRCh38
rs748540296 Chr.11: 9585320-9585320 on Build GRCh38
rs199808779 Chr.11: 9585330-9585330 on Build GRCh38
rs766452044 Chr.11: 9585395-9585395 on Build GRCh38
rs756660270 Chr.11: 9585577-9585577 on Build GRCh38
rs763239694 Chr.11: 9585390-9585390 on Build GRCh38
rs774011159 Chr.11: 9585207-9585207 on Build GRCh38
rs973157417 Chr.11: 9585626-9585626 on Build GRCh38
rs755550141 Chr.11: 9585302-9585304 on Build GRCh38
rs554525969 Chr.11: 9585312-9585312 on Build GRCh38
rs771783134 Chr.11: 9585462-9585462 on Build GRCh38
rs10770042 Chr.11: 9585485-9585485 on Build GRCh38
rs751525556 Chr.11: 9585309-9585309 on Build GRCh38
rs139320767 Chr.11: 9585468-9585468 on Build GRCh38
rs778206244 Chr.11: 9585436-9585436 on Build GRCh38
rs908881551 Chr.11: 9585333-9585333 on Build GRCh38
rs957362887 Chr.11: 9585476-9585476 on Build GRCh38
rs764070023 Chr.11: 9585239-9585239 on Build GRCh38
rs967600603 Chr.11: 9585249-9585249 on Build GRCh38
rs939919590 Chr.11: 9585617-9585617 on Build GRCh38
rs1002629228 Chr.11: 9585605-9585605 on Build GRCh38
rs767100836 Chr.11: 9585232-9585232 on Build GRCh38
rs893786368 Chr.11: 9585558-9585558 on Build GRCh38
rs200402463 Chr.11: 9585317-9585317 on Build GRCh38
rs753248931 Chr.11: 9585430-9585430 on Build GRCh38
rs1035396174 Chr.11: 9585286-9585286 on Build GRCh38
rs749561677 Chr.11: 9585460-9585460 on Build GRCh38
rs779428631 Chr.11: 9585439-9585439 on Build GRCh38
rs201185443 Chr.11: 9585536-9585536 on Build GRCh38
rs1011921375 Chr.11: 9585214-9585214 on Build GRCh38
rs189955202 Chr.11: 9585600-9585600 on Build GRCh38
rs1036992721 Chr.11: 9585238-9585238 on Build GRCh38
rs765138840 Chr.11: 9585384-9585384 on Build GRCh38
rs760217387 Chr.11: 9585363-9585363 on Build GRCh38
rs760694319 Chr.11: 9585236-9585236 on Build GRCh38
rs1007530343 Chr.11: 9585181-9585181 on Build GRCh38
rs780245459 Chr.11: 9585297-9585297 on Build GRCh38
rs752766522 Chr.11: 9585573-9585573 on Build GRCh38
rs56411856 Chr.11: 9585472-9585472 on Build GRCh38
rs1020230309 Chr.11: 9585223-9585223 on Build GRCh38
rs866677842 Chr.11: 9585474-9585474 on Build GRCh38
rs774592234 Chr.11: 9585544-9585544 on Build GRCh38
rs895299385 Chr.11: 9585313-9585313 on Build GRCh38
rs779537241 Chr.11: 9585464-9585464 on Build GRCh38
rs758537908 Chr.11: 9585296-9585296 on Build GRCh38
rs781213335 Chr.11: 9585425-9585425 on Build GRCh38
rs1032347866 Chr.11: 9585184-9585184 on Build GRCh38
rs972707136 Chr.11: 9585364-9585364 on Build GRCh38
rs912479137 Chr.11: 9585606-9585606 on Build GRCh38
rs185025093 Chr.11: 9585376-9585376 on Build GRCh38
rs898582113 Chr.11: 9585253-9585253 on Build GRCh38
rs753711776 Chr.11: 9585268-9585268 on Build GRCh38
rs768749832 Chr.11: 9585369-9585369 on Build GRCh38
rs775950178 Chr.11: 9585565-9585565 on Build GRCh38
rs756597519 Chr.11: 9585432-9585432 on Build GRCh38
rs140162029 Chr.11: 9585332-9585332 on Build GRCh38
rs372628405 Chr.11: 9585326-9585326 on Build GRCh38
rs34883479 Chr.11: 9585341-9585341 on Build GRCh38
rs766655330 Chr.11: 9585542-9585542 on Build GRCh38
rs767639383 Chr.11: 9585561-9585561 on Build GRCh38
rs376659774 Chr.11: 9585386-9585386 on Build GRCh38
rs912595519 Chr.11: 9585174-9585174 on Build GRCh38
rs761910453 Chr.11: 9585498-9585498 on Build GRCh38
rs368181613 Chr.11: 9585276-9585276 on Build GRCh38
rs774861059 Chr.11: 9585234-9585234 on Build GRCh38
rs779239560 Chr.11: 9585416-9585417 on Build GRCh38

View Full Product Details