Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66668862-66669410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM4B
Entrez Gene ID
83759
Gene Name
RNA binding motif protein 4B
Gene Aliases
RBM30, RBM4L, ZCCHC15, ZCCHC21B, ZCRB3B
UniGene
Hs.618713
Gene Chromosome Location
Chr.11: 66664994-66677921 on Build GRCh38
Transcripts
NM_001286135.1 NM_031492.3 XM_011545297.2 XM_017018402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6764172 c.840C>T p.G280G Chr.11: 66668864-66668864 on Build GRCh38
6764173 c.452G>A p.R151Q Chr.11: 66669252-66669252 on Build GRCh38
6589059 c.562G>A p.E188K Chr.11: 66669142-66669142 on Build GRCh38
4035795 c.668G>A p.R223Q Chr.11: 66669036-66669036 on Build GRCh38
6764168 c.475C>T p.Q159* Chr.11: 66669229-66669229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767147918 Chr.11: 66669290-66669290 on Build GRCh38
rs754773604 Chr.11: 66668989-66668989 on Build GRCh38
rs138212594 Chr.11: 66669017-66669017 on Build GRCh38
rs769122239 Chr.11: 66669077-66669077 on Build GRCh38
rs375898846 Chr.11: 66669320-66669320 on Build GRCh38
rs765336366 Chr.11: 66668897-66668897 on Build GRCh38
rs775384561 Chr.11: 66668901-66668901 on Build GRCh38
rs77173656 Chr.11: 66669218-66669218 on Build GRCh38
rs760475606 Chr.11: 66669306-66669306 on Build GRCh38
rs746374542 Chr.11: 66669063-66669063 on Build GRCh38
rs770741804 Chr.11: 66669315-66669318 on Build GRCh38
rs746641399 Chr.11: 66669060-66669060 on Build GRCh38
rs763194930 Chr.11: 66668908-66668908 on Build GRCh38
rs541735212 Chr.11: 66669311-66669311 on Build GRCh38
rs372917670 Chr.11: 66669271-66669271 on Build GRCh38
rs758074854 Chr.11: 66669050-66669050 on Build GRCh38
rs145285259 Chr.11: 66669213-66669213 on Build GRCh38
rs778581838 Chr.11: 66669043-66669043 on Build GRCh38
rs780748448 Chr.11: 66669062-66669062 on Build GRCh38
rs371878027 Chr.11: 66669012-66669012 on Build GRCh38
rs951406935 Chr.11: 66669026-66669026 on Build GRCh38
rs762126688 Chr.11: 66669079-66669079 on Build GRCh38
rs761181043 Chr.11: 66668951-66668951 on Build GRCh38
rs758456481 Chr.11: 66668902-66668902 on Build GRCh38
rs1046105309 Chr.11: 66669298-66669298 on Build GRCh38
rs138950956 Chr.11: 66668984-66668984 on Build GRCh38
rs569497432 Chr.11: 66669377-66669377 on Build GRCh38
rs370275000 Chr.11: 66669214-66669218 on Build GRCh38
rs200334725 Chr.11: 66668993-66668993 on Build GRCh38
rs768836554 Chr.11: 66669057-66669057 on Build GRCh38
rs765179407 Chr.11: 66669122-66669122 on Build GRCh38
rs944995817 Chr.11: 66669258-66669258 on Build GRCh38
rs912606424 Chr.11: 66669067-66669067 on Build GRCh38
rs747809725 Chr.11: 66668932-66668932 on Build GRCh38
rs759393862 Chr.11: 66669115-66669115 on Build GRCh38
rs754925060 Chr.11: 66669042-66669042 on Build GRCh38
rs759769612 Chr.11: 66668964-66668964 on Build GRCh38
rs559431794 Chr.11: 66669068-66669068 on Build GRCh38
rs61741747 Chr.11: 66668999-66668999 on Build GRCh38
rs747808883 Chr.11: 66668994-66668994 on Build GRCh38
rs116912258 Chr.11: 66669339-66669339 on Build GRCh38
rs369450559 Chr.11: 66669245-66669245 on Build GRCh38
rs368007657 Chr.11: 66668969-66668969 on Build GRCh38
rs748398594 Chr.11: 66669176-66669176 on Build GRCh38
rs375176983 Chr.11: 66669078-66669078 on Build GRCh38
rs574603433 Chr.11: 66668892-66668892 on Build GRCh38
rs752459002 Chr.11: 66669123-66669123 on Build GRCh38
rs372105336 Chr.11: 66669061-66669061 on Build GRCh38
rs780093249 Chr.11: 66668981-66668981 on Build GRCh38
rs773147865 Chr.11: 66669221-66669222 on Build GRCh38
rs767232648 Chr.11: 66669246-66669246 on Build GRCh38
rs780002507 Chr.11: 66668915-66668915 on Build GRCh38
rs748998928 Chr.11: 66668921-66668921 on Build GRCh38
rs562073155 Chr.11: 66668941-66668941 on Build GRCh38
rs760057266 Chr.11: 66669013-66669013 on Build GRCh38
rs139219386 Chr.11: 66669135-66669135 on Build GRCh38
rs748255777 Chr.11: 66668999-66669000 on Build GRCh38
rs376254090 Chr.11: 66669310-66669310 on Build GRCh38
rs779429638 Chr.11: 66669326-66669326 on Build GRCh38
rs374733996 Chr.11: 66669049-66669049 on Build GRCh38
rs749622862 Chr.11: 66669069-66669069 on Build GRCh38
rs772273886 Chr.11: 66669194-66669194 on Build GRCh38
rs777996819 Chr.11: 66668903-66668903 on Build GRCh38
rs141045331 Chr.11: 66668945-66668945 on Build GRCh38
rs763934522 Chr.11: 66669239-66669239 on Build GRCh38
rs779422627 Chr.11: 66669317-66669317 on Build GRCh38
rs756890495 Chr.11: 66669252-66669252 on Build GRCh38
rs764283037 Chr.11: 66668975-66668975 on Build GRCh38
rs146310422 Chr.11: 66669003-66669003 on Build GRCh38
rs776454290 Chr.11: 66669344-66669346 on Build GRCh38
rs746480100 Chr.11: 66669000-66669000 on Build GRCh38
rs764372966 Chr.11: 66669024-66669024 on Build GRCh38
rs906236610 Chr.11: 66669313-66669313 on Build GRCh38
rs645750 Chr.11: 66669357-66669357 on Build GRCh38
rs200192363 Chr.11: 66669161-66669161 on Build GRCh38
rs374593903 Chr.11: 66668966-66668966 on Build GRCh38
rs772074463 Chr.11: 66669199-66669201 on Build GRCh38
rs771633119 Chr.11: 66668943-66668943 on Build GRCh38
rs750387679 Chr.11: 66669247-66669247 on Build GRCh38
rs756599086 Chr.11: 66669165-66669165 on Build GRCh38
rs753789368 Chr.11: 66669037-66669037 on Build GRCh38
rs745858956 Chr.11: 66668912-66668912 on Build GRCh38
rs148731962 Chr.11: 66669030-66669030 on Build GRCh38
rs150762625 Chr.11: 66669125-66669125 on Build GRCh38
rs773281090 Chr.11: 66669106-66669106 on Build GRCh38
rs774839109 Chr.11: 66669175-66669175 on Build GRCh38
rs777008884 Chr.11: 66668961-66668961 on Build GRCh38
rs770270607 Chr.11: 66669065-66669065 on Build GRCh38
rs529466760 Chr.11: 66668976-66668976 on Build GRCh38
rs766349642 Chr.11: 66669216-66669216 on Build GRCh38
rs951642146 Chr.11: 66669378-66669378 on Build GRCh38
rs763846511 Chr.11: 66669131-66669131 on Build GRCh38
rs749778270 Chr.11: 66669169-66669169 on Build GRCh38
rs780571888 Chr.11: 66669159-66669159 on Build GRCh38
rs561405668 Chr.11: 66669336-66669336 on Build GRCh38
rs756844438 Chr.11: 66669145-66669145 on Build GRCh38
rs771651424 Chr.11: 66668997-66668997 on Build GRCh38
rs775447359 Chr.11: 66669217-66669217 on Build GRCh38
rs763091259 Chr.11: 66669020-66669020 on Build GRCh38
rs142178494 Chr.11: 66669094-66669094 on Build GRCh38
rs142896309 Chr.11: 66669019-66669019 on Build GRCh38
rs371760573 Chr.11: 66668913-66668913 on Build GRCh38
rs150237340 Chr.11: 66668980-66668980 on Build GRCh38
rs1053443610 Chr.11: 66668963-66668963 on Build GRCh38
rs374671861 Chr.11: 66669333-66669333 on Build GRCh38
rs768380966 Chr.11: 66668923-66668923 on Build GRCh38
rs746901293 Chr.11: 66669309-66669311 on Build GRCh38
rs771359086 Chr.11: 66668957-66668957 on Build GRCh38
rs778483374 Chr.11: 66668927-66668927 on Build GRCh38
rs761590001 Chr.11: 66669168-66669168 on Build GRCh38
rs1027169528 Chr.11: 66669372-66669372 on Build GRCh38
rs772480668 Chr.11: 66668944-66668944 on Build GRCh38
rs187877778 Chr.11: 66669389-66669389 on Build GRCh38
rs146095184 Chr.11: 66669025-66669025 on Build GRCh38
rs201202684 Chr.11: 66669036-66669036 on Build GRCh38

View Full Product Details