Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 162597733-162598331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UAP1
Entrez Gene ID
6675
Gene Name
UDP-N-acetylglucosamine pyrophosphorylase 1
Gene Aliases
AGX, AGX1, AGX2, SPAG2
UniGene
Hs.492859
Gene Chromosome Location
Chr.1: 162561440-162599843 on Build GRCh38
Transcripts
NM_001324113.1 NM_001324114.1 NM_001324115.1 NM_001324116.1 NM_001324117.1 NM_003115.4 XM_011509913.2 XM_011509914.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4636856 c.1427A>C p.D476A Chr.1: 162597809-162597809 on Build GRCh38
4636855 c.1376A>C p.D459A Chr.1: 162597809-162597809 on Build GRCh38
5623741 c.1402C>T p.P468S Chr.1: 162597835-162597835 on Build GRCh38
5729279 c.1444G>A p.E482K Chr.1: 162597826-162597826 on Build GRCh38
3477436 c.1400C>T p.S467F Chr.1: 162597833-162597833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020190046 Chr.1: 162598217-162598217 on Build GRCh38
rs754634378 Chr.1: 162597775-162597775 on Build GRCh38
rs781168091 Chr.1: 162598202-162598202 on Build GRCh38
rs142766548 Chr.1: 162597957-162597957 on Build GRCh38
rs776766778 Chr.1: 162598006-162598006 on Build GRCh38
rs1044786134 Chr.1: 162598166-162598166 on Build GRCh38
rs559742547 Chr.1: 162598142-162598142 on Build GRCh38
rs376276062 Chr.1: 162598239-162598240 on Build GRCh38
rs78789978 Chr.1: 162598239-162598239 on Build GRCh38
rs558459395 Chr.1: 162597817-162597817 on Build GRCh38
rs914603092 Chr.1: 162598106-162598106 on Build GRCh38
rs138482868 Chr.1: 162598230-162598231 on Build GRCh38
rs540948832 Chr.1: 162598041-162598041 on Build GRCh38
rs892983590 Chr.1: 162598206-162598207 on Build GRCh38
rs1017862984 Chr.1: 162597993-162597993 on Build GRCh38
rs907162902 Chr.1: 162598187-162598188 on Build GRCh38
rs776946984 Chr.1: 162597878-162597878 on Build GRCh38
rs766168320 Chr.1: 162597772-162597772 on Build GRCh38
rs937434689 Chr.1: 162598201-162598201 on Build GRCh38
rs555523876 Chr.1: 162598026-162598026 on Build GRCh38
rs993266056 Chr.1: 162598222-162598222 on Build GRCh38
rs751037440 Chr.1: 162597934-162597934 on Build GRCh38
rs374610378 Chr.1: 162597806-162597806 on Build GRCh38
rs1005925301 Chr.1: 162598260-162598260 on Build GRCh38
rs1011351116 Chr.1: 162598207-162598207 on Build GRCh38
rs762680661 Chr.1: 162597766-162597766 on Build GRCh38
rs373129729 Chr.1: 162598002-162598002 on Build GRCh38
rs773155826 Chr.1: 162597757-162597757 on Build GRCh38
rs535311410 Chr.1: 162597816-162597816 on Build GRCh38
rs758828289 Chr.1: 162597842-162597842 on Build GRCh38
rs746945439 Chr.1: 162597778-162597779 on Build GRCh38
rs146953035 Chr.1: 162598228-162598228 on Build GRCh38
rs905077719 Chr.1: 162597768-162597768 on Build GRCh38
rs953047302 Chr.1: 162598240-162598240 on Build GRCh38
rs976272259 Chr.1: 162598007-162598007 on Build GRCh38
rs773069798 Chr.1: 162597904-162597904 on Build GRCh38
rs985861920 Chr.1: 162598001-162598001 on Build GRCh38
rs941489217 Chr.1: 162598018-162598021 on Build GRCh38
rs12043619 Chr.1: 162598280-162598280 on Build GRCh38
rs776481514 Chr.1: 162597885-162597887 on Build GRCh38
rs768377505 Chr.1: 162597779-162597779 on Build GRCh38
rs541701130 Chr.1: 162598025-162598025 on Build GRCh38
rs564008429 Chr.1: 162598099-162598099 on Build GRCh38
rs574074824 Chr.1: 162598018-162598018 on Build GRCh38
rs542889457 Chr.1: 162597855-162597855 on Build GRCh38
rs905145361 Chr.1: 162598182-162598182 on Build GRCh38
rs951763032 Chr.1: 162597970-162597970 on Build GRCh38
rs765086048 Chr.1: 162598229-162598229 on Build GRCh38
rs767021497 Chr.1: 162597777-162597777 on Build GRCh38
rs771484016 Chr.1: 162597802-162597802 on Build GRCh38
rs993193885 Chr.1: 162597850-162597850 on Build GRCh38
rs755669200 Chr.1: 162597784-162597784 on Build GRCh38
rs184949893 Chr.1: 162598038-162598038 on Build GRCh38
rs887489928 Chr.1: 162598236-162598236 on Build GRCh38
rs751197837 Chr.1: 162597774-162597774 on Build GRCh38
rs768804773 Chr.1: 162597868-162597868 on Build GRCh38
rs923081347 Chr.1: 162597973-162597973 on Build GRCh38
rs557177820 Chr.1: 162597963-162597963 on Build GRCh38
rs984824023 Chr.1: 162597981-162597981 on Build GRCh38
rs1056254400 Chr.1: 162598193-162598193 on Build GRCh38
rs571680420 Chr.1: 162597880-162597880 on Build GRCh38
rs1011465561 Chr.1: 162597775-162597775 on Build GRCh38
rs937909511 Chr.1: 162598186-162598186 on Build GRCh38
rs975710229 Chr.1: 162597949-162597952 on Build GRCh38
rs370373682 Chr.1: 162598212-162598212 on Build GRCh38
rs746378990 Chr.1: 162597830-162597830 on Build GRCh38
rs748427935 Chr.1: 162597910-162597910 on Build GRCh38
rs780626587 Chr.1: 162597848-162597848 on Build GRCh38
rs747234554 Chr.1: 162597865-162597865 on Build GRCh38
rs375091827 Chr.1: 162597902-162597902 on Build GRCh38
rs779813694 Chr.1: 162597758-162597758 on Build GRCh38
rs202007828 Chr.1: 162597780-162597780 on Build GRCh38

View Full Product Details