Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10088313-10088894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Gene Symbol
FANCD2OS
Entrez Gene ID
115795
Gene Name
FANCD2 opposite strand
Gene Aliases
C3orf24
UniGene
Hs.190413
Gene Chromosome Location
Chr.3: 10081320-10108603 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361663 c.3493C>T p.R1165W Chr.3: 10088475-10088475 on Build GRCh38
7076453 c.3517G>C p.E1173Q Chr.3: 10088499-10088499 on Build GRCh38
3845678 c.3609T>C p.I1203I Chr.3: 10088876-10088876 on Build GRCh38
4975870 c.3470C>T p.S1157F Chr.3: 10088452-10088452 on Build GRCh38
1580887 c.3494G>A p.R1165Q Chr.3: 10088476-10088476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753527115 Chr.3: 10088844-10088844 on Build GRCh38
rs751864173 Chr.3: 10088831-10088831 on Build GRCh38
rs373757862 Chr.3: 10088874-10088874 on Build GRCh38
rs757027350 Chr.3: 10088805-10088806 on Build GRCh38
rs989705622 Chr.3: 10088623-10088623 on Build GRCh38
rs993970736 Chr.3: 10088466-10088466 on Build GRCh38
rs748776499 Chr.3: 10088778-10088779 on Build GRCh38
rs369777245 Chr.3: 10088584-10088584 on Build GRCh38
rs780304251 Chr.3: 10088534-10088534 on Build GRCh38
rs527995189 Chr.3: 10088350-10088350 on Build GRCh38
rs777042522 Chr.3: 10088796-10088796 on Build GRCh38
rs756713676 Chr.3: 10088845-10088845 on Build GRCh38
rs753106716 Chr.3: 10088463-10088464 on Build GRCh38
rs143403347 Chr.3: 10088465-10088465 on Build GRCh38
rs779632231 Chr.3: 10088415-10088415 on Build GRCh38
rs999879941 Chr.3: 10088787-10088787 on Build GRCh38
rs531334776 Chr.3: 10088470-10088470 on Build GRCh38
rs371571394 Chr.3: 10088687-10088687 on Build GRCh38
rs1007481549 Chr.3: 10088543-10088543 on Build GRCh38
rs201550150 Chr.3: 10088779-10088779 on Build GRCh38
rs377716703 Chr.3: 10088587-10088587 on Build GRCh38
rs780867042 Chr.3: 10088437-10088437 on Build GRCh38
rs937744739 Chr.3: 10088700-10088700 on Build GRCh38
rs372696571 Chr.3: 10088751-10088751 on Build GRCh38
rs753964157 Chr.3: 10088513-10088513 on Build GRCh38
rs759348779 Chr.3: 10088475-10088475 on Build GRCh38
rs774328826 Chr.3: 10088471-10088471 on Build GRCh38
rs151316403 Chr.3: 10088506-10088506 on Build GRCh38
rs549507714 Chr.3: 10088476-10088476 on Build GRCh38
rs766698512 Chr.3: 10088825-10088825 on Build GRCh38
rs958261095 Chr.3: 10088596-10088596 on Build GRCh38
rs760823688 Chr.3: 10088495-10088495 on Build GRCh38
rs763380585 Chr.3: 10088808-10088808 on Build GRCh38
rs745765337 Chr.3: 10088540-10088540 on Build GRCh38
rs139397618 Chr.3: 10088675-10088675 on Build GRCh38
rs778650034 Chr.3: 10088810-10088811 on Build GRCh38
rs764296912 Chr.3: 10088407-10088407 on Build GRCh38
rs966929624 Chr.3: 10088644-10088644 on Build GRCh38
rs937692272 Chr.3: 10088378-10088378 on Build GRCh38
rs781725867 Chr.3: 10088435-10088439 on Build GRCh38
rs55804542 Chr.3: 10088562-10088562 on Build GRCh38
rs982259285 Chr.3: 10088683-10088683 on Build GRCh38
rs760588840 Chr.3: 10088782-10088782 on Build GRCh38
rs1021210267 Chr.3: 10088743-10088743 on Build GRCh38
rs370944916 Chr.3: 10088516-10088516 on Build GRCh38
rs35503714 Chr.3: 10088599-10088599 on Build GRCh38
rs778303459 Chr.3: 10088863-10088863 on Build GRCh38
rs891286855 Chr.3: 10088684-10088685 on Build GRCh38
rs756183300 Chr.3: 10088405-10088405 on Build GRCh38
rs761955186 Chr.3: 10088797-10088797 on Build GRCh38
rs745660691 Chr.3: 10088814-10088815 on Build GRCh38
rs758749466 Chr.3: 10088531-10088531 on Build GRCh38
rs746223604 Chr.3: 10088423-10088423 on Build GRCh38
rs567825484 Chr.3: 10088520-10088520 on Build GRCh38
rs764205355 Chr.3: 10088509-10088509 on Build GRCh38
rs138409505 Chr.3: 10088525-10088525 on Build GRCh38
rs927603832 Chr.3: 10088332-10088332 on Build GRCh38
rs747786350 Chr.3: 10088454-10088454 on Build GRCh38
rs781077760 Chr.3: 10088418-10088418 on Build GRCh38
rs755725087 Chr.3: 10088553-10088553 on Build GRCh38
rs777378549 Chr.3: 10088843-10088843 on Build GRCh38
rs770473878 Chr.3: 10088571-10088571 on Build GRCh38
rs369022159 Chr.3: 10088463-10088463 on Build GRCh38
rs757471770 Chr.3: 10088519-10088519 on Build GRCh38
rs777197502 Chr.3: 10088578-10088579 on Build GRCh38
rs749039770 Chr.3: 10088720-10088720 on Build GRCh38
rs138189159 Chr.3: 10088399-10088399 on Build GRCh38
rs1030454628 Chr.3: 10088535-10088535 on Build GRCh38
rs866731784 Chr.3: 10088447-10088447 on Build GRCh38
rs1020405243 Chr.3: 10088384-10088384 on Build GRCh38
rs914046293 Chr.3: 10088634-10088634 on Build GRCh38
rs886038374 Chr.3: 10088584-10088585 on Build GRCh38
rs754341039 Chr.3: 10088413-10088413 on Build GRCh38
rs750892839 Chr.3: 10088803-10088803 on Build GRCh38
rs1051232662 Chr.3: 10088643-10088643 on Build GRCh38
rs149146745 Chr.3: 10088414-10088414 on Build GRCh38
rs1009809333 Chr.3: 10088366-10088366 on Build GRCh38
rs775736013 Chr.3: 10088477-10088477 on Build GRCh38
rs868320152 Chr.3: 10088345-10088345 on Build GRCh38
rs547204364 Chr.3: 10088651-10088651 on Build GRCh38
rs949980921 Chr.3: 10088517-10088517 on Build GRCh38
rs748781422 Chr.3: 10088566-10088566 on Build GRCh38
rs113469756 Chr.3: 10088381-10088381 on Build GRCh38
rs758739775 Chr.3: 10088424-10088424 on Build GRCh38
rs756030123 Chr.3: 10088694-10088694 on Build GRCh38
rs774237163 Chr.3: 10088577-10088577 on Build GRCh38
rs764399250 Chr.3: 10088786-10088786 on Build GRCh38
rs2241276 Chr.3: 10088704-10088704 on Build GRCh38
rs755673512 Chr.3: 10088530-10088531 on Build GRCh38
rs755389495 Chr.3: 10088834-10088834 on Build GRCh38
rs544697633 Chr.3: 10088425-10088425 on Build GRCh38

View Full Product Details