Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 44734775-44735281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF501
Entrez Gene ID
115560
Gene Name
zinc finger protein 501
Gene Aliases
ZNF, ZNF52
UniGene
Hs.401045
Gene Chromosome Location
Chr.3: 44729606-44737083 on Build GRCh38
Transcripts
NM_001258280.1 NM_145044.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
338745 c.386C>G p.T129S Chr.3: 44734807-44734807 on Build GRCh38
3067314 c.666G>A p.E222E Chr.3: 44735087-44735087 on Build GRCh38
3067313 c.661A>T p.S221C Chr.3: 44735082-44735082 on Build GRCh38
3067309 c.397C>T p.P133S Chr.3: 44734818-44734818 on Build GRCh38
5542271 c.461G>A p.R154K Chr.3: 44734882-44734882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769767445 Chr.3: 44735121-44735124 on Build GRCh38
rs919172687 Chr.3: 44734917-44734917 on Build GRCh38
rs753776632 Chr.3: 44735074-44735074 on Build GRCh38
rs199966866 Chr.3: 44735092-44735092 on Build GRCh38
rs780167684 Chr.3: 44734962-44734962 on Build GRCh38
rs200291719 Chr.3: 44734835-44734835 on Build GRCh38
rs375109474 Chr.3: 44734843-44734843 on Build GRCh38
rs753793868 Chr.3: 44734953-44734953 on Build GRCh38
rs201277677 Chr.3: 44735234-44735234 on Build GRCh38
rs114584722 Chr.3: 44735109-44735109 on Build GRCh38
rs199786232 Chr.3: 44735258-44735258 on Build GRCh38
rs866373213 Chr.3: 44735117-44735117 on Build GRCh38
rs58211979 Chr.3: 44734860-44734860 on Build GRCh38
rs372150222 Chr.3: 44735028-44735028 on Build GRCh38
rs772566008 Chr.3: 44735199-44735199 on Build GRCh38
rs773661097 Chr.3: 44734902-44734902 on Build GRCh38
rs748232737 Chr.3: 44734810-44734810 on Build GRCh38
rs767582266 Chr.3: 44735217-44735217 on Build GRCh38
rs770318534 Chr.3: 44734896-44734896 on Build GRCh38
rs777070729 Chr.3: 44734831-44734831 on Build GRCh38
rs747795074 Chr.3: 44735099-44735100 on Build GRCh38
rs771476887 Chr.3: 44735124-44735124 on Build GRCh38
rs998440880 Chr.3: 44735037-44735037 on Build GRCh38
rs778100714 Chr.3: 44735122-44735122 on Build GRCh38
rs889926664 Chr.3: 44735212-44735212 on Build GRCh38
rs762420869 Chr.3: 44734903-44734903 on Build GRCh38
rs747248884 Chr.3: 44734873-44734873 on Build GRCh38
rs780934418 Chr.3: 44735039-44735044 on Build GRCh38
rs865916022 Chr.3: 44734909-44734909 on Build GRCh38
rs867808087 Chr.3: 44735122-44735123 on Build GRCh38
rs533821258 Chr.3: 44734798-44734798 on Build GRCh38
rs138255914 Chr.3: 44735221-44735221 on Build GRCh38
rs762097467 Chr.3: 44734891-44734891 on Build GRCh38
rs376208947 Chr.3: 44734942-44734942 on Build GRCh38
rs375035661 Chr.3: 44734872-44734872 on Build GRCh38
rs368998322 Chr.3: 44734876-44734876 on Build GRCh38
rs202220207 Chr.3: 44735103-44735103 on Build GRCh38
rs578034408 Chr.3: 44734934-44734934 on Build GRCh38
rs752244448 Chr.3: 44735070-44735071 on Build GRCh38
rs367780010 Chr.3: 44735056-44735056 on Build GRCh38
rs756136194 Chr.3: 44734852-44734852 on Build GRCh38
rs758470817 Chr.3: 44734955-44734955 on Build GRCh38
rs749459435 Chr.3: 44735073-44735073 on Build GRCh38
rs564544043 Chr.3: 44734964-44734964 on Build GRCh38
rs745889183 Chr.3: 44734803-44734803 on Build GRCh38
rs776992185 Chr.3: 44734890-44734890 on Build GRCh38
rs150433704 Chr.3: 44734921-44734921 on Build GRCh38
rs763779707 Chr.3: 44735159-44735159 on Build GRCh38
rs772165442 Chr.3: 44734862-44734862 on Build GRCh38
rs761474570 Chr.3: 44735186-44735186 on Build GRCh38
rs750306502 Chr.3: 44735208-44735208 on Build GRCh38
rs372326306 Chr.3: 44734840-44734840 on Build GRCh38
rs781351187 Chr.3: 44734983-44734983 on Build GRCh38
rs776487643 Chr.3: 44735172-44735172 on Build GRCh38
rs764485565 Chr.3: 44734827-44734827 on Build GRCh38
rs781040517 Chr.3: 44734850-44734850 on Build GRCh38
rs200282883 Chr.3: 44735113-44735113 on Build GRCh38
rs755090894 Chr.3: 44735209-44735209 on Build GRCh38
rs754773526 Chr.3: 44734846-44734846 on Build GRCh38
rs919250056 Chr.3: 44735239-44735239 on Build GRCh38
rs142345135 Chr.3: 44735246-44735246 on Build GRCh38
rs376538464 Chr.3: 44735197-44735197 on Build GRCh38
rs774677875 Chr.3: 44735134-44735134 on Build GRCh38
rs777902839 Chr.3: 44734946-44734946 on Build GRCh38
rs752976754 Chr.3: 44735219-44735219 on Build GRCh38
rs770408994 Chr.3: 44734999-44734999 on Build GRCh38
rs369934306 Chr.3: 44735128-44735128 on Build GRCh38
rs201161138 Chr.3: 44735065-44735065 on Build GRCh38
rs765673635 Chr.3: 44734838-44734838 on Build GRCh38
rs757231292 Chr.3: 44735078-44735078 on Build GRCh38
rs760387064 Chr.3: 44735149-44735149 on Build GRCh38
rs929168781 Chr.3: 44734956-44734956 on Build GRCh38
rs752548297 Chr.3: 44734929-44734929 on Build GRCh38
rs763514876 Chr.3: 44734998-44734998 on Build GRCh38
rs772211994 Chr.3: 44734806-44734806 on Build GRCh38
rs202179449 Chr.3: 44734994-44734994 on Build GRCh38
rs140483745 Chr.3: 44734845-44734845 on Build GRCh38
rs201391564 Chr.3: 44735188-44735188 on Build GRCh38
rs201050548 Chr.3: 44735104-44735104 on Build GRCh38
rs1053286502 Chr.3: 44735043-44735043 on Build GRCh38
rs201711373 Chr.3: 44734996-44734996 on Build GRCh38
rs372723266 Chr.3: 44735080-44735080 on Build GRCh38
rs748536149 Chr.3: 44734992-44734992 on Build GRCh38
rs775346608 Chr.3: 44735146-44735146 on Build GRCh38
rs768926849 Chr.3: 44734966-44734966 on Build GRCh38
rs374876874 Chr.3: 44735052-44735052 on Build GRCh38
rs771673352 Chr.3: 44735140-44735140 on Build GRCh38
rs758906834 Chr.3: 44735030-44735030 on Build GRCh38
rs748157067 Chr.3: 44734851-44734851 on Build GRCh38
rs751239792 Chr.3: 44734912-44734912 on Build GRCh38
rs1035247305 Chr.3: 44735213-44735213 on Build GRCh38
rs767296773 Chr.3: 44735049-44735049 on Build GRCh38
rs372952554 Chr.3: 44734904-44734904 on Build GRCh38
rs764069805 Chr.3: 44735227-44735227 on Build GRCh38
rs767023875 Chr.3: 44734927-44734927 on Build GRCh38
rs368692015 Chr.3: 44734813-44734813 on Build GRCh38
rs749559913 Chr.3: 44735123-44735123 on Build GRCh38

View Full Product Details