Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52256864-52257413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR82
Entrez Gene ID
80335
Gene Name
WD repeat domain 82
Gene Aliases
MST107, MSTP107, PRO2730, PRO34047, SWD2, TMEM113, WDR82A
UniGene
Hs.194110
Gene Chromosome Location
Chr.3: 52254422-52278643 on Build GRCh38
Transcripts
NM_025222.3 XM_011534136.2

SNP Detail

SNP ID Chromosome Location
rs958120614 Chr.3: 52257280-52257280 on Build GRCh38
rs561641883 Chr.3: 52256954-52256954 on Build GRCh38
rs1028405300 Chr.3: 52257245-52257245 on Build GRCh38
rs1047060950 Chr.3: 52256897-52256897 on Build GRCh38
rs886244996 Chr.3: 52257152-52257152 on Build GRCh38
rs927673559 Chr.3: 52256945-52256945 on Build GRCh38
rs57823393 Chr.3: 52257062-52257062 on Build GRCh38
rs146545345 Chr.3: 52257208-52257208 on Build GRCh38
rs55653966 Chr.3: 52257267-52257267 on Build GRCh38
rs920224937 Chr.3: 52257370-52257370 on Build GRCh38
rs539175493 Chr.3: 52257118-52257118 on Build GRCh38
rs780053674 Chr.3: 52257215-52257216 on Build GRCh38
rs898913163 Chr.3: 52257114-52257114 on Build GRCh38
rs997247729 Chr.3: 52257213-52257213 on Build GRCh38
rs570391498 Chr.3: 52257039-52257039 on Build GRCh38
rs377579934 Chr.3: 52257190-52257190 on Build GRCh38
rs966829555 Chr.3: 52257334-52257334 on Build GRCh38
rs750905813 Chr.3: 52257181-52257181 on Build GRCh38
rs989491841 Chr.3: 52257300-52257300 on Build GRCh38
rs575385010 Chr.3: 52257371-52257371 on Build GRCh38
rs1019260796 Chr.3: 52257191-52257191 on Build GRCh38
rs529112539 Chr.3: 52256958-52256958 on Build GRCh38
rs780312530 Chr.3: 52257343-52257343 on Build GRCh38
rs994549428 Chr.3: 52257119-52257119 on Build GRCh38
rs532917834 Chr.3: 52256940-52256940 on Build GRCh38
rs552601295 Chr.3: 52257172-52257172 on Build GRCh38
rs764464555 Chr.3: 52256924-52256924 on Build GRCh38
rs145915837 Chr.3: 52257178-52257178 on Build GRCh38
rs555122744 Chr.3: 52257363-52257363 on Build GRCh38
rs982412049 Chr.3: 52257340-52257340 on Build GRCh38
rs764581752 Chr.3: 52257311-52257311 on Build GRCh38
rs34922530 Chr.3: 52257358-52257359 on Build GRCh38
rs937730350 Chr.3: 52257345-52257345 on Build GRCh38
rs751163085 Chr.3: 52257390-52257390 on Build GRCh38
rs760066127 Chr.3: 52257266-52257266 on Build GRCh38
rs562806674 Chr.3: 52257296-52257296 on Build GRCh38
rs1055201157 Chr.3: 52257360-52257360 on Build GRCh38
rs937660278 Chr.3: 52257032-52257032 on Build GRCh38
rs756001696 Chr.3: 52256943-52256943 on Build GRCh38
rs762051489 Chr.3: 52256994-52256994 on Build GRCh38
rs753411803 Chr.3: 52257127-52257127 on Build GRCh38
rs547194048 Chr.3: 52257027-52257027 on Build GRCh38

View Full Product Details