Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 53868220-53868766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR8
Entrez Gene ID
93973
Gene Name
ARP8 actin-related protein 8 homolog
Gene Aliases
ARP8, INO80N, hArp8
UniGene
Hs.412186
Gene Chromosome Location
Chr.3: 53863096-53882202 on Build GRCh38
Transcripts
NM_022899.4 XM_005265587.4 XM_011534249.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3775270 c.1852G>C p.E618Q Chr.3: 53868742-53868742 on Build GRCh38
5038016 c.1850G>C p.R617P Chr.3: 53868744-53868744 on Build GRCh38
6593141 c.955C>T p.R319C Chr.3: 53868754-53868754 on Build GRCh38
1424678 c.943C>T p.R315C Chr.3: 53868766-53868766 on Build GRCh38
1424677 c.1828C>T p.R610C Chr.3: 53868766-53868766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896007047 Chr.3: 53868421-53868421 on Build GRCh38
rs375331466 Chr.3: 53868683-53868683 on Build GRCh38
rs905942348 Chr.3: 53868344-53868344 on Build GRCh38
rs766618275 Chr.3: 53868369-53868369 on Build GRCh38
rs920515208 Chr.3: 53868584-53868584 on Build GRCh38
rs1046686 Chr.3: 53868698-53868698 on Build GRCh38
rs940586182 Chr.3: 53868301-53868301 on Build GRCh38
rs918564039 Chr.3: 53868330-53868330 on Build GRCh38
rs541015051 Chr.3: 53868541-53868541 on Build GRCh38
rs752293669 Chr.3: 53868473-53868473 on Build GRCh38
rs752272820 Chr.3: 53868725-53868725 on Build GRCh38
rs777148213 Chr.3: 53868728-53868728 on Build GRCh38
rs1057233167 Chr.3: 53868662-53868662 on Build GRCh38
rs746587919 Chr.3: 53868732-53868732 on Build GRCh38
rs1055349999 Chr.3: 53868586-53868587 on Build GRCh38
rs537688214 Chr.3: 53868340-53868340 on Build GRCh38
rs770117404 Chr.3: 53868259-53868259 on Build GRCh38
rs984115852 Chr.3: 53868650-53868650 on Build GRCh38
rs113423844 Chr.3: 53868575-53868575 on Build GRCh38
rs563160137 Chr.3: 53868697-53868697 on Build GRCh38
rs561171973 Chr.3: 53868660-53868661 on Build GRCh38
rs1057480933 Chr.3: 53868407-53868407 on Build GRCh38
rs373675886 Chr.3: 53868469-53868469 on Build GRCh38
rs976811562 Chr.3: 53868366-53868366 on Build GRCh38
rs12637539 Chr.3: 53868656-53868656 on Build GRCh38
rs764058239 Chr.3: 53868693-53868693 on Build GRCh38
rs192368042 Chr.3: 53868272-53868272 on Build GRCh38
rs753169136 Chr.3: 53868730-53868732 on Build GRCh38
rs532148065 Chr.3: 53868733-53868733 on Build GRCh38
rs548635089 Chr.3: 53868261-53868261 on Build GRCh38
rs1026652326 Chr.3: 53868273-53868273 on Build GRCh38
rs368263560 Chr.3: 53868670-53868670 on Build GRCh38
rs558590827 Chr.3: 53868512-53868512 on Build GRCh38
rs781042991 Chr.3: 53868738-53868738 on Build GRCh38
rs758113135 Chr.3: 53868727-53868727 on Build GRCh38
rs372921911 Chr.3: 53868718-53868718 on Build GRCh38
rs758998178 Chr.3: 53868365-53868365 on Build GRCh38
rs745838902 Chr.3: 53868739-53868739 on Build GRCh38
rs987262004 Chr.3: 53868534-53868534 on Build GRCh38
rs756477477 Chr.3: 53868717-53868717 on Build GRCh38
rs148353645 Chr.3: 53868323-53868323 on Build GRCh38
rs931310408 Chr.3: 53868339-53868339 on Build GRCh38
rs754118790 Chr.3: 53868657-53868657 on Build GRCh38
rs142187582 Chr.3: 53868364-53868364 on Build GRCh38
rs116180758 Chr.3: 53868320-53868320 on Build GRCh38
rs142533235 Chr.3: 53868304-53868305 on Build GRCh38
rs146364482 Chr.3: 53868439-53868439 on Build GRCh38
rs538751590 Chr.3: 53868461-53868461 on Build GRCh38
rs571971175 Chr.3: 53868540-53868540 on Build GRCh38
rs772869073 Chr.3: 53868276-53868276 on Build GRCh38
rs769527407 Chr.3: 53868745-53868745 on Build GRCh38
rs1015541367 Chr.3: 53868345-53868345 on Build GRCh38
rs760242128 Chr.3: 53868496-53868496 on Build GRCh38
rs774302529 Chr.3: 53868348-53868348 on Build GRCh38
rs774529412 Chr.3: 53868696-53868696 on Build GRCh38
rs762748263 Chr.3: 53868297-53868297 on Build GRCh38
rs921270859 Chr.3: 53868411-53868411 on Build GRCh38
rs1010879142 Chr.3: 53868424-53868424 on Build GRCh38
rs377090713 Chr.3: 53868615-53868615 on Build GRCh38
rs1028653962 Chr.3: 53868478-53868478 on Build GRCh38
rs199906479 Chr.3: 53868711-53868711 on Build GRCh38
rs1042930582 Chr.3: 53868548-53868548 on Build GRCh38
rs370679724 Chr.3: 53868678-53868678 on Build GRCh38
rs989580224 Chr.3: 53868470-53868470 on Build GRCh38

View Full Product Details