Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 108896472-108897070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDAR
Entrez Gene ID
10913
Gene Name
ectodysplasin A receptor
Gene Aliases
DL, ECTD10A, ECTD10B, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, HRM1
UniGene
Hs.171971
Gene Chromosome Location
Chr.2: 108894471-108989372 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Gene Symbol
RANBP2
Entrez Gene ID
5903
Gene Name
RAN binding protein 2
Gene Aliases
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2
UniGene
Hs.199561
Gene Chromosome Location
Chr.2: 108719446-109842301 on Build GRCh38
Transcripts
NM_022336.3 XM_006712204.1 XM_011510502.2 XM_011510503.2 XM_017004623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6153747 c.1323C>T p.A441A Chr.2: 108896931-108896931 on Build GRCh38
7260973 c.1259G>T p.R420L Chr.2: 108896995-108896995 on Build GRCh38
6192406 c.1236C>T p.L412L Chr.2: 108897018-108897018 on Build GRCh38
6200313 c.1199G>A p.R400H Chr.2: 108897055-108897055 on Build GRCh38
6153748 c.1419C>T p.A473A Chr.2: 108896931-108896931 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200198949 Chr.2: 108897045-108897045 on Build GRCh38
rs754345329 Chr.2: 108896881-108896881 on Build GRCh38
rs540186842 Chr.2: 108896696-108896696 on Build GRCh38
rs765733088 Chr.2: 108897005-108897005 on Build GRCh38
rs947430925 Chr.2: 108896533-108896546 on Build GRCh38
rs780909525 Chr.2: 108896716-108896716 on Build GRCh38
rs1043396400 Chr.2: 108896688-108896688 on Build GRCh38
rs1052320183 Chr.2: 108896725-108896725 on Build GRCh38
rs75703116 Chr.2: 108896579-108896579 on Build GRCh38
rs746416669 Chr.2: 108896936-108896936 on Build GRCh38
rs771596072 Chr.2: 108896977-108896977 on Build GRCh38
rs947851463 Chr.2: 108896670-108896670 on Build GRCh38
rs764443374 Chr.2: 108896896-108896896 on Build GRCh38
rs1008476829 Chr.2: 108896762-108896762 on Build GRCh38
rs889617202 Chr.2: 108896861-108896861 on Build GRCh38
rs1026836713 Chr.2: 108896576-108896576 on Build GRCh38
rs772690729 Chr.2: 108896982-108896982 on Build GRCh38
rs951398066 Chr.2: 108896618-108896618 on Build GRCh38
rs1055999776 Chr.2: 108896664-108896664 on Build GRCh38
rs758717240 Chr.2: 108896921-108896921 on Build GRCh38
rs746557228 Chr.2: 108896667-108896667 on Build GRCh38
rs886054744 Chr.2: 108896966-108896966 on Build GRCh38
rs928893721 Chr.2: 108896654-108896654 on Build GRCh38
rs534753987 Chr.2: 108896763-108896763 on Build GRCh38
rs897107382 Chr.2: 108896547-108896547 on Build GRCh38
rs879008158 Chr.2: 108896919-108896919 on Build GRCh38
rs982753996 Chr.2: 108896834-108896834 on Build GRCh38
rs947552141 Chr.2: 108896940-108896940 on Build GRCh38
rs368952407 Chr.2: 108896526-108896526 on Build GRCh38
rs121908453 Chr.2: 108896995-108896995 on Build GRCh38
rs781382867 Chr.2: 108896944-108896944 on Build GRCh38
rs111591018 Chr.2: 108896860-108896860 on Build GRCh38
rs1027045768 Chr.2: 108896805-108896805 on Build GRCh38
rs148961371 Chr.2: 108896653-108896653 on Build GRCh38
rs897053285 Chr.2: 108896839-108896839 on Build GRCh38
rs768401042 Chr.2: 108896719-108896719 on Build GRCh38
rs41280579 Chr.2: 108897008-108897008 on Build GRCh38
rs144996915 Chr.2: 108896945-108896945 on Build GRCh38
rs927493621 Chr.2: 108896712-108896712 on Build GRCh38
rs188257661 Chr.2: 108896838-108896838 on Build GRCh38
rs903490347 Chr.2: 108896714-108896714 on Build GRCh38
rs761459363 Chr.2: 108896963-108896963 on Build GRCh38
rs41280577 Chr.2: 108896955-108896955 on Build GRCh38
rs192939679 Chr.2: 108896916-108896916 on Build GRCh38
rs7607563 Chr.2: 108896865-108896865 on Build GRCh38
rs879053670 Chr.2: 108896619-108896619 on Build GRCh38
rs773885029 Chr.2: 108896954-108896954 on Build GRCh38
rs201228294 Chr.2: 108896503-108896504 on Build GRCh38
rs751748342 Chr.2: 108896930-108896930 on Build GRCh38
rs985570452 Chr.2: 108896590-108896590 on Build GRCh38
rs769830718 Chr.2: 108896946-108896946 on Build GRCh38
rs564195922 Chr.2: 108896949-108896949 on Build GRCh38
rs758376546 Chr.2: 108896505-108896505 on Build GRCh38
rs1018132137 Chr.2: 108896772-108896772 on Build GRCh38
rs965366133 Chr.2: 108896646-108896646 on Build GRCh38
rs748617098 Chr.2: 108896888-108896888 on Build GRCh38
rs757716764 Chr.2: 108896754-108896754 on Build GRCh38
rs368571532 Chr.2: 108896950-108896950 on Build GRCh38
rs951344216 Chr.2: 108896827-108896827 on Build GRCh38
rs199984607 Chr.2: 108896891-108896891 on Build GRCh38
rs768162724 Chr.2: 108896856-108896856 on Build GRCh38
rs748843905 Chr.2: 108896685-108896685 on Build GRCh38
rs760079482 Chr.2: 108896985-108896985 on Build GRCh38
rs995718536 Chr.2: 108896575-108896575 on Build GRCh38
rs370147384 Chr.2: 108896859-108896859 on Build GRCh38
rs559802013 Chr.2: 108896715-108896715 on Build GRCh38
rs886041005 Chr.2: 108896970-108896970 on Build GRCh38
rs757289455 Chr.2: 108896937-108896937 on Build GRCh38
rs965481198 Chr.2: 108896928-108896928 on Build GRCh38
rs938814773 Chr.2: 108896655-108896655 on Build GRCh38
rs112630241 Chr.2: 108896515-108896515 on Build GRCh38
rs1052141434 Chr.2: 108896991-108896991 on Build GRCh38
rs979302277 Chr.2: 108896710-108896710 on Build GRCh38
rs964464421 Chr.2: 108896788-108896788 on Build GRCh38
rs530945702 Chr.2: 108896898-108896898 on Build GRCh38
rs769136515 Chr.2: 108896793-108896793 on Build GRCh38
rs1019743215 Chr.2: 108896614-108896614 on Build GRCh38
rs528478080 Chr.2: 108896952-108896952 on Build GRCh38
rs777844792 Chr.2: 108896923-108896923 on Build GRCh38

View Full Product Details