Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77276358-77276880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMT2
Entrez Gene ID
29954
Gene Name
protein O-mannosyltransferase 2
Gene Aliases
LGMD2N, MDDGA2, MDDGB2, MDDGC2
UniGene
Hs.132989
Gene Chromosome Location
Chr.14: 77274956-77320885 on Build GRCh38
Transcripts
NM_013382.5 XM_011536675.2 XM_011536676.2 XM_011536677.2 XM_011536679.2 XM_017021244.1 XM_017021245.1

SNP Detail

SNP ID Chromosome Location
rs111886247 Chr.14: 77276839-77276839 on Build GRCh38
rs878922649 Chr.14: 77276703-77276703 on Build GRCh38
rs908829298 Chr.14: 77276793-77276793 on Build GRCh38
rs1012254972 Chr.14: 77276594-77276594 on Build GRCh38
rs948890154 Chr.14: 77276403-77276403 on Build GRCh38
rs886050819 Chr.14: 77276754-77276754 on Build GRCh38
rs1039223094 Chr.14: 77276487-77276487 on Build GRCh38
rs745944930 Chr.14: 77276803-77276803 on Build GRCh38
rs1045074868 Chr.14: 77276780-77276780 on Build GRCh38
rs762946655 Chr.14: 77276466-77276466 on Build GRCh38
rs546336993 Chr.14: 77276551-77276551 on Build GRCh38
rs980293755 Chr.14: 77276649-77276649 on Build GRCh38
rs956008298 Chr.14: 77276590-77276590 on Build GRCh38
rs376080296 Chr.14: 77276465-77276465 on Build GRCh38
rs886050821 Chr.14: 77276782-77276782 on Build GRCh38
rs975158823 Chr.14: 77276816-77276816 on Build GRCh38
rs752332125 Chr.14: 77276683-77276683 on Build GRCh38
rs1035134599 Chr.14: 77276567-77276567 on Build GRCh38
rs186897393 Chr.14: 77276771-77276771 on Build GRCh38
rs919029987 Chr.14: 77276688-77276688 on Build GRCh38
rs1039332345 Chr.14: 77276831-77276831 on Build GRCh38
rs750268552 Chr.14: 77276417-77276417 on Build GRCh38
rs897463477 Chr.14: 77276441-77276441 on Build GRCh38
rs141262145 Chr.14: 77276770-77276770 on Build GRCh38
rs765352349 Chr.14: 77276753-77276756 on Build GRCh38
rs371632843 Chr.14: 77276618-77276618 on Build GRCh38
rs574135706 Chr.14: 77276802-77276802 on Build GRCh38
rs559906734 Chr.14: 77276559-77276559 on Build GRCh38
rs1031233676 Chr.14: 77276679-77276679 on Build GRCh38
rs1056230440 Chr.14: 77276438-77276438 on Build GRCh38
rs1021621093 Chr.14: 77276773-77276773 on Build GRCh38
rs963658846 Chr.14: 77276702-77276702 on Build GRCh38
rs886050818 Chr.14: 77276449-77276449 on Build GRCh38
rs751053898 Chr.14: 77276530-77276530 on Build GRCh38
rs376620617 Chr.14: 77276427-77276427 on Build GRCh38
rs1044979698 Chr.14: 77276420-77276420 on Build GRCh38
rs935061893 Chr.14: 77276428-77276428 on Build GRCh38
rs928758799 Chr.14: 77276653-77276653 on Build GRCh38
rs572839285 Chr.14: 77276693-77276694 on Build GRCh38
rs550980783 Chr.14: 77276648-77276648 on Build GRCh38
rs1001183152 Chr.14: 77276786-77276786 on Build GRCh38
rs7159558 Chr.14: 77276586-77276586 on Build GRCh38
rs1056200721 Chr.14: 77276685-77276686 on Build GRCh38
rs538159298 Chr.14: 77276619-77276619 on Build GRCh38
rs942377864 Chr.14: 77276484-77276484 on Build GRCh38
rs531238117 Chr.14: 77276431-77276431 on Build GRCh38
rs45459800 Chr.14: 77276738-77276738 on Build GRCh38
rs886050820 Chr.14: 77276779-77276779 on Build GRCh38
rs1048985868 Chr.14: 77276469-77276469 on Build GRCh38
rs536738775 Chr.14: 77276740-77276740 on Build GRCh38
rs912315852 Chr.14: 77276689-77276689 on Build GRCh38
rs771371509 Chr.14: 77276778-77276778 on Build GRCh38
rs560549362 Chr.14: 77276792-77276792 on Build GRCh38
rs114926981 Chr.14: 77276781-77276781 on Build GRCh38
rs552352814 Chr.14: 77276610-77276610 on Build GRCh38
rs559126943 Chr.14: 77276776-77276776 on Build GRCh38
rs1012224040 Chr.14: 77276779-77276779 on Build GRCh38
rs181520065 Chr.14: 77276539-77276539 on Build GRCh38
rs560566070 Chr.14: 77276494-77276494 on Build GRCh38
rs914836313 Chr.14: 77276391-77276391 on Build GRCh38
rs539853899 Chr.14: 77276814-77276814 on Build GRCh38
rs756477627 Chr.14: 77276727-77276727 on Build GRCh38
rs777949514 Chr.14: 77276763-77276763 on Build GRCh38
rs376936628 Chr.14: 77276752-77276752 on Build GRCh38
rs912222368 Chr.14: 77276471-77276471 on Build GRCh38
rs900783693 Chr.14: 77276499-77276499 on Build GRCh38
rs1038951910 Chr.14: 77276710-77276710 on Build GRCh38
rs772087434 Chr.14: 77276821-77276821 on Build GRCh38
rs146570158 Chr.14: 77276677-77276677 on Build GRCh38
rs930875502 Chr.14: 77276737-77276737 on Build GRCh38
rs970087743 Chr.14: 77276774-77276774 on Build GRCh38
rs1048790445 Chr.14: 77276700-77276700 on Build GRCh38
rs150190025 Chr.14: 77276775-77276775 on Build GRCh38
rs956600436 Chr.14: 77276682-77276682 on Build GRCh38
rs1049361898 Chr.14: 77276754-77276754 on Build GRCh38
rs10141995 Chr.14: 77276789-77276789 on Build GRCh38
rs766421627 Chr.14: 77276526-77276526 on Build GRCh38

View Full Product Details