Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32643321-32643852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCG5
Entrez Gene ID
6447
Gene Name
secretogranin V
Gene Aliases
7B2, P7B2, SGNE1, SgV
UniGene
Hs.156540
Gene Chromosome Location
Chr.15: 32641669-32697097 on Build GRCh38
Transcripts
NM_001144757.1 NM_003020.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6444296 c.167C>T p.P56L Chr.15: 32643759-32643759 on Build GRCh38
1264928 c.66T>C p.T22T Chr.15: 32643658-32643658 on Build GRCh38
6008027 c.124A>T p.R42W Chr.15: 32643716-32643716 on Build GRCh38
433705 c.223G>A p.E75K Chr.15: 32643815-32643815 on Build GRCh38
4053999 c.213C>T p.P71P Chr.15: 32643805-32643805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053952906 Chr.15: 32643429-32643429 on Build GRCh38
rs747076418 Chr.15: 32643711-32643711 on Build GRCh38
rs749667874 Chr.15: 32643805-32643805 on Build GRCh38
rs753708324 Chr.15: 32643686-32643686 on Build GRCh38
rs79291170 Chr.15: 32643526-32643526 on Build GRCh38
rs971397743 Chr.15: 32643483-32643483 on Build GRCh38
rs201757263 Chr.15: 32643612-32643612 on Build GRCh38
rs753384664 Chr.15: 32643747-32643747 on Build GRCh38
rs757288577 Chr.15: 32643592-32643592 on Build GRCh38
rs939544899 Chr.15: 32643462-32643462 on Build GRCh38
rs746009181 Chr.15: 32643598-32643598 on Build GRCh38
rs1003473864 Chr.15: 32643520-32643520 on Build GRCh38
rs763651303 Chr.15: 32643683-32643683 on Build GRCh38
rs780816546 Chr.15: 32643492-32643492 on Build GRCh38
rs370104815 Chr.15: 32643559-32643559 on Build GRCh38
rs563982406 Chr.15: 32643539-32643539 on Build GRCh38
rs187406342 Chr.15: 32643664-32643664 on Build GRCh38
rs1052615122 Chr.15: 32643472-32643472 on Build GRCh38
rs946852436 Chr.15: 32643709-32643709 on Build GRCh38
rs771554856 Chr.15: 32643733-32643733 on Build GRCh38
rs1045303291 Chr.15: 32643408-32643408 on Build GRCh38
rs760233647 Chr.15: 32643739-32643739 on Build GRCh38
rs780286582 Chr.15: 32643605-32643605 on Build GRCh38
rs774978081 Chr.15: 32643811-32643811 on Build GRCh38
rs1025481670 Chr.15: 32643594-32643594 on Build GRCh38
rs763687220 Chr.15: 32643419-32643419 on Build GRCh38
rs977023045 Chr.15: 32643577-32643577 on Build GRCh38
rs754987387 Chr.15: 32643774-32643774 on Build GRCh38
rs767323267 Chr.15: 32643555-32643555 on Build GRCh38
rs764874026 Chr.15: 32643760-32643760 on Build GRCh38
rs761700525 Chr.15: 32643751-32643751 on Build GRCh38
rs200869533 Chr.15: 32643693-32643693 on Build GRCh38
rs766298083 Chr.15: 32643772-32643772 on Build GRCh38
rs553583807 Chr.15: 32643485-32643485 on Build GRCh38
rs762077268 Chr.15: 32643617-32643617 on Build GRCh38
rs780575685 Chr.15: 32643622-32643622 on Build GRCh38
rs753564941 Chr.15: 32643585-32643585 on Build GRCh38
rs541575696 Chr.15: 32643476-32643476 on Build GRCh38
rs545683183 Chr.15: 32643528-32643528 on Build GRCh38
rs199901818 Chr.15: 32643698-32643698 on Build GRCh38
rs373862155 Chr.15: 32643826-32643826 on Build GRCh38
rs900213329 Chr.15: 32643409-32643409 on Build GRCh38
rs148575688 Chr.15: 32643788-32643788 on Build GRCh38
rs532967380 Chr.15: 32643680-32643680 on Build GRCh38
rs763500021 Chr.15: 32643727-32643727 on Build GRCh38
rs751181390 Chr.15: 32643514-32643514 on Build GRCh38
rs756980769 Chr.15: 32643692-32643692 on Build GRCh38
rs371253301 Chr.15: 32643626-32643626 on Build GRCh38
rs374087954 Chr.15: 32643593-32643593 on Build GRCh38
rs950603009 Chr.15: 32643560-32643560 on Build GRCh38
rs758477629 Chr.15: 32643694-32643694 on Build GRCh38
rs781236977 Chr.15: 32643718-32643718 on Build GRCh38
rs755981497 Chr.15: 32643563-32643563 on Build GRCh38
rs772090948 Chr.15: 32643603-32643603 on Build GRCh38
rs1017953140 Chr.15: 32643781-32643781 on Build GRCh38
rs768930500 Chr.15: 32643717-32643717 on Build GRCh38
rs201908391 Chr.15: 32643540-32643540 on Build GRCh38
rs769999982 Chr.15: 32643720-32643720 on Build GRCh38
rs1038738933 Chr.15: 32643358-32643358 on Build GRCh38
rs777110263 Chr.15: 32643615-32643615 on Build GRCh38
rs758370744 Chr.15: 32643641-32643642 on Build GRCh38
rs370176611 Chr.15: 32643796-32643796 on Build GRCh38
rs759152849 Chr.15: 32643676-32643676 on Build GRCh38
rs554784380 Chr.15: 32643556-32643556 on Build GRCh38
rs779807688 Chr.15: 32643715-32643716 on Build GRCh38
rs775209230 Chr.15: 32643735-32643735 on Build GRCh38
rs543047571 Chr.15: 32643545-32643545 on Build GRCh38
rs894016240 Chr.15: 32643439-32643439 on Build GRCh38
rs948083376 Chr.15: 32643619-32643619 on Build GRCh38
rs999955377 Chr.15: 32643424-32643424 on Build GRCh38
rs944416096 Chr.15: 32643356-32643356 on Build GRCh38
rs1010216638 Chr.15: 32643473-32643473 on Build GRCh38
rs911528417 Chr.15: 32643353-32643353 on Build GRCh38
rs773779024 Chr.15: 32643721-32643721 on Build GRCh38
rs746540442 Chr.15: 32643813-32643813 on Build GRCh38
rs906780692 Chr.15: 32643455-32643455 on Build GRCh38
rs948033332 Chr.15: 32643370-32643370 on Build GRCh38
rs771607983 Chr.15: 32643806-32643806 on Build GRCh38
rs201023892 Chr.15: 32643551-32643551 on Build GRCh38
rs866939119 Chr.15: 32643740-32643740 on Build GRCh38
rs567248964 Chr.15: 32643762-32643762 on Build GRCh38
rs781495858 Chr.15: 32643775-32643775 on Build GRCh38
rs763857379 Chr.15: 32643745-32643745 on Build GRCh38
rs749616760 Chr.15: 32643650-32643650 on Build GRCh38
rs373839348 Chr.15: 32643681-32643681 on Build GRCh38
rs555464001 Chr.15: 32643719-32643719 on Build GRCh38
rs777655878 Chr.15: 32643573-32643573 on Build GRCh38
rs750372565 Chr.15: 32643761-32643761 on Build GRCh38

View Full Product Details