Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 130162967-130163562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD10
Entrez Gene ID
11211
Gene Name
frizzled class receptor 10
Gene Aliases
CD350, FZ-10, Fz10, FzE7, hFz10
UniGene
Hs.31664
Gene Chromosome Location
Chr.12: 130162459-130165740 on Build GRCh38
Transcripts
NM_007197.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3458069 c.549G>A p.K183K Chr.12: 130163491-130163491 on Build GRCh38
4647547 c.28C>T p.L10L Chr.12: 130162970-130162970 on Build GRCh38
5043849 c.203T>C p.L68S Chr.12: 130163145-130163145 on Build GRCh38
6135904 c.214G>C p.A72P Chr.12: 130163156-130163156 on Build GRCh38
4040104 c.357G>A p.P119P Chr.12: 130163299-130163299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576227403 Chr.12: 130163078-130163078 on Build GRCh38
rs752288264 Chr.12: 130163218-130163218 on Build GRCh38
rs756870051 Chr.12: 130163413-130163413 on Build GRCh38
rs765524304 Chr.12: 130163484-130163484 on Build GRCh38
rs776256274 Chr.12: 130163165-130163165 on Build GRCh38
rs749137419 Chr.12: 130163378-130163378 on Build GRCh38
rs563264151 Chr.12: 130163464-130163464 on Build GRCh38
rs765384828 Chr.12: 130163294-130163295 on Build GRCh38
rs751007002 Chr.12: 130163298-130163298 on Build GRCh38
rs200092271 Chr.12: 130163289-130163289 on Build GRCh38
rs1030145255 Chr.12: 130163338-130163338 on Build GRCh38
rs777449665 Chr.12: 130163320-130163320 on Build GRCh38
rs139041385 Chr.12: 130163193-130163193 on Build GRCh38
rs964564427 Chr.12: 130163410-130163410 on Build GRCh38
rs756854914 Chr.12: 130163330-130163330 on Build GRCh38
rs550083428 Chr.12: 130163179-130163179 on Build GRCh38
rs143887209 Chr.12: 130163404-130163404 on Build GRCh38
rs767188838 Chr.12: 130163533-130163533 on Build GRCh38
rs369253950 Chr.12: 130163212-130163212 on Build GRCh38
rs763279090 Chr.12: 130163528-130163528 on Build GRCh38
rs774773609 Chr.12: 130163390-130163390 on Build GRCh38
rs1022439632 Chr.12: 130163271-130163271 on Build GRCh38
rs202181794 Chr.12: 130163009-130163009 on Build GRCh38
rs765552086 Chr.12: 130163273-130163273 on Build GRCh38
rs1037469023 Chr.12: 130163062-130163062 on Build GRCh38
rs1030287197 Chr.12: 130163440-130163440 on Build GRCh38
rs761795300 Chr.12: 130163391-130163391 on Build GRCh38
rs767118059 Chr.12: 130163495-130163495 on Build GRCh38
rs746899085 Chr.12: 130163162-130163162 on Build GRCh38
rs200800452 Chr.12: 130163008-130163008 on Build GRCh38
rs768979118 Chr.12: 130163383-130163383 on Build GRCh38
rs772214243 Chr.12: 130163443-130163443 on Build GRCh38
rs767899069 Chr.12: 130163295-130163295 on Build GRCh38
rs768111324 Chr.12: 130163208-130163208 on Build GRCh38
rs751018133 Chr.12: 130163323-130163323 on Build GRCh38
rs757667097 Chr.12: 130163195-130163195 on Build GRCh38
rs556423688 Chr.12: 130163049-130163049 on Build GRCh38
rs939134736 Chr.12: 130163506-130163506 on Build GRCh38
rs749084019 Chr.12: 130163436-130163436 on Build GRCh38
rs771412434 Chr.12: 130163455-130163455 on Build GRCh38
rs895378064 Chr.12: 130163234-130163234 on Build GRCh38
rs755745804 Chr.12: 130163497-130163497 on Build GRCh38
rs754888654 Chr.12: 130163437-130163437 on Build GRCh38
rs745764318 Chr.12: 130163359-130163359 on Build GRCh38
rs747933777 Chr.12: 130163501-130163501 on Build GRCh38
rs527990944 Chr.12: 130162986-130162986 on Build GRCh38
rs766910008 Chr.12: 130163401-130163401 on Build GRCh38
rs777186624 Chr.12: 130163460-130163460 on Build GRCh38
rs201208311 Chr.12: 130163010-130163010 on Build GRCh38
rs780192582 Chr.12: 130163432-130163432 on Build GRCh38
rs943306926 Chr.12: 130163019-130163019 on Build GRCh38
rs374447637 Chr.12: 130163451-130163451 on Build GRCh38
rs564758683 Chr.12: 130163083-130163083 on Build GRCh38
rs752799541 Chr.12: 130163088-130163088 on Build GRCh38
rs767008901 Chr.12: 130163417-130163417 on Build GRCh38
rs766680908 Chr.12: 130163305-130163305 on Build GRCh38
rs775897205 Chr.12: 130163526-130163526 on Build GRCh38
rs748345546 Chr.12: 130163442-130163442 on Build GRCh38
rs758946231 Chr.12: 130162988-130162988 on Build GRCh38
rs369861300 Chr.12: 130163183-130163183 on Build GRCh38
rs759422064 Chr.12: 130163173-130163173 on Build GRCh38
rs1021343419 Chr.12: 130163268-130163268 on Build GRCh38
rs543498086 Chr.12: 130163439-130163439 on Build GRCh38
rs761092010 Chr.12: 130163299-130163299 on Build GRCh38
rs747697911 Chr.12: 130163029-130163029 on Build GRCh38
rs112917999 Chr.12: 130163332-130163332 on Build GRCh38
rs756092983 Chr.12: 130163427-130163427 on Build GRCh38
rs985841216 Chr.12: 130163402-130163402 on Build GRCh38
rs985505257 Chr.12: 130163015-130163015 on Build GRCh38
rs778426298 Chr.12: 130163217-130163217 on Build GRCh38
rs770750333 Chr.12: 130163254-130163254 on Build GRCh38
rs892018995 Chr.12: 130163450-130163450 on Build GRCh38
rs752049652 Chr.12: 130163115-130163115 on Build GRCh38
rs777914537 Chr.12: 130163503-130163503 on Build GRCh38
rs929032210 Chr.12: 130163096-130163096 on Build GRCh38
rs200216509 Chr.12: 130163018-130163018 on Build GRCh38
rs766029066 Chr.12: 130163061-130163061 on Build GRCh38
rs781620193 Chr.12: 130163011-130163011 on Build GRCh38
rs759948678 Chr.12: 130163406-130163406 on Build GRCh38
rs973296981 Chr.12: 130163428-130163428 on Build GRCh38
rs149428477 Chr.12: 130163281-130163281 on Build GRCh38
rs1011179139 Chr.12: 130163252-130163252 on Build GRCh38
rs199663032 Chr.12: 130163216-130163216 on Build GRCh38
rs777223482 Chr.12: 130163247-130163247 on Build GRCh38
rs555606586 Chr.12: 130163129-130163129 on Build GRCh38
rs757856444 Chr.12: 130163117-130163117 on Build GRCh38
rs201661032 Chr.12: 130163498-130163498 on Build GRCh38
rs775744371 Chr.12: 130163493-130163493 on Build GRCh38
rs754557108 Chr.12: 130163132-130163132 on Build GRCh38
rs920436507 Chr.12: 130163074-130163074 on Build GRCh38
rs781245363 Chr.12: 130163264-130163264 on Build GRCh38
rs764689557 Chr.12: 130163105-130163105 on Build GRCh38
rs771976377 Chr.12: 130163040-130163040 on Build GRCh38
rs376533946 Chr.12: 130163091-130163091 on Build GRCh38
rs749936930 Chr.12: 130163424-130163424 on Build GRCh38
rs773187403 Chr.12: 130163395-130163395 on Build GRCh38
rs769645215 Chr.12: 130163373-130163373 on Build GRCh38
rs142916136 Chr.12: 130163104-130163104 on Build GRCh38
rs773696876 Chr.12: 130163024-130163024 on Build GRCh38
rs1017978465 Chr.12: 130163530-130163530 on Build GRCh38
rs761203938 Chr.12: 130163398-130163398 on Build GRCh38
rs781365950 Chr.12: 130163121-130163121 on Build GRCh38
rs770928042 Chr.12: 130163507-130163507 on Build GRCh38
rs745641282 Chr.12: 130163272-130163272 on Build GRCh38
rs773617704 Chr.12: 130163495-130163495 on Build GRCh38
rs778729120 Chr.12: 130163499-130163499 on Build GRCh38
rs775442646 Chr.12: 130163280-130163280 on Build GRCh38
rs747106141 Chr.12: 130163449-130163449 on Build GRCh38
rs775916919 Chr.12: 130163181-130163181 on Build GRCh38
rs774544608 Chr.12: 130163291-130163291 on Build GRCh38
rs772807968 Chr.12: 130163543-130163543 on Build GRCh38
rs772122832 Chr.12: 130163392-130163392 on Build GRCh38
rs778678017 Chr.12: 130163160-130163160 on Build GRCh38
rs776701558 Chr.12: 130163511-130163511 on Build GRCh38
rs965418997 Chr.12: 130163532-130163532 on Build GRCh38
rs566016475 Chr.12: 130162992-130162992 on Build GRCh38
rs779957850 Chr.12: 130163375-130163375 on Build GRCh38
rs773192478 Chr.12: 130163048-130163048 on Build GRCh38
rs1003608704 Chr.12: 130163146-130163146 on Build GRCh38
rs763728060 Chr.12: 130163411-130163411 on Build GRCh38
rs758622904 Chr.12: 130163502-130163502 on Build GRCh38
rs758068557 Chr.12: 130163245-130163245 on Build GRCh38
rs770320717 Chr.12: 130163517-130163517 on Build GRCh38
rs765533368 Chr.12: 130163086-130163086 on Build GRCh38
rs572426561 Chr.12: 130163161-130163161 on Build GRCh38
rs762003488 Chr.12: 130163293-130163293 on Build GRCh38
rs746320895 Chr.12: 130163512-130163512 on Build GRCh38
rs769560603 Chr.12: 130163276-130163276 on Build GRCh38
rs746560003 Chr.12: 130163248-130163248 on Build GRCh38
rs778357040 Chr.12: 130163255-130163255 on Build GRCh38
rs747084549 Chr.12: 130163505-130163505 on Build GRCh38
rs750732340 Chr.12: 130163209-130163209 on Build GRCh38
rs201223026 Chr.12: 130163311-130163311 on Build GRCh38
rs10848026 Chr.12: 130163164-130163164 on Build GRCh38
rs534150403 Chr.12: 130162995-130162995 on Build GRCh38

View Full Product Details