Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30988088-30988669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD3B7
Entrez Gene ID
80270
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
Gene Aliases
CBAS1, PFIC4, SDR11E3
UniGene
Hs.460618
Gene Chromosome Location
Chr.16: 30985189-30989152 on Build GRCh38
Transcripts
NM_001142777.1 NM_001142778.1 NM_025193.3 XM_005255601.3 XM_011545960.2 XM_011545961.1 XM_011545962.2 XM_017023732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423959 c.1066C>A p.R356S Chr.16: 30988139-30988139 on Build GRCh38
3754851 c.1068T>C p.R356R Chr.16: 30988141-30988141 on Build GRCh38
4824428 c.1047G>A p.S349S Chr.16: 30988120-30988120 on Build GRCh38
1180415 c.1031A>G p.Y344C Chr.16: 30988104-30988104 on Build GRCh38
3113613 c.1061G>A p.R354Q Chr.16: 30988134-30988134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551492416 Chr.16: 30988583-30988583 on Build GRCh38
rs181123058 Chr.16: 30988536-30988536 on Build GRCh38
rs1007287217 Chr.16: 30988407-30988407 on Build GRCh38
rs958466441 Chr.16: 30988126-30988126 on Build GRCh38
rs547474493 Chr.16: 30988178-30988178 on Build GRCh38
rs757174407 Chr.16: 30988134-30988134 on Build GRCh38
rs747934703 Chr.16: 30988185-30988185 on Build GRCh38
rs762582807 Chr.16: 30988210-30988210 on Build GRCh38
rs529231958 Chr.16: 30988623-30988623 on Build GRCh38
rs201485331 Chr.16: 30988128-30988128 on Build GRCh38
rs747643646 Chr.16: 30988348-30988348 on Build GRCh38
rs773106348 Chr.16: 30988191-30988191 on Build GRCh38
rs915843122 Chr.16: 30988530-30988530 on Build GRCh38
rs990752936 Chr.16: 30988525-30988525 on Build GRCh38
rs765855524 Chr.16: 30988140-30988140 on Build GRCh38
rs780273226 Chr.16: 30988173-30988173 on Build GRCh38
rs1035029407 Chr.16: 30988503-30988505 on Build GRCh38
rs761402161 Chr.16: 30988194-30988194 on Build GRCh38
rs982168766 Chr.16: 30988496-30988497 on Build GRCh38
rs894042520 Chr.16: 30988409-30988409 on Build GRCh38
rs939803002 Chr.16: 30988558-30988558 on Build GRCh38
rs948744690 Chr.16: 30988569-30988569 on Build GRCh38
rs1037154088 Chr.16: 30988595-30988595 on Build GRCh38
rs769439604 Chr.16: 30988133-30988133 on Build GRCh38
rs145747267 Chr.16: 30988168-30988168 on Build GRCh38
rs768208043 Chr.16: 30988306-30988306 on Build GRCh38
rs376693465 Chr.16: 30988234-30988234 on Build GRCh38
rs560267801 Chr.16: 30988479-30988479 on Build GRCh38
rs762631905 Chr.16: 30988138-30988138 on Build GRCh38
rs141997902 Chr.16: 30988163-30988163 on Build GRCh38
rs934586416 Chr.16: 30988227-30988227 on Build GRCh38
rs948795612 Chr.16: 30988240-30988240 on Build GRCh38
rs367802544 Chr.16: 30988206-30988206 on Build GRCh38
rs547418000 Chr.16: 30988636-30988636 on Build GRCh38
rs533625860 Chr.16: 30988481-30988481 on Build GRCh38
rs1029003747 Chr.16: 30988406-30988406 on Build GRCh38
rs776417391 Chr.16: 30988124-30988124 on Build GRCh38
rs536762662 Chr.16: 30988505-30988505 on Build GRCh38
rs1011515769 Chr.16: 30988444-30988444 on Build GRCh38
rs996194796 Chr.16: 30988333-30988333 on Build GRCh38
rs368619380 Chr.16: 30988175-30988175 on Build GRCh38
rs990477806 Chr.16: 30988539-30988539 on Build GRCh38
rs574482852 Chr.16: 30988360-30988360 on Build GRCh38
rs1021120349 Chr.16: 30988471-30988471 on Build GRCh38
rs932332615 Chr.16: 30988621-30988621 on Build GRCh38
rs897337574 Chr.16: 30988616-30988616 on Build GRCh38
rs1027288247 Chr.16: 30988347-30988347 on Build GRCh38
rs914558334 Chr.16: 30988543-30988543 on Build GRCh38
rs140059922 Chr.16: 30988380-30988380 on Build GRCh38
rs577031784 Chr.16: 30988282-30988282 on Build GRCh38
rs397514442 Chr.16: 30988112-30988113 on Build GRCh38
rs907353521 Chr.16: 30988254-30988254 on Build GRCh38
rs746926891 Chr.16: 30988120-30988120 on Build GRCh38
rs773863220 Chr.16: 30988310-30988310 on Build GRCh38
rs564710803 Chr.16: 30988507-30988507 on Build GRCh38
rs151000245 Chr.16: 30988162-30988162 on Build GRCh38
rs897410739 Chr.16: 30988319-30988319 on Build GRCh38
rs965813116 Chr.16: 30988482-30988482 on Build GRCh38
rs562082021 Chr.16: 30988622-30988622 on Build GRCh38
rs915806796 Chr.16: 30988185-30988188 on Build GRCh38
rs761166824 Chr.16: 30988161-30988161 on Build GRCh38
rs758419723 Chr.16: 30988580-30988580 on Build GRCh38
rs781013516 Chr.16: 30988184-30988184 on Build GRCh38
rs778853666 Chr.16: 30988253-30988253 on Build GRCh38
rs34212827 Chr.16: 30988113-30988113 on Build GRCh38
rs769444237 Chr.16: 30988186-30988186 on Build GRCh38
rs959384495 Chr.16: 30988523-30988523 on Build GRCh38
rs372707052 Chr.16: 30988214-30988214 on Build GRCh38
rs757589014 Chr.16: 30988167-30988167 on Build GRCh38
rs147810715 Chr.16: 30988170-30988170 on Build GRCh38
rs980103106 Chr.16: 30988577-30988577 on Build GRCh38
rs144297694 Chr.16: 30988235-30988235 on Build GRCh38
rs572127353 Chr.16: 30988502-30988502 on Build GRCh38
rs2305880 Chr.16: 30988141-30988141 on Build GRCh38
rs556149521 Chr.16: 30988332-30988332 on Build GRCh38
rs538152984 Chr.16: 30988314-30988314 on Build GRCh38

View Full Product Details