Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9576448-9576882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP52
Entrez Gene ID
146845
Gene Name
cilia and flagella associated protein 52
Gene Aliases
WDR16, WDRPUH
UniGene
Hs.232270
Gene Chromosome Location
Chr.17: 9576627-9643459 on Build GRCh38
Transcripts
NM_001080556.1 NM_145054.4 XM_017024227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
708486 c.69T>G p.N23K Chr.17: 9576764-9576764 on Build GRCh38
4820078 c.60C>G p.I20M Chr.17: 9576755-9576755 on Build GRCh38
2882814 c.67A>G p.N23D Chr.17: 9576762-9576762 on Build GRCh38
6821560 c.26C>T p.A9V Chr.17: 9576721-9576721 on Build GRCh38
986418 c.17C>T p.S6L Chr.17: 9576712-9576712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745586152 Chr.17: 9576673-9576673 on Build GRCh38
rs924811129 Chr.17: 9576664-9576664 on Build GRCh38
rs773506070 Chr.17: 9576468-9576468 on Build GRCh38
rs566805732 Chr.17: 9576546-9576546 on Build GRCh38
rs34406554 Chr.17: 9576848-9576849 on Build GRCh38
rs1038701909 Chr.17: 9576541-9576541 on Build GRCh38
rs372640682 Chr.17: 9576734-9576734 on Build GRCh38
rs764091622 Chr.17: 9576725-9576725 on Build GRCh38
rs924700272 Chr.17: 9576497-9576497 on Build GRCh38
rs771596812 Chr.17: 9576681-9576681 on Build GRCh38
rs545226350 Chr.17: 9576823-9576823 on Build GRCh38
rs557961379 Chr.17: 9576492-9576492 on Build GRCh38
rs773119476 Chr.17: 9576713-9576713 on Build GRCh38
rs1037672880 Chr.17: 9576859-9576859 on Build GRCh38
rs1056259313 Chr.17: 9576618-9576618 on Build GRCh38
rs372786040 Chr.17: 9576646-9576646 on Build GRCh38
rs746523607 Chr.17: 9576695-9576695 on Build GRCh38
rs756159401 Chr.17: 9576723-9576723 on Build GRCh38
rs78608879 Chr.17: 9576490-9576490 on Build GRCh38
rs551772760 Chr.17: 9576594-9576594 on Build GRCh38
rs754404537 Chr.17: 9576731-9576731 on Build GRCh38
rs959006063 Chr.17: 9576540-9576540 on Build GRCh38
rs369821312 Chr.17: 9576645-9576645 on Build GRCh38
rs978842894 Chr.17: 9576496-9576496 on Build GRCh38
rs112474080 Chr.17: 9576490-9576491 on Build GRCh38
rs879715624 Chr.17: 9576624-9576624 on Build GRCh38
rs544154897 Chr.17: 9576709-9576709 on Build GRCh38
rs776716850 Chr.17: 9576700-9576700 on Build GRCh38
rs746253693 Chr.17: 9576736-9576736 on Build GRCh38
rs987679427 Chr.17: 9576510-9576510 on Build GRCh38
rs16958487 Chr.17: 9576560-9576560 on Build GRCh38
rs761836671 Chr.17: 9576704-9576704 on Build GRCh38
rs531439145 Chr.17: 9576491-9576491 on Build GRCh38
rs771302579 Chr.17: 9576570-9576570 on Build GRCh38
rs1022649622 Chr.17: 9576649-9576649 on Build GRCh38
rs143563239 Chr.17: 9576788-9576788 on Build GRCh38
rs375706369 Chr.17: 9576744-9576744 on Build GRCh38
rs370915625 Chr.17: 9576756-9576756 on Build GRCh38
rs751958256 Chr.17: 9576744-9576746 on Build GRCh38
rs140009881 Chr.17: 9576715-9576715 on Build GRCh38
rs768349856 Chr.17: 9576762-9576762 on Build GRCh38
rs930317908 Chr.17: 9576566-9576566 on Build GRCh38
rs376223932 Chr.17: 9576699-9576699 on Build GRCh38
rs374591347 Chr.17: 9576499-9576499 on Build GRCh38
rs776266650 Chr.17: 9576764-9576764 on Build GRCh38
rs1003472499 Chr.17: 9576512-9576512 on Build GRCh38
rs375846701 Chr.17: 9576556-9576556 on Build GRCh38
rs200153894 Chr.17: 9576721-9576721 on Build GRCh38
rs1034049166 Chr.17: 9576539-9576539 on Build GRCh38
rs761248950 Chr.17: 9576531-9576531 on Build GRCh38
rs778270536 Chr.17: 9576658-9576658 on Build GRCh38
rs370454094 Chr.17: 9576666-9576666 on Build GRCh38
rs769822678 Chr.17: 9576816-9576816 on Build GRCh38
rs779586425 Chr.17: 9576749-9576749 on Build GRCh38
rs746608233 Chr.17: 9576751-9576751 on Build GRCh38
rs758741653 Chr.17: 9576738-9576738 on Build GRCh38
rs974962669 Chr.17: 9576818-9576818 on Build GRCh38
rs150592698 Chr.17: 9576596-9576596 on Build GRCh38
rs775171372 Chr.17: 9576694-9576694 on Build GRCh38
rs533317760 Chr.17: 9576593-9576593 on Build GRCh38
rs943632067 Chr.17: 9576537-9576537 on Build GRCh38

View Full Product Details