Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103586990-103587530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCEAL4
Entrez Gene ID
79921
Gene Name
transcription elongation factor A like 4
Gene Aliases
NPD017, WEX7
UniGene
Hs.194329
Gene Chromosome Location
Chr.X: 103576231-103587736 on Build GRCh38
Transcripts
NM_001006935.2 NM_001006937.2 NM_001300901.1 NM_001305840.1 NM_001305841.1 NM_001305842.1 NM_024863.5 XM_017029856.1 XM_017029857.1

SNP Detail

SNP ID Chromosome Location
rs549909784 Chr.X: 103587343-103587343 on Build GRCh38
rs774690181 Chr.X: 103587067-103587067 on Build GRCh38
rs377158977 Chr.X: 103587248-103587248 on Build GRCh38
rs1028331229 Chr.X: 103587389-103587389 on Build GRCh38
rs780165569 Chr.X: 103587268-103587268 on Build GRCh38
rs764772966 Chr.X: 103587115-103587115 on Build GRCh38
rs11010 Chr.X: 103587113-103587113 on Build GRCh38
rs766030159 Chr.X: 103587081-103587081 on Build GRCh38
rs201993261 Chr.X: 103587301-103587301 on Build GRCh38
rs773569149 Chr.X: 103587361-103587361 on Build GRCh38
rs776391685 Chr.X: 103587317-103587317 on Build GRCh38
rs752626147 Chr.X: 103587158-103587158 on Build GRCh38
rs769153116 Chr.X: 103587295-103587295 on Build GRCh38
rs764151895 Chr.X: 103587366-103587366 on Build GRCh38
rs767622680 Chr.X: 103587481-103587481 on Build GRCh38
rs777788429 Chr.X: 103587171-103587171 on Build GRCh38
rs756177089 Chr.X: 103587130-103587130 on Build GRCh38
rs367788883 Chr.X: 103587085-103587085 on Build GRCh38
rs17850896 Chr.X: 103587108-103587108 on Build GRCh38
rs758406612 Chr.X: 103587166-103587166 on Build GRCh38
rs773501291 Chr.X: 103587196-103587197 on Build GRCh38
rs763432036 Chr.X: 103587064-103587064 on Build GRCh38
rs775905970 Chr.X: 103587044-103587044 on Build GRCh38
rs199974804 Chr.X: 103587053-103587053 on Build GRCh38
rs368477560 Chr.X: 103587290-103587290 on Build GRCh38
rs772148763 Chr.X: 103587184-103587185 on Build GRCh38
rs751414028 Chr.X: 103587214-103587214 on Build GRCh38
rs749856659 Chr.X: 103587285-103587285 on Build GRCh38
rs771285046 Chr.X: 103587035-103587035 on Build GRCh38
rs111976427 Chr.X: 103587204-103587204 on Build GRCh38
rs769063260 Chr.X: 103587065-103587065 on Build GRCh38
rs769880298 Chr.X: 103587041-103587041 on Build GRCh38
rs531689402 Chr.X: 103587342-103587342 on Build GRCh38
rs375153902 Chr.X: 103587227-103587227 on Build GRCh38
rs778993256 Chr.X: 103587025-103587025 on Build GRCh38
rs758997247 Chr.X: 103587114-103587114 on Build GRCh38
rs781418427 Chr.X: 103587036-103587036 on Build GRCh38
rs779249856 Chr.X: 103587340-103587340 on Build GRCh38
rs759710510 Chr.X: 103587476-103587476 on Build GRCh38
rs762097073 Chr.X: 103587068-103587068 on Build GRCh38
rs954037550 Chr.X: 103587403-103587403 on Build GRCh38
rs766893431 Chr.X: 103587382-103587382 on Build GRCh38
rs766851210 Chr.X: 103587364-103587365 on Build GRCh38
rs1015763397 Chr.X: 103587499-103587500 on Build GRCh38
rs374709861 Chr.X: 103587015-103587015 on Build GRCh38
rs373009602 Chr.X: 103587242-103587242 on Build GRCh38
rs752260147 Chr.X: 103587362-103587362 on Build GRCh38
rs221933 Chr.X: 103587239-103587239 on Build GRCh38
rs763393492 Chr.X: 103587372-103587376 on Build GRCh38
rs751562599 Chr.X: 103587456-103587456 on Build GRCh38
rs745960691 Chr.X: 103587039-103587039 on Build GRCh38
rs202054247 Chr.X: 103587300-103587300 on Build GRCh38
rs761059818 Chr.X: 103587202-103587203 on Build GRCh38
rs17850895 Chr.X: 103587071-103587071 on Build GRCh38

View Full Product Details