Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67924034-67924397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS11A
Entrez Gene ID
339967
Gene Name
transmembrane protease, serine 11A
Gene Aliases
ECRG1
UniGene
Hs.450110
Gene Chromosome Location
Chr.4: 67910301-67963514 on Build GRCh38
Transcripts
NM_001114387.1 NM_182606.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7320951 c.509G>A p.G170E Chr.4: 67924148-67924148 on Build GRCh38
6846666 c.495G>A p.M165I Chr.4: 67924162-67924162 on Build GRCh38
126978 c.511G>T p.E171* Chr.4: 67924146-67924146 on Build GRCh38
1694438 c.523C>T p.Q175* Chr.4: 67924134-67924134 on Build GRCh38
5819385 c.508G>T p.G170W Chr.4: 67924149-67924149 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754035699 Chr.4: 67924139-67924139 on Build GRCh38
rs566293897 Chr.4: 67924243-67924243 on Build GRCh38
rs755777467 Chr.4: 67924122-67924125 on Build GRCh38
rs975830723 Chr.4: 67924073-67924073 on Build GRCh38
rs755335925 Chr.4: 67924143-67924143 on Build GRCh38
rs373447572 Chr.4: 67924135-67924135 on Build GRCh38
rs898256537 Chr.4: 67924228-67924228 on Build GRCh38
rs770552818 Chr.4: 67924095-67924095 on Build GRCh38
rs369884437 Chr.4: 67924115-67924115 on Build GRCh38
rs552432223 Chr.4: 67924318-67924318 on Build GRCh38
rs764974472 Chr.4: 67924111-67924111 on Build GRCh38
rs6839279 Chr.4: 67924060-67924060 on Build GRCh38
rs565352774 Chr.4: 67924340-67924340 on Build GRCh38
rs1031560485 Chr.4: 67924373-67924373 on Build GRCh38
rs547831857 Chr.4: 67924372-67924372 on Build GRCh38
rs940179687 Chr.4: 67924121-67924121 on Build GRCh38
rs748647985 Chr.4: 67924149-67924149 on Build GRCh38
rs747068696 Chr.4: 67924300-67924300 on Build GRCh38
rs563617746 Chr.4: 67924258-67924258 on Build GRCh38
rs920943225 Chr.4: 67924252-67924252 on Build GRCh38
rs144009146 Chr.4: 67924148-67924148 on Build GRCh38
rs370139605 Chr.4: 67924191-67924191 on Build GRCh38
rs776163752 Chr.4: 67924099-67924099 on Build GRCh38
rs921302381 Chr.4: 67924079-67924079 on Build GRCh38
rs762926876 Chr.4: 67924122-67924122 on Build GRCh38
rs376811083 Chr.4: 67924106-67924106 on Build GRCh38
rs186038072 Chr.4: 67924081-67924081 on Build GRCh38
rs200746363 Chr.4: 67924188-67924188 on Build GRCh38
rs767578490 Chr.4: 67924130-67924130 on Build GRCh38
rs766573543 Chr.4: 67924137-67924137 on Build GRCh38
rs909355646 Chr.4: 67924317-67924317 on Build GRCh38
rs28434494 Chr.4: 67924348-67924348 on Build GRCh38
rs772919150 Chr.4: 67924089-67924089 on Build GRCh38
rs201795879 Chr.4: 67924198-67924198 on Build GRCh38
rs142521133 Chr.4: 67924299-67924299 on Build GRCh38
rs965291787 Chr.4: 67924152-67924152 on Build GRCh38
rs987087393 Chr.4: 67924093-67924093 on Build GRCh38
rs975041305 Chr.4: 67924227-67924227 on Build GRCh38
rs527801981 Chr.4: 67924364-67924364 on Build GRCh38
rs777356354 Chr.4: 67924207-67924208 on Build GRCh38
rs746659046 Chr.4: 67924090-67924090 on Build GRCh38

View Full Product Details