Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 139454074-139454576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB33B
Entrez Gene ID
83452
Gene Name
RAB33B, member RAS oncogene family
Gene Aliases
SMC2
UniGene
Hs.591679
Gene Chromosome Location
Chr.4: 139453232-139475916 on Build GRCh38
Transcripts
NM_031296.2 XM_011532299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7003285 c.83C>A p.P28H Chr.4: 139454278-139454278 on Build GRCh38
4719800 c.64G>A p.A22T Chr.4: 139454259-139454259 on Build GRCh38
7082468 c.45C>T p.S15S Chr.4: 139454240-139454240 on Build GRCh38
7144366 c.102C>T p.F34F Chr.4: 139454297-139454297 on Build GRCh38
1051388 c.53C>T p.A18V Chr.4: 139454248-139454248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771215024 Chr.4: 139454161-139454161 on Build GRCh38
rs1038769242 Chr.4: 139454477-139454477 on Build GRCh38
rs746037476 Chr.4: 139454356-139454356 on Build GRCh38
rs913370870 Chr.4: 139454144-139454144 on Build GRCh38
rs745958807 Chr.4: 139454441-139454441 on Build GRCh38
rs772457148 Chr.4: 139454358-139454358 on Build GRCh38
rs202128260 Chr.4: 139454426-139454426 on Build GRCh38
rs747936685 Chr.4: 139454360-139454360 on Build GRCh38
rs779011534 Chr.4: 139454440-139454440 on Build GRCh38
rs954788812 Chr.4: 139454534-139454534 on Build GRCh38
rs147523243 Chr.4: 139454291-139454291 on Build GRCh38
rs368228699 Chr.4: 139454225-139454225 on Build GRCh38
rs751831855 Chr.4: 139454204-139454209 on Build GRCh38
rs764433053 Chr.4: 139454418-139454418 on Build GRCh38
rs769703005 Chr.4: 139454455-139454455 on Build GRCh38
rs201382820 Chr.4: 139454261-139454261 on Build GRCh38
rs587776958 Chr.4: 139454331-139454331 on Build GRCh38
rs765290544 Chr.4: 139454275-139454275 on Build GRCh38
rs201281099 Chr.4: 139454417-139454417 on Build GRCh38
rs766613518 Chr.4: 139454177-139454177 on Build GRCh38
rs979821067 Chr.4: 139454102-139454102 on Build GRCh38
rs764775221 Chr.4: 139454193-139454193 on Build GRCh38
rs759584805 Chr.4: 139454475-139454476 on Build GRCh38
rs886674153 Chr.4: 139454431-139454431 on Build GRCh38
rs1011302437 Chr.4: 139454156-139454156 on Build GRCh38
rs757940962 Chr.4: 139454206-139454206 on Build GRCh38
rs774601028 Chr.4: 139454252-139454252 on Build GRCh38
rs760496697 Chr.4: 139454165-139454165 on Build GRCh38
rs563342565 Chr.4: 139454141-139454141 on Build GRCh38
rs911276815 Chr.4: 139454436-139454436 on Build GRCh38
rs772891750 Chr.4: 139454463-139454463 on Build GRCh38
rs777789062 Chr.4: 139454237-139454237 on Build GRCh38
rs551015649 Chr.4: 139454099-139454099 on Build GRCh38
rs926179435 Chr.4: 139454131-139454131 on Build GRCh38
rs776574187 Chr.4: 139454265-139454265 on Build GRCh38
rs762852071 Chr.4: 139454370-139454370 on Build GRCh38
rs748718122 Chr.4: 139454148-139454148 on Build GRCh38
rs921956920 Chr.4: 139454094-139454094 on Build GRCh38
rs759147084 Chr.4: 139454395-139454395 on Build GRCh38
rs757709966 Chr.4: 139454437-139454437 on Build GRCh38
rs182647425 Chr.4: 139454160-139454160 on Build GRCh38
rs747305009 Chr.4: 139454454-139454454 on Build GRCh38
rs1035382343 Chr.4: 139454412-139454412 on Build GRCh38
rs773859206 Chr.4: 139454493-139454493 on Build GRCh38
rs761857133 Chr.4: 139454192-139454192 on Build GRCh38
rs905629538 Chr.4: 139454366-139454366 on Build GRCh38
rs149649513 Chr.4: 139454361-139454361 on Build GRCh38
rs752394542 Chr.4: 139454405-139454405 on Build GRCh38
rs749986787 Chr.4: 139454201-139454201 on Build GRCh38
rs753814273 Chr.4: 139454190-139454190 on Build GRCh38
rs368860024 Chr.4: 139454448-139454448 on Build GRCh38
rs767052914 Chr.4: 139454495-139454495 on Build GRCh38
rs776521254 Chr.4: 139454359-139454359 on Build GRCh38
rs553383693 Chr.4: 139454506-139454506 on Build GRCh38
rs765854412 Chr.4: 139454381-139454381 on Build GRCh38
rs769846792 Chr.4: 139454154-139454154 on Build GRCh38
rs773200487 Chr.4: 139454365-139454365 on Build GRCh38
rs777094745 Chr.4: 139454146-139454146 on Build GRCh38
rs917025427 Chr.4: 139454096-139454096 on Build GRCh38
rs1051104549 Chr.4: 139454153-139454153 on Build GRCh38
rs187072518 Chr.4: 139454469-139454469 on Build GRCh38
rs886251366 Chr.4: 139454209-139454209 on Build GRCh38
rs936307967 Chr.4: 139454140-139454140 on Build GRCh38
rs748593700 Chr.4: 139454223-139454223 on Build GRCh38
rs753943143 Chr.4: 139454340-139454340 on Build GRCh38
rs367812678 Chr.4: 139454297-139454297 on Build GRCh38
rs13128486 Chr.4: 139454142-139454142 on Build GRCh38
rs1052970617 Chr.4: 139454112-139454112 on Build GRCh38
rs371561776 Chr.4: 139454323-139454323 on Build GRCh38
rs778229509 Chr.4: 139454324-139454324 on Build GRCh38
rs753089290 Chr.4: 139454300-139454300 on Build GRCh38
rs974858262 Chr.4: 139454232-139454232 on Build GRCh38
rs921925813 Chr.4: 139454245-139454245 on Build GRCh38
rs375211226 Chr.4: 139454222-139454222 on Build GRCh38
rs867952323 Chr.4: 139454377-139454377 on Build GRCh38
rs138534367 Chr.4: 139454330-139454330 on Build GRCh38
rs980175514 Chr.4: 139454322-139454322 on Build GRCh38
rs568847426 Chr.4: 139454528-139454528 on Build GRCh38
rs755362234 Chr.4: 139454221-139454221 on Build GRCh38
rs377532895 Chr.4: 139454215-139454215 on Build GRCh38
rs759734964 Chr.4: 139454259-139454259 on Build GRCh38
rs773290825 Chr.4: 139454155-139454155 on Build GRCh38
rs974910643 Chr.4: 139454522-139454522 on Build GRCh38
rs780395539 Chr.4: 139454452-139454452 on Build GRCh38
rs755124749 Chr.4: 139454385-139454385 on Build GRCh38
rs370813224 Chr.4: 139454246-139454246 on Build GRCh38
rs576086336 Chr.4: 139454553-139454553 on Build GRCh38
rs1038394433 Chr.4: 139454172-139454172 on Build GRCh38
rs764020976 Chr.4: 139454168-139454168 on Build GRCh38
rs761803997 Chr.4: 139454270-139454270 on Build GRCh38
rs375245585 Chr.4: 139454282-139454282 on Build GRCh38
rs568007401 Chr.4: 139454396-139454396 on Build GRCh38
rs1047651350 Chr.4: 139454523-139454523 on Build GRCh38
rs770801753 Chr.4: 139454478-139454478 on Build GRCh38
rs770964994 Chr.4: 139454247-139454247 on Build GRCh38
rs529385824 Chr.4: 139454095-139454095 on Build GRCh38
rs766581102 Chr.4: 139454292-139454292 on Build GRCh38
rs370996949 Chr.4: 139454219-139454219 on Build GRCh38
rs771039039 Chr.4: 139454336-139454336 on Build GRCh38
rs1035871076 Chr.4: 139454194-139454194 on Build GRCh38
rs898461275 Chr.4: 139454485-139454485 on Build GRCh38
rs762497408 Chr.4: 139454283-139454283 on Build GRCh38

View Full Product Details