Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 90622490-90623089 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
UniGene
Hs.591777
Gene Chromosome Location
Chr.5: 90558681-91164226 on Build GRCh38
Transcripts
NM_032119.3 NR_003149.1 XM_017009963.1 XM_017009964.1 XM_017009965.1 XM_017009966.1 XM_017009967.1 XM_017009968.1 XM_017009969.1 XM_017009970.1 XM_017009971.1 XM_017009974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399060 c.550A>G p.T184A Chr.5: 90622693-90622693 on Build GRCh38
738967 c.525G>T p.K175N Chr.5: 90622668-90622668 on Build GRCh38
1071111 c.465C>T p.I155I Chr.5: 90622608-90622608 on Build GRCh38
3618531 c.535G>A p.G179R Chr.5: 90622678-90622678 on Build GRCh38
5591153 c.488G>A p.R163K Chr.5: 90622631-90622631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185757607 Chr.5: 90622764-90622764 on Build GRCh38
rs6891275 Chr.5: 90622887-90622887 on Build GRCh38
rs932331675 Chr.5: 90622540-90622540 on Build GRCh38
rs199840088 Chr.5: 90622823-90622824 on Build GRCh38
rs981191086 Chr.5: 90622656-90622656 on Build GRCh38
rs201758835 Chr.5: 90622643-90622643 on Build GRCh38
rs895855635 Chr.5: 90622816-90622816 on Build GRCh38
rs780134978 Chr.5: 90622612-90622612 on Build GRCh38
rs372450736 Chr.5: 90622562-90622562 on Build GRCh38
rs774679925 Chr.5: 90622817-90622817 on Build GRCh38
rs1007982362 Chr.5: 90622549-90622549 on Build GRCh38
rs13162720 Chr.5: 90622805-90622805 on Build GRCh38
rs1002170513 Chr.5: 90623009-90623009 on Build GRCh38
rs1013831026 Chr.5: 90623059-90623059 on Build GRCh38
rs777700739 Chr.5: 90622749-90622751 on Build GRCh38
rs201180985 Chr.5: 90622609-90622609 on Build GRCh38
rs6891287 Chr.5: 90622908-90622908 on Build GRCh38
rs539443837 Chr.5: 90622727-90622727 on Build GRCh38
rs769036174 Chr.5: 90622820-90622820 on Build GRCh38
rs73181620 Chr.5: 90622541-90622541 on Build GRCh38
rs777780563 Chr.5: 90622593-90622593 on Build GRCh38
rs952062982 Chr.5: 90622648-90622648 on Build GRCh38
rs568691248 Chr.5: 90622546-90622546 on Build GRCh38
rs560104130 Chr.5: 90622560-90622560 on Build GRCh38
rs571045299 Chr.5: 90623049-90623049 on Build GRCh38
rs189534872 Chr.5: 90622804-90622804 on Build GRCh38
rs879298294 Chr.5: 90622645-90622645 on Build GRCh38
rs756400005 Chr.5: 90622804-90622824 on Build GRCh38
rs1014747239 Chr.5: 90622834-90622834 on Build GRCh38
rs772421314 Chr.5: 90622652-90622652 on Build GRCh38
rs755990677 Chr.5: 90622742-90622742 on Build GRCh38
rs1033676600 Chr.5: 90623018-90623018 on Build GRCh38
rs753440266 Chr.5: 90623051-90623051 on Build GRCh38
rs115936618 Chr.5: 90622516-90622516 on Build GRCh38
rs199873924 Chr.5: 90622606-90622606 on Build GRCh38
rs534742658 Chr.5: 90622529-90622529 on Build GRCh38
rs754960654 Chr.5: 90622683-90622683 on Build GRCh38
rs932307241 Chr.5: 90622964-90622964 on Build GRCh38
rs1014386176 Chr.5: 90622545-90622545 on Build GRCh38
rs552930944 Chr.5: 90623063-90623063 on Build GRCh38
rs368285759 Chr.5: 90622561-90622561 on Build GRCh38
rs752430578 Chr.5: 90622569-90622569 on Build GRCh38
rs911549562 Chr.5: 90622956-90622956 on Build GRCh38
rs543662961 Chr.5: 90622865-90622865 on Build GRCh38
rs1049113653 Chr.5: 90622995-90622995 on Build GRCh38
rs764343409 Chr.5: 90622815-90622815 on Build GRCh38
rs943957048 Chr.5: 90622983-90622983 on Build GRCh38
rs905131099 Chr.5: 90622800-90622800 on Build GRCh38
rs758259022 Chr.5: 90622573-90622573 on Build GRCh38
rs537249795 Chr.5: 90623016-90623016 on Build GRCh38
rs1000385189 Chr.5: 90622896-90622896 on Build GRCh38
rs111696128 Chr.5: 90622967-90622967 on Build GRCh38
rs754607038 Chr.5: 90622558-90622558 on Build GRCh38
rs768822378 Chr.5: 90622620-90622620 on Build GRCh38
rs576057560 Chr.5: 90622804-90622811 on Build GRCh38
rs897035152 Chr.5: 90622608-90622608 on Build GRCh38
rs150089006 Chr.5: 90622988-90622988 on Build GRCh38
rs1033235901 Chr.5: 90622905-90622905 on Build GRCh38
rs16868864 Chr.5: 90622517-90622517 on Build GRCh38
rs374917122 Chr.5: 90622687-90622687 on Build GRCh38
rs911611736 Chr.5: 90622642-90622642 on Build GRCh38
rs1048941209 Chr.5: 90622796-90622796 on Build GRCh38
rs761111332 Chr.5: 90622678-90622678 on Build GRCh38
rs774414461 Chr.5: 90622623-90622623 on Build GRCh38
rs926218257 Chr.5: 90622594-90622594 on Build GRCh38
rs572988724 Chr.5: 90622803-90622803 on Build GRCh38
rs766815254 Chr.5: 90622706-90622706 on Build GRCh38
rs574309591 Chr.5: 90622835-90622835 on Build GRCh38
rs979186434 Chr.5: 90622894-90622894 on Build GRCh38
rs758933880 Chr.5: 90622741-90622741 on Build GRCh38
rs767517901 Chr.5: 90622726-90622727 on Build GRCh38
rs191082699 Chr.5: 90623060-90623060 on Build GRCh38
rs6870337 Chr.5: 90622996-90622996 on Build GRCh38
rs752540803 Chr.5: 90622732-90622737 on Build GRCh38
rs759460034 Chr.5: 90623020-90623020 on Build GRCh38
rs749359697 Chr.5: 90622614-90622614 on Build GRCh38
rs551198938 Chr.5: 90623040-90623040 on Build GRCh38
rs986417786 Chr.5: 90622925-90622925 on Build GRCh38
rs140696061 Chr.5: 90622987-90622987 on Build GRCh38
rs960731160 Chr.5: 90623023-90623023 on Build GRCh38
rs186178552 Chr.5: 90622984-90622984 on Build GRCh38
rs773569257 Chr.5: 90622676-90622676 on Build GRCh38
rs912441334 Chr.5: 90622974-90622974 on Build GRCh38
rs181767947 Chr.5: 90622814-90622814 on Build GRCh38
rs71614759 Chr.5: 90622804-90622805 on Build GRCh38

View Full Product Details