Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63684543-63685142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF3
Entrez Gene ID
23469
Gene Name
PHD finger protein 3
Gene Aliases
-
UniGene
Hs.348921
Gene Chromosome Location
Chr.6: 63635802-63715522 on Build GRCh38
Transcripts
NM_001290259.1 NM_001290260.1 NM_015153.3 XM_005248701.3 XM_005248702.3 XM_005248703.3 XM_006715426.3 XM_011535648.2 XM_017010626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3629174 c.1034C>T p.S345F Chr.6: 63684756-63684756 on Build GRCh38
1445548 c.1252A>G p.K418E Chr.6: 63684974-63684974 on Build GRCh38
6205427 c.942A>T p.A314A Chr.6: 63684664-63684664 on Build GRCh38
1445547 c.930A>C p.E310D Chr.6: 63684652-63684652 on Build GRCh38
4976297 c.834C>G p.A278A Chr.6: 63684556-63684556 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762248281 Chr.6: 63685047-63685047 on Build GRCh38
rs1002884221 Chr.6: 63684739-63684739 on Build GRCh38
rs770030568 Chr.6: 63684583-63684583 on Build GRCh38
rs777894990 Chr.6: 63684992-63684992 on Build GRCh38
rs201542595 Chr.6: 63685051-63685051 on Build GRCh38
rs201392787 Chr.6: 63684688-63684688 on Build GRCh38
rs376295445 Chr.6: 63684940-63684940 on Build GRCh38
rs947003235 Chr.6: 63684895-63684895 on Build GRCh38
rs753307394 Chr.6: 63684841-63684841 on Build GRCh38
rs775942686 Chr.6: 63684888-63684888 on Build GRCh38
rs759809824 Chr.6: 63685015-63685015 on Build GRCh38
rs758072478 Chr.6: 63684860-63684860 on Build GRCh38
rs770908929 Chr.6: 63685001-63685001 on Build GRCh38
rs970748719 Chr.6: 63684731-63684731 on Build GRCh38
rs745547553 Chr.6: 63684736-63684736 on Build GRCh38
rs748116849 Chr.6: 63684891-63684891 on Build GRCh38
rs765035004 Chr.6: 63684924-63684924 on Build GRCh38
rs750711376 Chr.6: 63684678-63684678 on Build GRCh38
rs144653440 Chr.6: 63684575-63684575 on Build GRCh38
rs144904237 Chr.6: 63684986-63684986 on Build GRCh38
rs762845653 Chr.6: 63685092-63685093 on Build GRCh38
rs961867260 Chr.6: 63684802-63684802 on Build GRCh38
rs138552118 Chr.6: 63684858-63684858 on Build GRCh38
rs773081572 Chr.6: 63684904-63684904 on Build GRCh38
rs754516098 Chr.6: 63684972-63684972 on Build GRCh38
rs200317912 Chr.6: 63685057-63685057 on Build GRCh38
rs760718754 Chr.6: 63685023-63685023 on Build GRCh38
rs752653189 Chr.6: 63684935-63684935 on Build GRCh38
rs769325218 Chr.6: 63684991-63684991 on Build GRCh38
rs369388279 Chr.6: 63684576-63684576 on Build GRCh38
rs1010461307 Chr.6: 63684649-63684649 on Build GRCh38
rs41271583 Chr.6: 63684574-63684574 on Build GRCh38
rs576956849 Chr.6: 63684734-63684734 on Build GRCh38
rs960336372 Chr.6: 63684867-63684867 on Build GRCh38
rs760949714 Chr.6: 63685115-63685115 on Build GRCh38
rs926281982 Chr.6: 63684863-63684863 on Build GRCh38
rs774232033 Chr.6: 63685014-63685014 on Build GRCh38
rs145882970 Chr.6: 63685054-63685054 on Build GRCh38
rs561539302 Chr.6: 63684980-63684980 on Build GRCh38
rs112493279 Chr.6: 63685082-63685082 on Build GRCh38
rs13197356 Chr.6: 63684788-63684788 on Build GRCh38
rs763981549 Chr.6: 63684642-63684642 on Build GRCh38
rs376420192 Chr.6: 63684806-63684806 on Build GRCh38
rs370046747 Chr.6: 63684907-63684907 on Build GRCh38
rs980376150 Chr.6: 63684782-63684782 on Build GRCh38
rs764463727 Chr.6: 63684937-63684937 on Build GRCh38
rs759821272 Chr.6: 63684918-63684918 on Build GRCh38
rs757661828 Chr.6: 63684954-63684954 on Build GRCh38
rs202212437 Chr.6: 63684580-63684580 on Build GRCh38
rs200854574 Chr.6: 63684915-63684915 on Build GRCh38
rs756521354 Chr.6: 63684844-63684844 on Build GRCh38
rs769577866 Chr.6: 63684900-63684900 on Build GRCh38
rs984571295 Chr.6: 63684751-63684751 on Build GRCh38
rs774927472 Chr.6: 63684633-63684633 on Build GRCh38
rs75261369 Chr.6: 63685016-63685016 on Build GRCh38
rs755563050 Chr.6: 63684699-63684699 on Build GRCh38
rs200635324 Chr.6: 63685092-63685092 on Build GRCh38
rs767492096 Chr.6: 63684840-63684840 on Build GRCh38
rs370463326 Chr.6: 63685049-63685049 on Build GRCh38
rs923216911 Chr.6: 63684828-63684828 on Build GRCh38
rs779732543 Chr.6: 63684866-63684866 on Build GRCh38
rs766571844 Chr.6: 63684691-63684691 on Build GRCh38
rs186370923 Chr.6: 63684750-63684750 on Build GRCh38
rs771883304 Chr.6: 63685094-63685094 on Build GRCh38
rs756258382 Chr.6: 63684774-63684774 on Build GRCh38
rs775617680 Chr.6: 63685104-63685104 on Build GRCh38
rs751649183 Chr.6: 63684697-63684697 on Build GRCh38
rs765089871 Chr.6: 63684789-63684789 on Build GRCh38
rs749253375 Chr.6: 63685081-63685081 on Build GRCh38
rs775015544 Chr.6: 63684737-63684737 on Build GRCh38
rs755147200 Chr.6: 63684834-63684834 on Build GRCh38
rs757149871 Chr.6: 63685068-63685068 on Build GRCh38
rs751224112 Chr.6: 63684963-63684963 on Build GRCh38
rs766659806 Chr.6: 63684821-63684821 on Build GRCh38
rs778204152 Chr.6: 63684847-63684847 on Build GRCh38
rs768134802 Chr.6: 63684766-63684766 on Build GRCh38
rs774783696 Chr.6: 63684916-63684916 on Build GRCh38
rs779233451 Chr.6: 63684955-63684955 on Build GRCh38
rs753599947 Chr.6: 63684648-63684648 on Build GRCh38
rs188655023 Chr.6: 63685035-63685035 on Build GRCh38
rs764933415 Chr.6: 63684663-63684663 on Build GRCh38
rs768266800 Chr.6: 63684882-63684882 on Build GRCh38
rs776695139 Chr.6: 63684768-63684768 on Build GRCh38
rs771629040 Chr.6: 63684609-63684609 on Build GRCh38
rs755645412 Chr.6: 63685073-63685073 on Build GRCh38
rs756569769 Chr.6: 63684718-63684718 on Build GRCh38
rs745604429 Chr.6: 63685089-63685089 on Build GRCh38
rs1034838889 Chr.6: 63684740-63684740 on Build GRCh38
rs751808696 Chr.6: 63684831-63684831 on Build GRCh38
rs746476185 Chr.6: 63684753-63684753 on Build GRCh38
rs775664529 Chr.6: 63685021-63685021 on Build GRCh38
rs763298881 Chr.6: 63684815-63684815 on Build GRCh38
rs749231054 Chr.6: 63684995-63684995 on Build GRCh38
rs748573789 Chr.6: 63684717-63684717 on Build GRCh38
rs760614750 Chr.6: 63684639-63684639 on Build GRCh38
rs781746178 Chr.6: 63684707-63684707 on Build GRCh38
rs761705041 Chr.6: 63684662-63684662 on Build GRCh38
rs140230222 Chr.6: 63684936-63684936 on Build GRCh38
rs373163320 Chr.6: 63684741-63684741 on Build GRCh38
rs759107522 Chr.6: 63684605-63684605 on Build GRCh38
rs778883202 Chr.6: 63685086-63685086 on Build GRCh38
rs767648874 Chr.6: 63684933-63684933 on Build GRCh38
rs574493951 Chr.6: 63684833-63684833 on Build GRCh38
rs746657831 Chr.6: 63684872-63684872 on Build GRCh38
rs754398341 Chr.6: 63685040-63685040 on Build GRCh38
rs774095026 Chr.6: 63684588-63684588 on Build GRCh38
rs752185911 Chr.6: 63685081-63685086 on Build GRCh38
rs141773184 Chr.6: 63685055-63685055 on Build GRCh38

View Full Product Details