Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143443764-143444305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA1-AS1
Entrez Gene ID
285965
Gene Name
EPHA1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.642649
Gene Chromosome Location
Chr.7: 143407813-143523449 on Build GRCh38
Transcripts
NR_033897.1 NM_177437.1

Gene Symbol
TAS2R60
Entrez Gene ID
338398
Gene Name
taste 2 receptor member 60
Gene Aliases
T2R56, T2R60
UniGene
Hs.688222
Gene Chromosome Location
Chr.7: 143443453-143444409 on Build GRCh38
Transcripts
NR_033897.1 NM_177437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381952 c.388T>G p.F130V Chr.7: 143443840-143443840 on Build GRCh38
1086803 c.572A>T p.K191I Chr.7: 143444024-143444024 on Build GRCh38
5605080 c.569A>G p.E190G Chr.7: 143444021-143444021 on Build GRCh38
3950109 c.661G>T p.G221* Chr.7: 143444113-143444113 on Build GRCh38
6176845 c.736C>G p.L246V Chr.7: 143444188-143444188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377595209 Chr.7: 143444212-143444212 on Build GRCh38
rs1006698533 Chr.7: 143443949-143443949 on Build GRCh38
rs1049692335 Chr.7: 143443833-143443833 on Build GRCh38
rs146596643 Chr.7: 143444001-143444001 on Build GRCh38
rs780562531 Chr.7: 143443926-143443926 on Build GRCh38
rs759903548 Chr.7: 143444225-143444225 on Build GRCh38
rs947438184 Chr.7: 143443927-143443927 on Build GRCh38
rs149539350 Chr.7: 143443881-143443881 on Build GRCh38
rs557957226 Chr.7: 143443997-143443997 on Build GRCh38
rs770845479 Chr.7: 143444050-143444050 on Build GRCh38
rs376147576 Chr.7: 143443887-143443887 on Build GRCh38
rs768161504 Chr.7: 143443934-143443934 on Build GRCh38
rs757555218 Chr.7: 143443910-143443910 on Build GRCh38
rs778073838 Chr.7: 143443905-143443905 on Build GRCh38
rs758887139 Chr.7: 143444267-143444267 on Build GRCh38
rs138525749 Chr.7: 143443902-143443902 on Build GRCh38
rs574740580 Chr.7: 143444009-143444009 on Build GRCh38
rs776993333 Chr.7: 143444170-143444171 on Build GRCh38
rs80017500 Chr.7: 143444047-143444047 on Build GRCh38
rs751069916 Chr.7: 143444274-143444274 on Build GRCh38
rs571748180 Chr.7: 143444263-143444263 on Build GRCh38
rs772926612 Chr.7: 143444039-143444039 on Build GRCh38
rs780149646 Chr.7: 143444221-143444221 on Build GRCh38
rs36004042 Chr.7: 143444275-143444276 on Build GRCh38
rs181824636 Chr.7: 143443941-143443941 on Build GRCh38
rs748742818 Chr.7: 143443933-143443933 on Build GRCh38
rs369309653 Chr.7: 143443903-143443903 on Build GRCh38
rs35195910 Chr.7: 143443838-143443840 on Build GRCh38
rs1039530609 Chr.7: 143444150-143444150 on Build GRCh38
rs199958730 Chr.7: 143444169-143444169 on Build GRCh38
rs767812355 Chr.7: 143443999-143443999 on Build GRCh38
rs372814575 Chr.7: 143444159-143444159 on Build GRCh38
rs191105252 Chr.7: 143444002-143444002 on Build GRCh38
rs267601365 Chr.7: 143444266-143444266 on Build GRCh38
rs397890297 Chr.7: 143443844-143443846 on Build GRCh38
rs1024707709 Chr.7: 143444089-143444089 on Build GRCh38
rs777682450 Chr.7: 143444156-143444156 on Build GRCh38
rs766615770 Chr.7: 143444217-143444217 on Build GRCh38
rs972168369 Chr.7: 143444088-143444088 on Build GRCh38
rs141559059 Chr.7: 143444091-143444091 on Build GRCh38
rs1000747422 Chr.7: 143444041-143444041 on Build GRCh38
rs777313008 Chr.7: 143444272-143444272 on Build GRCh38
rs764254335 Chr.7: 143443990-143443990 on Build GRCh38
rs759537973 Chr.7: 143443924-143443924 on Build GRCh38
rs117057228 Chr.7: 143444032-143444032 on Build GRCh38
rs758723466 Chr.7: 143443845-143443845 on Build GRCh38
rs138664103 Chr.7: 143444111-143444111 on Build GRCh38
rs752772800 Chr.7: 143443899-143443899 on Build GRCh38
rs770159996 Chr.7: 143443920-143443920 on Build GRCh38
rs761735975 Chr.7: 143444185-143444185 on Build GRCh38
rs756172558 Chr.7: 143444155-143444155 on Build GRCh38
rs138984388 Chr.7: 143444016-143444016 on Build GRCh38
rs774950147 Chr.7: 143443842-143443842 on Build GRCh38
rs189427885 Chr.7: 143443876-143443876 on Build GRCh38
rs373953807 Chr.7: 143443884-143443884 on Build GRCh38
rs746349687 Chr.7: 143443813-143443813 on Build GRCh38
rs987892629 Chr.7: 143444126-143444126 on Build GRCh38
rs547166075 Chr.7: 143443855-143443855 on Build GRCh38
rs761974777 Chr.7: 143444079-143444079 on Build GRCh38
rs765663321 Chr.7: 143444242-143444242 on Build GRCh38
rs746005734 Chr.7: 143444240-143444240 on Build GRCh38
rs369687381 Chr.7: 143443806-143443806 on Build GRCh38
rs755540104 Chr.7: 143444030-143444030 on Build GRCh38
rs781390432 Chr.7: 143443916-143443916 on Build GRCh38
rs771121061 Chr.7: 143444053-143444053 on Build GRCh38
rs753242345 Chr.7: 143444261-143444261 on Build GRCh38
rs771795316 Chr.7: 143443954-143443954 on Build GRCh38
rs772838631 Chr.7: 143443955-143443955 on Build GRCh38
rs549056725 Chr.7: 143443879-143443879 on Build GRCh38
rs866531341 Chr.7: 143444049-143444049 on Build GRCh38
rs760537750 Chr.7: 143443959-143443959 on Build GRCh38
rs1016879612 Chr.7: 143444065-143444065 on Build GRCh38
rs753830861 Chr.7: 143443973-143443973 on Build GRCh38
rs139329687 Chr.7: 143444137-143444137 on Build GRCh38
rs767577890 Chr.7: 143444104-143444104 on Build GRCh38
rs773920845 Chr.7: 143443935-143443935 on Build GRCh38
rs919299515 Chr.7: 143443854-143443854 on Build GRCh38
rs200500918 Chr.7: 143444106-143444106 on Build GRCh38
rs775511147 Chr.7: 143444183-143444183 on Build GRCh38
rs150586355 Chr.7: 143444053-143444053 on Build GRCh38
rs770383194 Chr.7: 143443822-143443822 on Build GRCh38
rs149293624 Chr.7: 143444122-143444122 on Build GRCh38
rs773226153 Chr.7: 143444195-143444195 on Build GRCh38
rs759611864 Chr.7: 143443979-143443979 on Build GRCh38
rs754238914 Chr.7: 143444117-143444117 on Build GRCh38
rs201185058 Chr.7: 143444008-143444008 on Build GRCh38
rs112054197 Chr.7: 143444128-143444128 on Build GRCh38
rs567694653 Chr.7: 143443843-143443843 on Build GRCh38
rs1036406988 Chr.7: 143443956-143443956 on Build GRCh38
rs576422213 Chr.7: 143443998-143443998 on Build GRCh38
rs550023940 Chr.7: 143444146-143444146 on Build GRCh38
rs144187592 Chr.7: 143443901-143443901 on Build GRCh38
rs765306918 Chr.7: 143444067-143444067 on Build GRCh38
rs746285550 Chr.7: 143443919-143443919 on Build GRCh38
rs774257099 Chr.7: 143444068-143444068 on Build GRCh38
rs767634505 Chr.7: 143444189-143444189 on Build GRCh38
rs146233225 Chr.7: 143444136-143444136 on Build GRCh38
rs754450920 Chr.7: 143443893-143443893 on Build GRCh38
rs749129324 Chr.7: 143444179-143444179 on Build GRCh38
rs766170232 Chr.7: 143443968-143443968 on Build GRCh38
rs778150797 Chr.7: 143443919-143443919 on Build GRCh38
rs747210213 Chr.7: 143444043-143444043 on Build GRCh38
rs759522062 Chr.7: 143444054-143444054 on Build GRCh38
rs771570444 Chr.7: 143444038-143444038 on Build GRCh38
rs751825125 Chr.7: 143443908-143443908 on Build GRCh38
rs535583913 Chr.7: 143443847-143443847 on Build GRCh38

View Full Product Details