Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27949099-27949685 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINGO2
Entrez Gene ID
158038
Gene Name
leucine rich repeat and Ig domain containing 2
Gene Aliases
LERN3, LRRN6C
UniGene
Hs.444665
Gene Chromosome Location
Chr.9: 27937617-29213981 on Build GRCh38
Transcripts
NM_001258282.1 NM_152570.2 XM_011517719.1 XM_011517724.2 XM_011517728.2 XM_017014303.1 XM_017014304.1 XM_017014305.1 XM_017014306.1 XM_017014307.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6115557 c.1142T>G p.M381R Chr.9: 27949530-27949530 on Build GRCh38
3664295 c.995T>A p.L332Q Chr.9: 27949677-27949677 on Build GRCh38
4698095 c.1218C>T p.T406T Chr.9: 27949454-27949454 on Build GRCh38
6183384 c.1312C>T p.P438S Chr.9: 27949360-27949360 on Build GRCh38
2771739 c.1519C>T p.R507C Chr.9: 27949153-27949153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777523147 Chr.9: 27949142-27949142 on Build GRCh38
rs199551773 Chr.9: 27949444-27949444 on Build GRCh38
rs866067533 Chr.9: 27949260-27949260 on Build GRCh38
rs747318931 Chr.9: 27949277-27949277 on Build GRCh38
rs760521373 Chr.9: 27949397-27949397 on Build GRCh38
rs759537365 Chr.9: 27949132-27949132 on Build GRCh38
rs117465750 Chr.9: 27949250-27949250 on Build GRCh38
rs774298229 Chr.9: 27949301-27949301 on Build GRCh38
rs112622234 Chr.9: 27949539-27949539 on Build GRCh38
rs756497213 Chr.9: 27949469-27949469 on Build GRCh38
rs144926745 Chr.9: 27949489-27949489 on Build GRCh38
rs546990838 Chr.9: 27949165-27949165 on Build GRCh38
rs748323322 Chr.9: 27949488-27949488 on Build GRCh38
rs749322030 Chr.9: 27949371-27949371 on Build GRCh38
rs778843125 Chr.9: 27949478-27949478 on Build GRCh38
rs752823711 Chr.9: 27949222-27949222 on Build GRCh38
rs377706661 Chr.9: 27949580-27949580 on Build GRCh38
rs887954577 Chr.9: 27949502-27949502 on Build GRCh38
rs148834097 Chr.9: 27949520-27949520 on Build GRCh38
rs746312003 Chr.9: 27949363-27949363 on Build GRCh38
rs771459715 Chr.9: 27949381-27949381 on Build GRCh38
rs150860330 Chr.9: 27949220-27949220 on Build GRCh38
rs565403765 Chr.9: 27949337-27949337 on Build GRCh38
rs772216428 Chr.9: 27949559-27949559 on Build GRCh38
rs761188651 Chr.9: 27949190-27949190 on Build GRCh38
rs147302902 Chr.9: 27949355-27949355 on Build GRCh38
rs761625344 Chr.9: 27949307-27949307 on Build GRCh38
rs949080582 Chr.9: 27949374-27949374 on Build GRCh38
rs181261791 Chr.9: 27949555-27949555 on Build GRCh38
rs373044342 Chr.9: 27949353-27949353 on Build GRCh38
rs377599950 Chr.9: 27949445-27949445 on Build GRCh38
rs564870453 Chr.9: 27949268-27949268 on Build GRCh38
rs557858322 Chr.9: 27949421-27949421 on Build GRCh38
rs758289947 Chr.9: 27949140-27949140 on Build GRCh38
rs755826820 Chr.9: 27949253-27949253 on Build GRCh38
rs746055552 Chr.9: 27949282-27949282 on Build GRCh38
rs983323601 Chr.9: 27949297-27949297 on Build GRCh38
rs780846144 Chr.9: 27949245-27949245 on Build GRCh38
rs780213616 Chr.9: 27949471-27949471 on Build GRCh38
rs763964876 Chr.9: 27949299-27949299 on Build GRCh38
rs759411279 Chr.9: 27949611-27949611 on Build GRCh38
rs746922513 Chr.9: 27949566-27949566 on Build GRCh38
rs543561013 Chr.9: 27949326-27949326 on Build GRCh38
rs201237761 Chr.9: 27949139-27949139 on Build GRCh38
rs779811591 Chr.9: 27949325-27949325 on Build GRCh38
rs766071387 Chr.9: 27949398-27949398 on Build GRCh38
rs754547326 Chr.9: 27949340-27949340 on Build GRCh38
rs748117682 Chr.9: 27949553-27949553 on Build GRCh38
rs748631245 Chr.9: 27949170-27949170 on Build GRCh38
rs148299908 Chr.9: 27949185-27949185 on Build GRCh38
rs894798215 Chr.9: 27949655-27949655 on Build GRCh38
rs75120285 Chr.9: 27949361-27949361 on Build GRCh38
rs142556336 Chr.9: 27949550-27949550 on Build GRCh38
rs768750698 Chr.9: 27949372-27949372 on Build GRCh38
rs762334750 Chr.9: 27949636-27949636 on Build GRCh38
rs763633223 Chr.9: 27949638-27949638 on Build GRCh38
rs779685785 Chr.9: 27949261-27949261 on Build GRCh38
rs370255317 Chr.9: 27949507-27949507 on Build GRCh38
rs751291528 Chr.9: 27949235-27949235 on Build GRCh38
rs751076098 Chr.9: 27949643-27949643 on Build GRCh38
rs772702001 Chr.9: 27949263-27949263 on Build GRCh38
rs777531806 Chr.9: 27949564-27949564 on Build GRCh38
rs997559529 Chr.9: 27949531-27949531 on Build GRCh38
rs775232627 Chr.9: 27949635-27949635 on Build GRCh38
rs369907065 Chr.9: 27949163-27949163 on Build GRCh38
rs779198594 Chr.9: 27949551-27949551 on Build GRCh38
rs965089089 Chr.9: 27949175-27949175 on Build GRCh38
rs537474007 Chr.9: 27949145-27949145 on Build GRCh38
rs17506843 Chr.9: 27949152-27949152 on Build GRCh38
rs777096048 Chr.9: 27949281-27949281 on Build GRCh38
rs113809022 Chr.9: 27949419-27949419 on Build GRCh38
rs769275016 Chr.9: 27949628-27949628 on Build GRCh38
rs775547230 Chr.9: 27949288-27949288 on Build GRCh38
rs753565851 Chr.9: 27949412-27949412 on Build GRCh38
rs141466894 Chr.9: 27949191-27949191 on Build GRCh38
rs187957678 Chr.9: 27949519-27949519 on Build GRCh38
rs748864410 Chr.9: 27949262-27949262 on Build GRCh38
rs1003222855 Chr.9: 27949631-27949631 on Build GRCh38
rs753946666 Chr.9: 27949131-27949131 on Build GRCh38
rs145458168 Chr.9: 27949506-27949506 on Build GRCh38
rs776566052 Chr.9: 27949610-27949610 on Build GRCh38
rs866834150 Chr.9: 27949147-27949147 on Build GRCh38
rs145524081 Chr.9: 27949329-27949329 on Build GRCh38
rs750277332 Chr.9: 27949323-27949323 on Build GRCh38
rs1047911784 Chr.9: 27949475-27949475 on Build GRCh38
rs767183276 Chr.9: 27949319-27949319 on Build GRCh38
rs775706847 Chr.9: 27949370-27949370 on Build GRCh38
rs539237459 Chr.9: 27949364-27949364 on Build GRCh38
rs768911785 Chr.9: 27949476-27949476 on Build GRCh38
rs772877318 Chr.9: 27949396-27949396 on Build GRCh38
rs879175880 Chr.9: 27949456-27949456 on Build GRCh38
rs767546427 Chr.9: 27949395-27949395 on Build GRCh38
rs780715183 Chr.9: 27949153-27949153 on Build GRCh38
rs767866594 Chr.9: 27949518-27949518 on Build GRCh38
rs763029018 Chr.9: 27949289-27949289 on Build GRCh38
rs749477204 Chr.9: 27949474-27949474 on Build GRCh38
rs1055305227 Chr.9: 27949646-27949646 on Build GRCh38
rs749644042 Chr.9: 27949643-27949645 on Build GRCh38
rs771116741 Chr.9: 27949193-27949193 on Build GRCh38
rs200772912 Chr.9: 27949358-27949358 on Build GRCh38
rs200667527 Chr.9: 27949391-27949391 on Build GRCh38
rs754257328 Chr.9: 27949549-27949549 on Build GRCh38
rs146048453 Chr.9: 27949162-27949162 on Build GRCh38
rs374960769 Chr.9: 27949508-27949508 on Build GRCh38
rs776730719 Chr.9: 27949219-27949219 on Build GRCh38
rs139346811 Chr.9: 27949279-27949279 on Build GRCh38
rs770113244 Chr.9: 27949283-27949283 on Build GRCh38
rs778374208 Chr.9: 27949350-27949350 on Build GRCh38
rs772751190 Chr.9: 27949176-27949176 on Build GRCh38
rs756650483 Chr.9: 27949541-27949541 on Build GRCh38
rs766567208 Chr.9: 27949121-27949121 on Build GRCh38
rs757019380 Chr.9: 27949238-27949238 on Build GRCh38
rs765051334 Chr.9: 27949516-27949516 on Build GRCh38
rs774449926 Chr.9: 27949375-27949375 on Build GRCh38
rs764089619 Chr.9: 27949234-27949234 on Build GRCh38
rs759250293 Chr.9: 27949509-27949509 on Build GRCh38
rs201950549 Chr.9: 27949434-27949434 on Build GRCh38
rs868744633 Chr.9: 27949455-27949455 on Build GRCh38
rs929456987 Chr.9: 27949424-27949424 on Build GRCh38
rs754718931 Chr.9: 27949418-27949418 on Build GRCh38

View Full Product Details