Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21333388-21333987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL9
Entrez Gene ID
55958
Gene Name
kelch like family member 9
Gene Aliases
-
UniGene
Hs.701778
Gene Chromosome Location
Chr.9: 21331019-21335430 on Build GRCh38
Transcripts
NM_018847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
360495 c.1273G>T p.E425* Chr.9: 21333587-21333587 on Build GRCh38
753767 c.1220G>T p.R407L Chr.9: 21333640-21333640 on Build GRCh38
6696911 c.1471G>A p.V491I Chr.9: 21333389-21333389 on Build GRCh38
7425153 c.1225G>T p.A409S Chr.9: 21333635-21333635 on Build GRCh38
77978 c.1219C>T p.R407C Chr.9: 21333641-21333641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746062333 Chr.9: 21333887-21333887 on Build GRCh38
rs540989730 Chr.9: 21333710-21333710 on Build GRCh38
rs201886256 Chr.9: 21333956-21333956 on Build GRCh38
rs777005831 Chr.9: 21333885-21333885 on Build GRCh38
rs1042635512 Chr.9: 21333416-21333416 on Build GRCh38
rs1003042332 Chr.9: 21333913-21333913 on Build GRCh38
rs761789349 Chr.9: 21333471-21333471 on Build GRCh38
rs773912458 Chr.9: 21333918-21333918 on Build GRCh38
rs757312612 Chr.9: 21333530-21333531 on Build GRCh38
rs757850895 Chr.9: 21333589-21333589 on Build GRCh38
rs113463325 Chr.9: 21333667-21333667 on Build GRCh38
rs928232719 Chr.9: 21333435-21333435 on Build GRCh38
rs772128717 Chr.9: 21333493-21333493 on Build GRCh38
rs556068145 Chr.9: 21333454-21333454 on Build GRCh38
rs377006324 Chr.9: 21333926-21333926 on Build GRCh38
rs183913881 Chr.9: 21333601-21333601 on Build GRCh38
rs749652681 Chr.9: 21333655-21333655 on Build GRCh38
rs985321884 Chr.9: 21333615-21333615 on Build GRCh38
rs1021583562 Chr.9: 21333534-21333534 on Build GRCh38
rs762839952 Chr.9: 21333903-21333903 on Build GRCh38
rs764090374 Chr.9: 21333917-21333917 on Build GRCh38
rs767797835 Chr.9: 21333933-21333933 on Build GRCh38
rs191333137 Chr.9: 21333466-21333466 on Build GRCh38
rs780349937 Chr.9: 21333458-21333458 on Build GRCh38
rs142721860 Chr.9: 21333420-21333420 on Build GRCh38
rs762591796 Chr.9: 21333810-21333810 on Build GRCh38
rs138147934 Chr.9: 21333681-21333681 on Build GRCh38
rs1038417828 Chr.9: 21333725-21333725 on Build GRCh38
rs939350829 Chr.9: 21333442-21333442 on Build GRCh38
rs767566812 Chr.9: 21333756-21333757 on Build GRCh38
rs533792568 Chr.9: 21333965-21333965 on Build GRCh38
rs761330125 Chr.9: 21333921-21333921 on Build GRCh38
rs747079959 Chr.9: 21333870-21333870 on Build GRCh38
rs368659358 Chr.9: 21333815-21333815 on Build GRCh38
rs762900457 Chr.9: 21333780-21333780 on Build GRCh38
rs200991754 Chr.9: 21333962-21333962 on Build GRCh38
rs755569998 Chr.9: 21333674-21333674 on Build GRCh38
rs200657430 Chr.9: 21333804-21333804 on Build GRCh38
rs751236400 Chr.9: 21333609-21333609 on Build GRCh38
rs773308378 Chr.9: 21333507-21333507 on Build GRCh38
rs139573231 Chr.9: 21333889-21333889 on Build GRCh38
rs779292562 Chr.9: 21333858-21333858 on Build GRCh38
rs766297148 Chr.9: 21333525-21333525 on Build GRCh38
rs753662484 Chr.9: 21333531-21333531 on Build GRCh38
rs150401147 Chr.9: 21333638-21333638 on Build GRCh38
rs779305744 Chr.9: 21333680-21333680 on Build GRCh38
rs776633696 Chr.9: 21333718-21333718 on Build GRCh38
rs774560249 Chr.9: 21333468-21333468 on Build GRCh38
rs369605312 Chr.9: 21333927-21333927 on Build GRCh38
rs199935926 Chr.9: 21333863-21333863 on Build GRCh38
rs77402394 Chr.9: 21333448-21333448 on Build GRCh38
rs775335120 Chr.9: 21333900-21333900 on Build GRCh38
rs76705600 Chr.9: 21333868-21333868 on Build GRCh38
rs750024292 Chr.9: 21333840-21333840 on Build GRCh38
rs771336123 Chr.9: 21333877-21333877 on Build GRCh38
rs759585223 Chr.9: 21333547-21333547 on Build GRCh38
rs772072891 Chr.9: 21333683-21333683 on Build GRCh38
rs755297963 Chr.9: 21333852-21333852 on Build GRCh38
rs142392704 Chr.9: 21333720-21333720 on Build GRCh38
rs869312950 Chr.9: 21333897-21333897 on Build GRCh38
rs751040318 Chr.9: 21333816-21333816 on Build GRCh38
rs756973113 Chr.9: 21333826-21333826 on Build GRCh38
rs760936795 Chr.9: 21333524-21333524 on Build GRCh38
rs530304630 Chr.9: 21333748-21333748 on Build GRCh38
rs112686685 Chr.9: 21333829-21333829 on Build GRCh38
rs1049033361 Chr.9: 21333721-21333721 on Build GRCh38
rs747282144 Chr.9: 21333707-21333707 on Build GRCh38
rs147375233 Chr.9: 21333536-21333536 on Build GRCh38
rs780440002 Chr.9: 21333644-21333644 on Build GRCh38
rs765080867 Chr.9: 21333549-21333549 on Build GRCh38
rs912349715 Chr.9: 21333633-21333633 on Build GRCh38
rs767146994 Chr.9: 21333834-21333834 on Build GRCh38
rs773373636 Chr.9: 21333694-21333694 on Build GRCh38
rs752309889 Chr.9: 21333550-21333550 on Build GRCh38
rs766950905 Chr.9: 21333925-21333925 on Build GRCh38
rs758913815 Chr.9: 21333867-21333867 on Build GRCh38

View Full Product Details