Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35834320-35834787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NM_001042589.2 NM_001042590.2 NM_016446.3 XM_005251483.4 XM_011517901.2 XM_011517902.2 XM_011517903.2 XM_011517904.2 XM_011517905.1 XM_011517908.2 XM_011517910.2 XM_011517911.2 XM_011517912.2 XM_011517913.2 XM_011517914.2 XM_011517915.2 XM_017014806.1

SNP Detail

SNP ID Chromosome Location
rs965789679 Chr.9: 35834431-35834431 on Build GRCh38
rs888944451 Chr.9: 35834351-35834351 on Build GRCh38
rs371008067 Chr.9: 35834733-35834733 on Build GRCh38
rs762762998 Chr.9: 35834343-35834343 on Build GRCh38
rs930281321 Chr.9: 35834710-35834710 on Build GRCh38
rs543865814 Chr.9: 35834534-35834534 on Build GRCh38
rs760858360 Chr.9: 35834718-35834719 on Build GRCh38
rs569414078 Chr.9: 35834567-35834567 on Build GRCh38
rs981016873 Chr.9: 35834568-35834568 on Build GRCh38
rs375569374 Chr.9: 35834751-35834751 on Build GRCh38
rs940884357 Chr.9: 35834673-35834673 on Build GRCh38
rs939574176 Chr.9: 35834589-35834589 on Build GRCh38
rs761322101 Chr.9: 35834750-35834750 on Build GRCh38
rs894191678 Chr.9: 35834747-35834747 on Build GRCh38
rs559214958 Chr.9: 35834400-35834400 on Build GRCh38
rs1031398885 Chr.9: 35834474-35834474 on Build GRCh38
rs372483201 Chr.9: 35834379-35834379 on Build GRCh38
rs1014483753 Chr.9: 35834436-35834436 on Build GRCh38
rs928249923 Chr.9: 35834584-35834584 on Build GRCh38
rs118098159 Chr.9: 35834722-35834722 on Build GRCh38
rs971758549 Chr.9: 35834613-35834613 on Build GRCh38
rs376988494 Chr.9: 35834693-35834693 on Build GRCh38
rs532438591 Chr.9: 35834734-35834734 on Build GRCh38
rs1048839419 Chr.9: 35834720-35834720 on Build GRCh38
rs989491735 Chr.9: 35834518-35834518 on Build GRCh38
rs754408843 Chr.9: 35834760-35834760 on Build GRCh38
rs999487449 Chr.9: 35834477-35834477 on Build GRCh38
rs1031003786 Chr.9: 35834483-35834483 on Build GRCh38
rs988017251 Chr.9: 35834620-35834620 on Build GRCh38
rs1024242424 Chr.9: 35834443-35834443 on Build GRCh38
rs562632310 Chr.9: 35834753-35834753 on Build GRCh38
rs1036612761 Chr.9: 35834690-35834690 on Build GRCh38
rs73444847 Chr.9: 35834702-35834702 on Build GRCh38
rs956818142 Chr.9: 35834517-35834517 on Build GRCh38
rs575439045 Chr.9: 35834499-35834499 on Build GRCh38
rs1007521939 Chr.9: 35834392-35834392 on Build GRCh38
rs201628999 Chr.9: 35834636-35834636 on Build GRCh38
rs185796708 Chr.9: 35834752-35834752 on Build GRCh38
rs998212338 Chr.9: 35834447-35834447 on Build GRCh38
rs867027378 Chr.9: 35834403-35834403 on Build GRCh38
rs1055942371 Chr.9: 35834376-35834376 on Build GRCh38

View Full Product Details