Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202300715-202301313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGR6
Entrez Gene ID
59352
Gene Name
leucine rich repeat containing G protein-coupled receptor 6
Gene Aliases
GPCR, VTS20631
UniGene
Hs.497402
Gene Chromosome Location
Chr.1: 202193990-202319761 on Build GRCh38
Transcripts
NM_001017403.1 NM_001017404.1 NM_021636.2 XM_005245404.3 XM_011509838.2 XM_011509839.2 XM_011509840.2 XM_011509841.2 XM_011509842.2 XM_011509843.2 XM_011509844.2 XM_011509846.2 XM_017001996.1 XM_017001997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997800 c.702-4C>A p.? Chr.1: 202301160-202301160 on Build GRCh38
5533743 c.836G>A p.G279E Chr.1: 202300899-202300899 on Build GRCh38
4229215 c.694C>T p.Q232* Chr.1: 202300913-202300913 on Build GRCh38
7077734 c.699G>A p.T233T Chr.1: 202300918-202300918 on Build GRCh38
4027021 c.394G>A p.A132T Chr.1: 202300874-202300874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747208810 Chr.1: 202301273-202301273 on Build GRCh38
rs769239277 Chr.1: 202300855-202300855 on Build GRCh38
rs765967561 Chr.1: 202300835-202300835 on Build GRCh38
rs774492417 Chr.1: 202300814-202300814 on Build GRCh38
rs1033452644 Chr.1: 202301135-202301135 on Build GRCh38
rs377364000 Chr.1: 202300820-202300820 on Build GRCh38
rs767233720 Chr.1: 202301203-202301203 on Build GRCh38
rs769069124 Chr.1: 202301149-202301149 on Build GRCh38
rs535217138 Chr.1: 202300957-202300957 on Build GRCh38
rs188651046 Chr.1: 202300877-202300877 on Build GRCh38
rs375015427 Chr.1: 202301131-202301131 on Build GRCh38
rs573677155 Chr.1: 202301032-202301032 on Build GRCh38
rs770198053 Chr.1: 202300804-202300804 on Build GRCh38
rs756926299 Chr.1: 202301120-202301120 on Build GRCh38
rs998551428 Chr.1: 202301086-202301086 on Build GRCh38
rs140481467 Chr.1: 202301099-202301099 on Build GRCh38
rs753778515 Chr.1: 202300830-202300830 on Build GRCh38
rs370716767 Chr.1: 202300919-202300919 on Build GRCh38
rs201828130 Chr.1: 202300807-202300807 on Build GRCh38
rs1045544495 Chr.1: 202301031-202301031 on Build GRCh38
rs750014816 Chr.1: 202301229-202301229 on Build GRCh38
rs200743881 Chr.1: 202300952-202300952 on Build GRCh38
rs761470187 Chr.1: 202301202-202301202 on Build GRCh38
rs200044070 Chr.1: 202300953-202300953 on Build GRCh38
rs755672763 Chr.1: 202301241-202301241 on Build GRCh38
rs1028594232 Chr.1: 202300747-202300747 on Build GRCh38
rs770302410 Chr.1: 202300901-202300901 on Build GRCh38
rs762967039 Chr.1: 202301175-202301175 on Build GRCh38
rs763956004 Chr.1: 202300971-202300971 on Build GRCh38
rs866514672 Chr.1: 202300917-202300917 on Build GRCh38
rs771190946 Chr.1: 202301282-202301282 on Build GRCh38
rs547512713 Chr.1: 202300788-202300788 on Build GRCh38
rs926249526 Chr.1: 202300762-202300762 on Build GRCh38
rs762082967 Chr.1: 202300938-202300938 on Build GRCh38
rs759191889 Chr.1: 202300915-202300915 on Build GRCh38
rs758216363 Chr.1: 202300852-202300852 on Build GRCh38
rs752552373 Chr.1: 202300851-202300851 on Build GRCh38
rs753081203 Chr.1: 202301055-202301055 on Build GRCh38
rs968299811 Chr.1: 202301112-202301112 on Build GRCh38
rs760867154 Chr.1: 202301166-202301166 on Build GRCh38
rs757648365 Chr.1: 202301271-202301271 on Build GRCh38
rs939017480 Chr.1: 202300791-202300791 on Build GRCh38
rs986596296 Chr.1: 202301146-202301146 on Build GRCh38
rs748566693 Chr.1: 202301261-202301261 on Build GRCh38
rs775614276 Chr.1: 202301174-202301174 on Build GRCh38
rs190134164 Chr.1: 202301160-202301160 on Build GRCh38
rs763858097 Chr.1: 202301180-202301180 on Build GRCh38
rs780581243 Chr.1: 202301134-202301134 on Build GRCh38
rs780735961 Chr.1: 202300866-202300866 on Build GRCh38
rs191347825 Chr.1: 202300814-202300814 on Build GRCh38
rs1029528335 Chr.1: 202300975-202300975 on Build GRCh38
rs371853249 Chr.1: 202301162-202301162 on Build GRCh38
rs186931980 Chr.1: 202300744-202300744 on Build GRCh38
rs909351497 Chr.1: 202301210-202301210 on Build GRCh38
rs956689638 Chr.1: 202301052-202301052 on Build GRCh38
rs772111614 Chr.1: 202300944-202300944 on Build GRCh38
rs758648270 Chr.1: 202301270-202301270 on Build GRCh38
rs929534261 Chr.1: 202301188-202301188 on Build GRCh38
rs755481197 Chr.1: 202301259-202301259 on Build GRCh38
rs763249127 Chr.1: 202300806-202300806 on Build GRCh38
rs764363529 Chr.1: 202301132-202301132 on Build GRCh38
rs779331350 Chr.1: 202301151-202301151 on Build GRCh38
rs374763129 Chr.1: 202300935-202300935 on Build GRCh38
rs184343559 Chr.1: 202300842-202300842 on Build GRCh38
rs759477997 Chr.1: 202300967-202300969 on Build GRCh38
rs7553800 Chr.1: 202300864-202300864 on Build GRCh38
rs985479292 Chr.1: 202301291-202301291 on Build GRCh38
rs765198632 Chr.1: 202300846-202300846 on Build GRCh38
rs771101043 Chr.1: 202301170-202301170 on Build GRCh38
rs758810089 Chr.1: 202301078-202301078 on Build GRCh38
rs892780656 Chr.1: 202301106-202301106 on Build GRCh38
rs779527512 Chr.1: 202301245-202301245 on Build GRCh38
rs751225109 Chr.1: 202301114-202301114 on Build GRCh38
rs369047348 Chr.1: 202301254-202301254 on Build GRCh38
rs185185831 Chr.1: 202301136-202301136 on Build GRCh38
rs748393899 Chr.1: 202301155-202301155 on Build GRCh38
rs1011911535 Chr.1: 202301115-202301115 on Build GRCh38
rs749687454 Chr.1: 202300918-202300918 on Build GRCh38
rs192833728 Chr.1: 202300969-202300969 on Build GRCh38
rs756762118 Chr.1: 202300856-202300856 on Build GRCh38
rs767613918 Chr.1: 202300821-202300821 on Build GRCh38
rs1049267213 Chr.1: 202300853-202300853 on Build GRCh38
rs567636323 Chr.1: 202300797-202300797 on Build GRCh38
rs202195510 Chr.1: 202301260-202301260 on Build GRCh38

View Full Product Details