Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 244423566-244424099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
UniGene
Hs.498313
Gene Chromosome Location
Chr.1: 244408492-244452134 on Build GRCh38
Transcripts
NM_001126.3 XM_011544108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
25651 c.479G>A p.G160D Chr.1: 244424055-244424055 on Build GRCh38
7300035 c.491_492insA p.I166fs*22 Chr.1: 244424042-244424043 on Build GRCh38
7147645 c.577G>C p.E193Q Chr.1: 244423957-244423957 on Build GRCh38
268608 c.546C>T p.C182C Chr.1: 244423988-244423988 on Build GRCh38
80835 c.495C>A p.G165G Chr.1: 244424039-244424039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1018876455 Chr.1: 244423791-244423791 on Build GRCh38
rs909925861 Chr.1: 244423897-244423898 on Build GRCh38
rs555900143 Chr.1: 244423753-244423753 on Build GRCh38
rs766235390 Chr.1: 244423985-244423985 on Build GRCh38
rs528269285 Chr.1: 244424004-244424004 on Build GRCh38
rs774923379 Chr.1: 244423790-244423790 on Build GRCh38
rs948003705 Chr.1: 244423680-244423680 on Build GRCh38
rs533491651 Chr.1: 244423849-244423849 on Build GRCh38
rs985281642 Chr.1: 244423672-244423672 on Build GRCh38
rs749735276 Chr.1: 244424022-244424022 on Build GRCh38
rs749788469 Chr.1: 244423918-244423918 on Build GRCh38
rs113277726 Chr.1: 244423771-244423771 on Build GRCh38
rs998172424 Chr.1: 244423897-244423897 on Build GRCh38
rs903586661 Chr.1: 244423872-244423872 on Build GRCh38
rs35548214 Chr.1: 244423859-244423860 on Build GRCh38
rs140739234 Chr.1: 244423597-244423597 on Build GRCh38
rs150166816 Chr.1: 244423687-244423687 on Build GRCh38
rs771622158 Chr.1: 244423923-244423923 on Build GRCh38
rs12134870 Chr.1: 244423999-244423999 on Build GRCh38
rs142986789 Chr.1: 244424005-244424005 on Build GRCh38
rs749471591 Chr.1: 244423868-244423868 on Build GRCh38
rs1007141183 Chr.1: 244423773-244423773 on Build GRCh38
rs992040280 Chr.1: 244423775-244423775 on Build GRCh38
rs371020647 Chr.1: 244423942-244423942 on Build GRCh38
rs909919315 Chr.1: 244423626-244423626 on Build GRCh38
rs544112122 Chr.1: 244423891-244423891 on Build GRCh38
rs774484934 Chr.1: 244423933-244423933 on Build GRCh38
rs1026604306 Chr.1: 244423887-244423887 on Build GRCh38
rs935020144 Chr.1: 244423889-244423889 on Build GRCh38
rs147807505 Chr.1: 244423817-244423834 on Build GRCh38
rs761553060 Chr.1: 244423961-244423961 on Build GRCh38
rs57748551 Chr.1: 244423893-244423893 on Build GRCh38
rs754469099 Chr.1: 244423991-244423991 on Build GRCh38
rs947876037 Chr.1: 244423869-244423869 on Build GRCh38
rs142254155 Chr.1: 244423685-244423685 on Build GRCh38
rs565481613 Chr.1: 244423970-244423970 on Build GRCh38
rs764982618 Chr.1: 244423969-244423969 on Build GRCh38
rs569810715 Chr.1: 244423657-244423657 on Build GRCh38
rs778267794 Chr.1: 244424016-244424016 on Build GRCh38
rs3127459 Chr.1: 244423764-244423764 on Build GRCh38
rs749633068 Chr.1: 244423938-244423939 on Build GRCh38
rs541684650 Chr.1: 244423766-244423766 on Build GRCh38
rs199525756 Chr.1: 244423806-244423811 on Build GRCh38
rs939546916 Chr.1: 244423689-244423689 on Build GRCh38
rs1053060154 Chr.1: 244423725-244423725 on Build GRCh38
rs1010649637 Chr.1: 244423742-244423742 on Build GRCh38
rs755881622 Chr.1: 244424009-244424009 on Build GRCh38
rs972035885 Chr.1: 244423735-244423735 on Build GRCh38
rs201478770 Chr.1: 244423988-244423988 on Build GRCh38
rs771401174 Chr.1: 244424028-244424028 on Build GRCh38
rs773753614 Chr.1: 244423907-244423907 on Build GRCh38
rs1043876866 Chr.1: 244423870-244423870 on Build GRCh38
rs757411387 Chr.1: 244424021-244424021 on Build GRCh38
rs199878104 Chr.1: 244423941-244423941 on Build GRCh38
rs1029566954 Chr.1: 244423587-244423587 on Build GRCh38
rs775593460 Chr.1: 244423959-244423959 on Build GRCh38
rs1029207233 Chr.1: 244423898-244423898 on Build GRCh38
rs758330279 Chr.1: 244423972-244423972 on Build GRCh38

View Full Product Details