Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 68482931-68483496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A16
Entrez Gene ID
8034
Gene Name
solute carrier family 25 member 16
Gene Aliases
D10S105E, GDA, GDC, HGT.1, ML7, hML7
UniGene
Hs.180408
Gene Chromosome Location
Chr.10: 68479961-68527523 on Build GRCh38
Transcripts
NM_001324312.1 NM_001324314.1 NM_001324315.1 NM_001324317.1 NM_152707.3 NR_136737.1 XM_011540211.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913076 c.942T>A p.S314S Chr.10: 68483489-68483489 on Build GRCh38
4844734 c.993C>G p.L331L Chr.10: 68483438-68483438 on Build GRCh38
396663 c.975G>T p.M325I Chr.10: 68483456-68483456 on Build GRCh38
5600487 c.937C>T p.P313S Chr.10: 68483494-68483494 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766865694 Chr.10: 68483407-68483409 on Build GRCh38
rs765303690 Chr.10: 68483425-68483426 on Build GRCh38
rs761922105 Chr.10: 68483471-68483471 on Build GRCh38
rs746410508 Chr.10: 68483440-68483440 on Build GRCh38
rs925720421 Chr.10: 68483364-68483364 on Build GRCh38
rs781164793 Chr.10: 68483414-68483414 on Build GRCh38
rs954587067 Chr.10: 68483008-68483008 on Build GRCh38
rs369766981 Chr.10: 68483465-68483465 on Build GRCh38
rs996545024 Chr.10: 68482989-68482989 on Build GRCh38
rs367914649 Chr.10: 68483398-68483398 on Build GRCh38
rs954748065 Chr.10: 68483241-68483241 on Build GRCh38
rs150740752 Chr.10: 68483422-68483422 on Build GRCh38
rs747313620 Chr.10: 68483464-68483464 on Build GRCh38
rs779516559 Chr.10: 68483205-68483205 on Build GRCh38
rs988283103 Chr.10: 68483392-68483392 on Build GRCh38
rs562328327 Chr.10: 68483012-68483012 on Build GRCh38
rs916276460 Chr.10: 68483102-68483102 on Build GRCh38
rs1017680539 Chr.10: 68483251-68483251 on Build GRCh38
rs752493008 Chr.10: 68483421-68483421 on Build GRCh38
rs10998219 Chr.10: 68483148-68483148 on Build GRCh38
rs751536409 Chr.10: 68483015-68483015 on Build GRCh38
rs765221928 Chr.10: 68483393-68483393 on Build GRCh38
rs913526751 Chr.10: 68483221-68483221 on Build GRCh38
rs749852857 Chr.10: 68483149-68483149 on Build GRCh38
rs4350264 Chr.10: 68483189-68483189 on Build GRCh38
rs973297885 Chr.10: 68483063-68483063 on Build GRCh38
rs769006055 Chr.10: 68483275-68483275 on Build GRCh38
rs910221540 Chr.10: 68482980-68482980 on Build GRCh38
rs1004919084 Chr.10: 68483359-68483359 on Build GRCh38
rs1046787477 Chr.10: 68483124-68483124 on Build GRCh38
rs181611849 Chr.10: 68483259-68483259 on Build GRCh38
rs779292837 Chr.10: 68483284-68483284 on Build GRCh38
rs892681257 Chr.10: 68483090-68483090 on Build GRCh38
rs752905262 Chr.10: 68483443-68483444 on Build GRCh38
rs549328403 Chr.10: 68483378-68483378 on Build GRCh38
rs963198126 Chr.10: 68483050-68483050 on Build GRCh38
rs914075218 Chr.10: 68483416-68483416 on Build GRCh38
rs142972256 Chr.10: 68483358-68483358 on Build GRCh38
rs996293724 Chr.10: 68483277-68483277 on Build GRCh38
rs529685622 Chr.10: 68483377-68483377 on Build GRCh38
rs935026915 Chr.10: 68483158-68483158 on Build GRCh38
rs943078895 Chr.10: 68482993-68482993 on Build GRCh38
rs572182292 Chr.10: 68483242-68483242 on Build GRCh38
rs1052305372 Chr.10: 68483322-68483322 on Build GRCh38
rs757754624 Chr.10: 68483069-68483070 on Build GRCh38
rs748886574 Chr.10: 68483427-68483427 on Build GRCh38
rs183860204 Chr.10: 68483456-68483456 on Build GRCh38
rs564902165 Chr.10: 68483225-68483225 on Build GRCh38
rs576418962 Chr.10: 68483188-68483188 on Build GRCh38
rs947519487 Chr.10: 68483227-68483227 on Build GRCh38
rs772525059 Chr.10: 68483441-68483441 on Build GRCh38
rs536645371 Chr.10: 68483118-68483118 on Build GRCh38
rs574124010 Chr.10: 68483067-68483067 on Build GRCh38
rs963290932 Chr.10: 68483380-68483380 on Build GRCh38
rs749688021 Chr.10: 68483468-68483468 on Build GRCh38
rs779697586 Chr.10: 68483424-68483425 on Build GRCh38
rs544224165 Chr.10: 68482974-68482974 on Build GRCh38
rs754318009 Chr.10: 68483425-68483425 on Build GRCh38
rs756305318 Chr.10: 68483358-68483359 on Build GRCh38
rs3200066 Chr.10: 68482960-68482960 on Build GRCh38
rs1046676512 Chr.10: 68483433-68483433 on Build GRCh38
rs1052674710 Chr.10: 68483088-68483088 on Build GRCh38
rs770532162 Chr.10: 68483428-68483428 on Build GRCh38
rs368621299 Chr.10: 68483038-68483038 on Build GRCh38
rs79458978 Chr.10: 68483047-68483047 on Build GRCh38

View Full Product Details