Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95610109-95610532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6551192 c.2168C>A p.A723D Chr.10: 95610235-95610235 on Build GRCh38
4951032 c.2206+2T>C p.? Chr.10: 95610195-95610195 on Build GRCh38
7160920 c.2189G>A p.G730D Chr.10: 95610214-95610214 on Build GRCh38
921811 c.2194C>T p.R732C Chr.10: 95610209-95610209 on Build GRCh38
7285522 c.2148T>C p.S716S Chr.10: 95610255-95610255 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs544907804 Chr.10: 95610376-95610376 on Build GRCh38
rs770249022 Chr.10: 95610217-95610217 on Build GRCh38
rs566962570 Chr.10: 95610390-95610390 on Build GRCh38
rs376649056 Chr.10: 95610308-95610308 on Build GRCh38
rs546924848 Chr.10: 95610387-95610387 on Build GRCh38
rs987254679 Chr.10: 95610163-95610163 on Build GRCh38
rs1028497188 Chr.10: 95610472-95610472 on Build GRCh38
rs745966631 Chr.10: 95610154-95610154 on Build GRCh38
rs1047521252 Chr.10: 95610306-95610306 on Build GRCh38
rs147842812 Chr.10: 95610264-95610264 on Build GRCh38
rs186124841 Chr.10: 95610322-95610322 on Build GRCh38
rs537885854 Chr.10: 95610421-95610421 on Build GRCh38
rs1052752911 Chr.10: 95610454-95610454 on Build GRCh38
rs751028815 Chr.10: 95610146-95610146 on Build GRCh38
rs771584878 Chr.10: 95610202-95610202 on Build GRCh38
rs771578568 Chr.10: 95610295-95610295 on Build GRCh38
rs904689735 Chr.10: 95610427-95610427 on Build GRCh38
rs751255086 Chr.10: 95610340-95610340 on Build GRCh38
rs752669339 Chr.10: 95610260-95610260 on Build GRCh38
rs200513474 Chr.10: 95610180-95610180 on Build GRCh38
rs531085704 Chr.10: 95610362-95610362 on Build GRCh38
rs746451513 Chr.10: 95610304-95610304 on Build GRCh38
rs763574839 Chr.10: 95610302-95610304 on Build GRCh38
rs757102092 Chr.10: 95610283-95610283 on Build GRCh38
rs773714478 Chr.10: 95610226-95610226 on Build GRCh38
rs771294999 Chr.10: 95610331-95610331 on Build GRCh38
rs542177817 Chr.10: 95610299-95610299 on Build GRCh38
rs766787482 Chr.10: 95610241-95610241 on Build GRCh38
rs943343 Chr.10: 95610486-95610486 on Build GRCh38
rs774916138 Chr.10: 95610208-95610208 on Build GRCh38
rs749048624 Chr.10: 95610149-95610149 on Build GRCh38
rs571497010 Chr.10: 95610450-95610450 on Build GRCh38
rs187285529 Chr.10: 95610465-95610465 on Build GRCh38
rs943342 Chr.10: 95610440-95610440 on Build GRCh38
rs759965579 Chr.10: 95610340-95610340 on Build GRCh38
rs587777858 Chr.10: 95610272-95610272 on Build GRCh38
rs530752063 Chr.10: 95610383-95610383 on Build GRCh38
rs374052426 Chr.10: 95610243-95610243 on Build GRCh38
rs759717025 Chr.10: 95610250-95610250 on Build GRCh38
rs190649781 Chr.10: 95610391-95610391 on Build GRCh38
rs182758188 Chr.10: 95610398-95610398 on Build GRCh38
rs373708599 Chr.10: 95610396-95610396 on Build GRCh38
rs753837028 Chr.10: 95610282-95610282 on Build GRCh38
rs893406039 Chr.10: 95610455-95610455 on Build GRCh38
rs760203992 Chr.10: 95610209-95610209 on Build GRCh38
rs770851681 Chr.10: 95610169-95610169 on Build GRCh38
rs780180232 Chr.10: 95610232-95610232 on Build GRCh38
rs10882640 Chr.10: 95610182-95610182 on Build GRCh38
rs113068766 Chr.10: 95610385-95610385 on Build GRCh38
rs749860654 Chr.10: 95610329-95610329 on Build GRCh38
rs906949111 Chr.10: 95610430-95610430 on Build GRCh38
rs900113576 Chr.10: 95610402-95610402 on Build GRCh38
rs144531891 Chr.10: 95610207-95610207 on Build GRCh38
rs774626215 Chr.10: 95610337-95610337 on Build GRCh38
rs569288667 Chr.10: 95610420-95610420 on Build GRCh38
rs573358145 Chr.10: 95610487-95610487 on Build GRCh38
rs202169492 Chr.10: 95610227-95610227 on Build GRCh38
rs778636647 Chr.10: 95610287-95610287 on Build GRCh38
rs960027872 Chr.10: 95610485-95610485 on Build GRCh38
rs756291410 Chr.10: 95610263-95610263 on Build GRCh38

View Full Product Details