Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19219372-19219971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
UniGene
Hs.439200
Gene Chromosome Location
Chr.1: 19215664-19251559 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

Gene Symbol
LOC101927895
Entrez Gene ID
101927895
Gene Name
uncharacterized LOC101927895
Gene Aliases
-
UniGene
Hs.660534
Gene Chromosome Location
Chr.1: 19210386-19240704 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1211830 c.2800T>G p.L934V Chr.1: 19219571-19219571 on Build GRCh38
6996593 c.2887G>A p.V963M Chr.1: 19219398-19219398 on Build GRCh38
1601449 c.2758C>T p.R920* Chr.1: 19219613-19219613 on Build GRCh38
3728040 c.2780C>T p.S927L Chr.1: 19219591-19219591 on Build GRCh38
5025200 c.2724A>T p.R908R Chr.1: 19219647-19219647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747982066 Chr.1: 19219703-19219703 on Build GRCh38
rs775064384 Chr.1: 19219495-19219495 on Build GRCh38
rs773451001 Chr.1: 19219534-19219534 on Build GRCh38
rs922706368 Chr.1: 19219565-19219565 on Build GRCh38
rs527808847 Chr.1: 19219880-19219880 on Build GRCh38
rs754657954 Chr.1: 19219867-19219867 on Build GRCh38
rs368104086 Chr.1: 19219702-19219702 on Build GRCh38
rs34350799 Chr.1: 19219399-19219399 on Build GRCh38
rs984611979 Chr.1: 19219524-19219524 on Build GRCh38
rs765270968 Chr.1: 19219530-19219530 on Build GRCh38
rs763072595 Chr.1: 19219551-19219551 on Build GRCh38
rs562770564 Chr.1: 19219612-19219612 on Build GRCh38
rs751831249 Chr.1: 19219558-19219558 on Build GRCh38
rs370639188 Chr.1: 19219913-19219913 on Build GRCh38
rs548595466 Chr.1: 19219613-19219613 on Build GRCh38
rs767596155 Chr.1: 19219655-19219655 on Build GRCh38
rs34207326 Chr.1: 19219569-19219569 on Build GRCh38
rs201239769 Chr.1: 19219695-19219695 on Build GRCh38
rs751573733 Chr.1: 19219515-19219518 on Build GRCh38
rs139335524 Chr.1: 19219619-19219619 on Build GRCh38
rs752890450 Chr.1: 19219682-19219683 on Build GRCh38
rs766717020 Chr.1: 19219554-19219554 on Build GRCh38
rs946570469 Chr.1: 19219648-19219648 on Build GRCh38
rs776565472 Chr.1: 19219398-19219398 on Build GRCh38
rs768489406 Chr.1: 19219507-19219507 on Build GRCh38
rs778457223 Chr.1: 19219490-19219490 on Build GRCh38
rs764588699 Chr.1: 19219660-19219660 on Build GRCh38
rs374448290 Chr.1: 19219448-19219448 on Build GRCh38
rs757835614 Chr.1: 19219605-19219605 on Build GRCh38
rs778091054 Chr.1: 19219593-19219593 on Build GRCh38
rs759487899 Chr.1: 19219733-19219733 on Build GRCh38
rs765343007 Chr.1: 19219402-19219402 on Build GRCh38
rs374950905 Chr.1: 19219658-19219658 on Build GRCh38
rs140097250 Chr.1: 19219883-19219884 on Build GRCh38
rs767904848 Chr.1: 19219602-19219602 on Build GRCh38
rs756676817 Chr.1: 19219591-19219591 on Build GRCh38
rs767902629 Chr.1: 19219580-19219580 on Build GRCh38
rs746811083 Chr.1: 19219503-19219503 on Build GRCh38
rs778017096 Chr.1: 19219695-19219696 on Build GRCh38
rs779513279 Chr.1: 19219680-19219680 on Build GRCh38
rs1023507564 Chr.1: 19219460-19219460 on Build GRCh38
rs781137936 Chr.1: 19219658-19219659 on Build GRCh38
rs758659439 Chr.1: 19219445-19219445 on Build GRCh38
rs755331088 Chr.1: 19219449-19219449 on Build GRCh38
rs144354996 Chr.1: 19219623-19219623 on Build GRCh38
rs748675045 Chr.1: 19219458-19219458 on Build GRCh38
rs755027275 Chr.1: 19219556-19219556 on Build GRCh38
rs749952140 Chr.1: 19219893-19219893 on Build GRCh38
rs894478957 Chr.1: 19219767-19219767 on Build GRCh38
rs747855639 Chr.1: 19219618-19219618 on Build GRCh38
rs1012376410 Chr.1: 19219841-19219841 on Build GRCh38
rs961387374 Chr.1: 19219451-19219451 on Build GRCh38
rs756275805 Chr.1: 19219692-19219694 on Build GRCh38
rs371109445 Chr.1: 19219544-19219544 on Build GRCh38
rs140520413 Chr.1: 19219671-19219671 on Build GRCh38
rs766662046 Chr.1: 19219446-19219446 on Build GRCh38
rs1013026997 Chr.1: 19219850-19219850 on Build GRCh38
rs753186319 Chr.1: 19219590-19219590 on Build GRCh38
rs184804579 Chr.1: 19219886-19219886 on Build GRCh38
rs566785135 Chr.1: 19219721-19219721 on Build GRCh38
rs781655970 Chr.1: 19219452-19219452 on Build GRCh38
rs369101824 Chr.1: 19219914-19219914 on Build GRCh38
rs772351253 Chr.1: 19219737-19219737 on Build GRCh38
rs753478603 Chr.1: 19219798-19219798 on Build GRCh38
rs529971469 Chr.1: 19219622-19219622 on Build GRCh38
rs749216366 Chr.1: 19219715-19219715 on Build GRCh38
rs754337469 Chr.1: 19219667-19219667 on Build GRCh38
rs954229339 Chr.1: 19219572-19219572 on Build GRCh38
rs1006468808 Chr.1: 19219902-19219903 on Build GRCh38
rs771125704 Chr.1: 19219641-19219641 on Build GRCh38
rs146583625 Chr.1: 19219681-19219681 on Build GRCh38
rs769475130 Chr.1: 19219707-19219707 on Build GRCh38
rs371960293 Chr.1: 19219709-19219709 on Build GRCh38
rs1027497197 Chr.1: 19219879-19219879 on Build GRCh38
rs759773407 Chr.1: 19219645-19219645 on Build GRCh38
rs755451780 Chr.1: 19219563-19219563 on Build GRCh38
rs781006015 Chr.1: 19219617-19219617 on Build GRCh38
rs775507363 Chr.1: 19219739-19219739 on Build GRCh38
rs368493850 Chr.1: 19219522-19219522 on Build GRCh38
rs1048796664 Chr.1: 19219849-19219849 on Build GRCh38
rs143947690 Chr.1: 19219626-19219626 on Build GRCh38
rs766882757 Chr.1: 19219423-19219423 on Build GRCh38
rs753129813 Chr.1: 19219656-19219656 on Build GRCh38
rs745344544 Chr.1: 19219491-19219491 on Build GRCh38
rs376616400 Chr.1: 19219727-19219727 on Build GRCh38
rs560254786 Chr.1: 19219742-19219742 on Build GRCh38
rs756767859 Chr.1: 19219485-19219485 on Build GRCh38
rs371761348 Chr.1: 19219499-19219499 on Build GRCh38
rs749946960 Chr.1: 19219600-19219600 on Build GRCh38

View Full Product Details