Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54758346-54758890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARS2
Entrez Gene ID
25973
Gene Name
prolyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
MT-PRORS, proRS
UniGene
Hs.380169
Gene Chromosome Location
Chr.1: 54756898-54764553 on Build GRCh38
Transcripts
NM_152268.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
464784 c.628T>C p.Y210H Chr.1: 54758534-54758534 on Build GRCh38
1748520 c.657C>T p.A219A Chr.1: 54758505-54758505 on Build GRCh38
2194148 c.718C>T p.P240S Chr.1: 54758444-54758444 on Build GRCh38
910951 c.656C>T p.A219V Chr.1: 54758506-54758506 on Build GRCh38
6488697 c.719C>T p.P240L Chr.1: 54758443-54758443 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780812512 Chr.1: 54758681-54758681 on Build GRCh38
rs771021398 Chr.1: 54758424-54758424 on Build GRCh38
rs1013500637 Chr.1: 54758477-54758477 on Build GRCh38
rs769659836 Chr.1: 54758560-54758560 on Build GRCh38
rs749996806 Chr.1: 54758684-54758684 on Build GRCh38
rs772609589 Chr.1: 54758420-54758420 on Build GRCh38
rs747777009 Chr.1: 54758536-54758536 on Build GRCh38
rs770846623 Chr.1: 54758834-54758834 on Build GRCh38
rs1047341439 Chr.1: 54758390-54758390 on Build GRCh38
rs767329181 Chr.1: 54758790-54758790 on Build GRCh38
rs768267306 Chr.1: 54758692-54758692 on Build GRCh38
rs1013615420 Chr.1: 54758724-54758724 on Build GRCh38
rs772080916 Chr.1: 54758646-54758646 on Build GRCh38
rs199529531 Chr.1: 54758596-54758596 on Build GRCh38
rs550757432 Chr.1: 54758503-54758503 on Build GRCh38
rs747976308 Chr.1: 54758823-54758823 on Build GRCh38
rs369889041 Chr.1: 54758796-54758796 on Build GRCh38
rs778638516 Chr.1: 54758822-54758822 on Build GRCh38
rs757784646 Chr.1: 54758388-54758388 on Build GRCh38
rs371891675 Chr.1: 54758473-54758473 on Build GRCh38
rs771454009 Chr.1: 54758751-54758751 on Build GRCh38
rs777294427 Chr.1: 54758544-54758544 on Build GRCh38
rs373414784 Chr.1: 54758690-54758690 on Build GRCh38
rs777068604 Chr.1: 54758764-54758764 on Build GRCh38
rs149840204 Chr.1: 54758839-54758839 on Build GRCh38
rs886759126 Chr.1: 54758399-54758399 on Build GRCh38
rs765798606 Chr.1: 54758775-54758775 on Build GRCh38
rs141760650 Chr.1: 54758558-54758558 on Build GRCh38
rs775719456 Chr.1: 54758487-54758487 on Build GRCh38
rs770966124 Chr.1: 54758557-54758557 on Build GRCh38
rs778691533 Chr.1: 54758710-54758710 on Build GRCh38
rs200135533 Chr.1: 54758597-54758597 on Build GRCh38
rs200044481 Chr.1: 54758507-54758507 on Build GRCh38
rs772256400 Chr.1: 54758825-54758825 on Build GRCh38
rs771542110 Chr.1: 54758413-54758413 on Build GRCh38
rs368867351 Chr.1: 54758663-54758663 on Build GRCh38
rs746501383 Chr.1: 54758613-54758613 on Build GRCh38
rs750862062 Chr.1: 54758592-54758592 on Build GRCh38
rs766831879 Chr.1: 54758659-54758659 on Build GRCh38
rs1029901014 Chr.1: 54758510-54758510 on Build GRCh38
rs761129871 Chr.1: 54758656-54758656 on Build GRCh38
rs898758725 Chr.1: 54758683-54758683 on Build GRCh38
rs756763627 Chr.1: 54758617-54758617 on Build GRCh38
rs915243147 Chr.1: 54758604-54758604 on Build GRCh38
rs969394666 Chr.1: 54758484-54758484 on Build GRCh38
rs749950887 Chr.1: 54758765-54758765 on Build GRCh38
rs747105461 Chr.1: 54758828-54758828 on Build GRCh38
rs763099969 Chr.1: 54758490-54758490 on Build GRCh38
rs536531769 Chr.1: 54758579-54758579 on Build GRCh38
rs761939434 Chr.1: 54758586-54758586 on Build GRCh38
rs559553257 Chr.1: 54758868-54758868 on Build GRCh38
rs757733146 Chr.1: 54758511-54758511 on Build GRCh38
rs754963725 Chr.1: 54758817-54758817 on Build GRCh38
rs928111486 Chr.1: 54758655-54758655 on Build GRCh38
rs777336137 Chr.1: 54758606-54758606 on Build GRCh38
rs957295551 Chr.1: 54758583-54758583 on Build GRCh38
rs768910389 Chr.1: 54758637-54758637 on Build GRCh38
rs759872268 Chr.1: 54758670-54758670 on Build GRCh38
rs764189076 Chr.1: 54758497-54758497 on Build GRCh38
rs775173656 Chr.1: 54758575-54758575 on Build GRCh38
rs747606931 Chr.1: 54758412-54758412 on Build GRCh38
rs755649256 Chr.1: 54758792-54758792 on Build GRCh38
rs955262716 Chr.1: 54758516-54758516 on Build GRCh38
rs775978240 Chr.1: 54758854-54758854 on Build GRCh38
rs774505231 Chr.1: 54758639-54758639 on Build GRCh38
rs746538075 Chr.1: 54758546-54758546 on Build GRCh38
rs1033319393 Chr.1: 54758784-54758784 on Build GRCh38
rs143143865 Chr.1: 54758720-54758720 on Build GRCh38
rs998668714 Chr.1: 54758855-54758855 on Build GRCh38
rs750805913 Chr.1: 54758500-54758501 on Build GRCh38
rs746271687 Chr.1: 54758404-54758404 on Build GRCh38
rs758573965 Chr.1: 54758376-54758376 on Build GRCh38
rs201021871 Chr.1: 54758859-54758859 on Build GRCh38
rs1023714299 Chr.1: 54758734-54758734 on Build GRCh38
rs957313936 Chr.1: 54758793-54758793 on Build GRCh38
rs759450188 Chr.1: 54758459-54758459 on Build GRCh38
rs1031563655 Chr.1: 54758548-54758548 on Build GRCh38
rs745754363 Chr.1: 54758621-54758621 on Build GRCh38
rs1038656349 Chr.1: 54758378-54758378 on Build GRCh38
rs780867581 Chr.1: 54758619-54758619 on Build GRCh38
rs768275963 Chr.1: 54758408-54758408 on Build GRCh38
rs779915944 Chr.1: 54758688-54758688 on Build GRCh38
rs768061369 Chr.1: 54758520-54758520 on Build GRCh38
rs2270004 Chr.1: 54758458-54758458 on Build GRCh38
rs768006151 Chr.1: 54758587-54758587 on Build GRCh38
rs199684359 Chr.1: 54758625-54758625 on Build GRCh38
rs575692913 Chr.1: 54758742-54758742 on Build GRCh38
rs746323251 Chr.1: 54758849-54758849 on Build GRCh38
rs145967472 Chr.1: 54758673-54758673 on Build GRCh38
rs758008147 Chr.1: 54758543-54758543 on Build GRCh38
rs1000886583 Chr.1: 54758779-54758779 on Build GRCh38
rs778023056 Chr.1: 54758826-54758826 on Build GRCh38
rs756996613 Chr.1: 54758674-54758674 on Build GRCh38
rs762025468 Chr.1: 54758643-54758643 on Build GRCh38
rs201109807 Chr.1: 54758578-54758578 on Build GRCh38
rs755605747 Chr.1: 54758772-54758772 on Build GRCh38
rs763794803 Chr.1: 54758672-54758672 on Build GRCh38
rs775325245 Chr.1: 54758624-54758624 on Build GRCh38
rs777724053 Chr.1: 54758379-54758379 on Build GRCh38
rs368556353 Chr.1: 54758409-54758409 on Build GRCh38
rs756056413 Chr.1: 54758687-54758687 on Build GRCh38
rs770467588 Chr.1: 54758852-54758852 on Build GRCh38
rs538373098 Chr.1: 54758747-54758747 on Build GRCh38
rs753147236 Chr.1: 54758767-54758767 on Build GRCh38
rs748319911 Chr.1: 54758713-54758713 on Build GRCh38
rs753658351 Chr.1: 54758814-54758814 on Build GRCh38
rs773300855 Chr.1: 54758649-54758649 on Build GRCh38
rs376605388 Chr.1: 54758605-54758605 on Build GRCh38
rs369838026 Chr.1: 54758581-54758581 on Build GRCh38
rs557179791 Chr.1: 54758830-54758830 on Build GRCh38
rs1023599710 Chr.1: 54758479-54758479 on Build GRCh38
rs377289123 Chr.1: 54758527-54758527 on Build GRCh38
rs760609265 Chr.1: 54758423-54758423 on Build GRCh38
rs780285964 Chr.1: 54758531-54758531 on Build GRCh38
rs193204977 Chr.1: 54758602-54758602 on Build GRCh38
rs376396001 Chr.1: 54758791-54758791 on Build GRCh38
rs908765565 Chr.1: 54758750-54758750 on Build GRCh38
rs538975170 Chr.1: 54758622-54758622 on Build GRCh38
rs781498443 Chr.1: 54758393-54758393 on Build GRCh38
rs146434154 Chr.1: 54758383-54758383 on Build GRCh38

View Full Product Details