Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145739248-145738649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD160
Entrez Gene ID
11126
Gene Name
CD160 molecule
Gene Aliases
BY55, NK1, NK28
UniGene
Hs.488237
Gene Chromosome Location
Chr.1: 145719312-145739288 on Build GRCh38
Transcripts
NM_007053.3 NR_103845.1

View Full Product Details