Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1992411-1992809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC727993
Entrez Gene ID
727993
Gene Name
uncharacterized LOC727993
Gene Aliases
-
UniGene
Hs.551466
Gene Chromosome Location
Chr.20: 1947210-2007517 on Build GRCh38
Transcripts
NR_134520.1 NM_001190892.1 NM_001190898.2 NM_001190899.2 NM_001190900.1 NM_024411.4

Gene Symbol
PDYN
Entrez Gene ID
5173
Gene Name
prodynorphin
Gene Aliases
ADCA, PENKB, SCA23
UniGene
Hs.22584
Gene Chromosome Location
Chr.20: 1978756-1994285 on Build GRCh38
Transcripts
NR_134520.1 NM_001190892.1 NM_001190898.2 NM_001190899.2 NM_001190900.1 NM_024411.4

SNP Detail

SNP ID Chromosome Location
rs142977649 Chr.20: 1992734-1992734 on Build GRCh38
rs947395569 Chr.20: 1992607-1992607 on Build GRCh38
rs536052604 Chr.20: 1992554-1992554 on Build GRCh38
rs964349138 Chr.20: 1992523-1992523 on Build GRCh38
rs193093593 Chr.20: 1992767-1992767 on Build GRCh38
rs375582792 Chr.20: 1992544-1992544 on Build GRCh38
rs967401538 Chr.20: 1992463-1992463 on Build GRCh38
rs981200325 Chr.20: 1992768-1992768 on Build GRCh38
rs961816204 Chr.20: 1992763-1992763 on Build GRCh38
rs541434374 Chr.20: 1992684-1992684 on Build GRCh38
rs937160775 Chr.20: 1992588-1992588 on Build GRCh38
rs1019808314 Chr.20: 1992651-1992651 on Build GRCh38
rs1027652985 Chr.20: 1992709-1992709 on Build GRCh38
rs940415889 Chr.20: 1992573-1992573 on Build GRCh38
rs552521046 Chr.20: 1992610-1992610 on Build GRCh38
rs61519908 Chr.20: 1992585-1992585 on Build GRCh38
rs1020325843 Chr.20: 1992455-1992455 on Build GRCh38
rs571761851 Chr.20: 1992545-1992545 on Build GRCh38
rs900926722 Chr.20: 1992607-1992607 on Build GRCh38
rs984492038 Chr.20: 1992737-1992737 on Build GRCh38
rs893012015 Chr.20: 1992605-1992605 on Build GRCh38
rs373426569 Chr.20: 1992612-1992612 on Build GRCh38
rs980154000 Chr.20: 1992764-1992764 on Build GRCh38
rs937171295 Chr.20: 1992779-1992779 on Build GRCh38
rs1011551159 Chr.20: 1992453-1992453 on Build GRCh38
rs879616340 Chr.20: 1992575-1992576 on Build GRCh38
rs925980775 Chr.20: 1992765-1992765 on Build GRCh38
rs903315479 Chr.20: 1992614-1992614 on Build GRCh38
rs73573508 Chr.20: 1992682-1992682 on Build GRCh38
rs1019737255 Chr.20: 1992488-1992488 on Build GRCh38
rs917371747 Chr.20: 1992724-1992724 on Build GRCh38
rs886056536 Chr.20: 1992606-1992606 on Build GRCh38
rs553501210 Chr.20: 1992537-1992537 on Build GRCh38
rs1056134165 Chr.20: 1992589-1992589 on Build GRCh38
rs1014597776 Chr.20: 1992753-1992753 on Build GRCh38
rs1008247597 Chr.20: 1992619-1992619 on Build GRCh38
rs976058185 Chr.20: 1992534-1992534 on Build GRCh38
rs554716006 Chr.20: 1992584-1992584 on Build GRCh38
rs928378992 Chr.20: 1992775-1992775 on Build GRCh38
rs543727277 Chr.20: 1992597-1992597 on Build GRCh38
rs970202342 Chr.20: 1992757-1992757 on Build GRCh38
rs868790882 Chr.20: 1992489-1992489 on Build GRCh38
rs961908461 Chr.20: 1992681-1992681 on Build GRCh38
rs1008383560 Chr.20: 1992464-1992464 on Build GRCh38
rs997001818 Chr.20: 1992700-1992700 on Build GRCh38
rs893138469 Chr.20: 1992442-1992442 on Build GRCh38
rs541987715 Chr.20: 1992615-1992615 on Build GRCh38
rs559887144 Chr.20: 1992708-1992708 on Build GRCh38
rs925949689 Chr.20: 1992576-1992576 on Build GRCh38

View Full Product Details