Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41184419-41185012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZHX3
Entrez Gene ID
23051
Gene Name
zinc fingers and homeoboxes 3
Gene Aliases
TIX1
UniGene
Hs.380133
Gene Chromosome Location
Chr.20: 41178449-41317606 on Build GRCh38
Transcripts
NM_015035.3 XM_005260341.3 XM_005260343.4 XM_006723748.3 XM_006723749.3 XM_011528701.2 XM_011528702.2 XM_011528703.2 XM_011528705.2 XM_011528706.2 XM_011528707.2 XM_011528708.2 XM_011528709.2 XM_011528711.2 XM_011528713.2 XM_011528714.2 XM_011528717.2 XM_011528718.2 XM_011528720.1 XM_017027736.1 XM_017027737.1 XM_017027738.1

SNP Detail

SNP ID Chromosome Location
rs945998118 Chr.20: 41184857-41184857 on Build GRCh38
rs566855614 Chr.20: 41184685-41184685 on Build GRCh38
rs868690930 Chr.20: 41184458-41184458 on Build GRCh38
rs551168514 Chr.20: 41184555-41184555 on Build GRCh38
rs759240981 Chr.20: 41184816-41184816 on Build GRCh38
rs535593114 Chr.20: 41184887-41184887 on Build GRCh38
rs367975521 Chr.20: 41184652-41184653 on Build GRCh38
rs1050164994 Chr.20: 41184904-41184904 on Build GRCh38
rs143338249 Chr.20: 41184503-41184503 on Build GRCh38
rs913167507 Chr.20: 41184854-41184854 on Build GRCh38
rs1042929387 Chr.20: 41184862-41184862 on Build GRCh38
rs182098267 Chr.20: 41184473-41184473 on Build GRCh38
rs1021277675 Chr.20: 41184579-41184579 on Build GRCh38
rs1015359240 Chr.20: 41184959-41184959 on Build GRCh38
rs938554641 Chr.20: 41184827-41184827 on Build GRCh38
rs919945333 Chr.20: 41184721-41184721 on Build GRCh38
rs762417532 Chr.20: 41184944-41184944 on Build GRCh38
rs531131986 Chr.20: 41184550-41184550 on Build GRCh38
rs555448324 Chr.20: 41184924-41184924 on Build GRCh38
rs927354906 Chr.20: 41184825-41184825 on Build GRCh38
rs903417207 Chr.20: 41184476-41184476 on Build GRCh38
rs754244007 Chr.20: 41184865-41184865 on Build GRCh38
rs1044476932 Chr.20: 41184493-41184493 on Build GRCh38
rs924869311 Chr.20: 41184817-41184817 on Build GRCh38
rs769415637 Chr.20: 41184480-41184480 on Build GRCh38
rs986977554 Chr.20: 41184661-41184661 on Build GRCh38
rs374969125 Chr.20: 41184756-41184756 on Build GRCh38
rs745837571 Chr.20: 41184469-41184469 on Build GRCh38
rs766192631 Chr.20: 41184611-41184611 on Build GRCh38
rs762502738 Chr.20: 41184546-41184546 on Build GRCh38
rs1028658592 Chr.20: 41184653-41184653 on Build GRCh38
rs776974739 Chr.20: 41184911-41184911 on Build GRCh38
rs1004017246 Chr.20: 41184923-41184923 on Build GRCh38
rs973368663 Chr.20: 41184687-41184687 on Build GRCh38
rs577036963 Chr.20: 41184453-41184453 on Build GRCh38
rs183600182 Chr.20: 41184962-41184962 on Build GRCh38
rs1056947225 Chr.20: 41184838-41184838 on Build GRCh38
rs890207589 Chr.20: 41184917-41184917 on Build GRCh38
rs991622567 Chr.20: 41184732-41184732 on Build GRCh38
rs571073350 Chr.20: 41184585-41184585 on Build GRCh38
rs920038519 Chr.20: 41184491-41184491 on Build GRCh38
rs950220299 Chr.20: 41184776-41184776 on Build GRCh38
rs954185291 Chr.20: 41184457-41184457 on Build GRCh38
rs931264214 Chr.20: 41184787-41184787 on Build GRCh38
rs761370594 Chr.20: 41184673-41184673 on Build GRCh38
rs1003986267 Chr.20: 41184553-41184553 on Build GRCh38
rs1029915686 Chr.20: 41184523-41184523 on Build GRCh38
rs920694032 Chr.20: 41184442-41184442 on Build GRCh38
rs746937476 Chr.20: 41184908-41184909 on Build GRCh38
rs190711701 Chr.20: 41184658-41184658 on Build GRCh38
rs995761776 Chr.20: 41184866-41184866 on Build GRCh38
rs920568011 Chr.20: 41184652-41184652 on Build GRCh38
rs6072302 Chr.20: 41184616-41184616 on Build GRCh38
rs561727461 Chr.20: 41184534-41184534 on Build GRCh38
rs1054529768 Chr.20: 41184849-41184849 on Build GRCh38
rs6093452 Chr.20: 41184573-41184573 on Build GRCh38
rs912778462 Chr.20: 41184471-41184471 on Build GRCh38
rs187794034 Chr.20: 41184492-41184492 on Build GRCh38
rs1015496616 Chr.20: 41184562-41184562 on Build GRCh38
rs528224513 Chr.20: 41184495-41184495 on Build GRCh38
rs1019795256 Chr.20: 41184960-41184960 on Build GRCh38
rs767159731 Chr.20: 41184752-41184752 on Build GRCh38
rs980190988 Chr.20: 41184788-41184788 on Build GRCh38

View Full Product Details