Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60644677-60645152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIX1
Entrez Gene ID
6495
Gene Name
SIX homeobox 1
Gene Aliases
BOS3, DFNA23, TIP39
UniGene
Hs.633506
Gene Chromosome Location
Chr.14: 60644698-60649489 on Build GRCh38
Transcripts
NM_005982.3 XM_017021602.1

SNP Detail

SNP ID Chromosome Location
rs1049888091 Chr.14: 60645117-60645117 on Build GRCh38
rs907252831 Chr.14: 60644722-60644722 on Build GRCh38
rs946413885 Chr.14: 60644768-60644768 on Build GRCh38
rs887064797 Chr.14: 60644848-60644848 on Build GRCh38
rs61993831 Chr.14: 60644787-60644787 on Build GRCh38
rs137994957 Chr.14: 60644907-60644907 on Build GRCh38
rs886050567 Chr.14: 60644712-60644712 on Build GRCh38
rs996987391 Chr.14: 60644808-60644808 on Build GRCh38
rs146967331 Chr.14: 60644805-60644805 on Build GRCh38
rs982122567 Chr.14: 60644964-60644964 on Build GRCh38
rs564608572 Chr.14: 60644705-60644705 on Build GRCh38
rs938722386 Chr.14: 60645122-60645122 on Build GRCh38
rs1028056922 Chr.14: 60644977-60644977 on Build GRCh38
rs1008554191 Chr.14: 60644802-60644802 on Build GRCh38
rs547309317 Chr.14: 60644765-60644765 on Build GRCh38
rs550089904 Chr.14: 60644945-60644945 on Build GRCh38
rs779226095 Chr.14: 60645000-60645000 on Build GRCh38
rs558532789 Chr.14: 60645051-60645051 on Build GRCh38
rs3832952 Chr.14: 60645012-60645013 on Build GRCh38
rs1042111674 Chr.14: 60644720-60644720 on Build GRCh38
rs1005556303 Chr.14: 60644939-60644939 on Build GRCh38
rs536496013 Chr.14: 60644944-60644944 on Build GRCh38
rs888656744 Chr.14: 60645125-60645125 on Build GRCh38
rs990659039 Chr.14: 60645016-60645016 on Build GRCh38
rs140088005 Chr.14: 60644760-60644760 on Build GRCh38
rs995368945 Chr.14: 60644800-60644800 on Build GRCh38
rs914785412 Chr.14: 60645043-60645043 on Build GRCh38
rs965132403 Chr.14: 60644936-60644936 on Build GRCh38
rs1042563695 Chr.14: 60645034-60645034 on Build GRCh38
rs746385772 Chr.14: 60645032-60645032 on Build GRCh38
rs997021988 Chr.14: 60645097-60645100 on Build GRCh38
rs780378618 Chr.14: 60645087-60645087 on Build GRCh38
rs1040014700 Chr.14: 60645086-60645086 on Build GRCh38
rs1055614826 Chr.14: 60644775-60644775 on Build GRCh38
rs757802086 Chr.14: 60644999-60644999 on Build GRCh38
rs1003844969 Chr.14: 60644891-60644891 on Build GRCh38
rs901405689 Chr.14: 60644806-60644806 on Build GRCh38
rs915036970 Chr.14: 60645018-60645018 on Build GRCh38
rs569954956 Chr.14: 60644767-60644767 on Build GRCh38
rs952808390 Chr.14: 60644934-60644934 on Build GRCh38
rs181833190 Chr.14: 60645068-60645068 on Build GRCh38
rs1054948020 Chr.14: 60644713-60644713 on Build GRCh38

View Full Product Details