Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57213607-57214108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
UniGene
Hs.460885
Gene Chromosome Location
Chr.16: 57186284-57240475 on Build GRCh38
Transcripts
NM_001305163.1 NM_001305164.1 NM_133368.2 XM_005256220.1 XM_011523427.1 XM_011523428.1 XM_011523430.1 XM_017023844.1

SNP Detail

SNP ID Chromosome Location
rs773809073 Chr.16: 57213866-57213866 on Build GRCh38
rs1013424916 Chr.16: 57213869-57213869 on Build GRCh38
rs560867821 Chr.16: 57213674-57213674 on Build GRCh38
rs772260362 Chr.16: 57214004-57214004 on Build GRCh38
rs990409668 Chr.16: 57213999-57213999 on Build GRCh38
rs756094594 Chr.16: 57213937-57213937 on Build GRCh38
rs770286447 Chr.16: 57213759-57213759 on Build GRCh38
rs773471644 Chr.16: 57213989-57213989 on Build GRCh38
rs372798463 Chr.16: 57213867-57213867 on Build GRCh38
rs551619118 Chr.16: 57213791-57213791 on Build GRCh38
rs750331517 Chr.16: 57213934-57213934 on Build GRCh38
rs780710129 Chr.16: 57213932-57213932 on Build GRCh38
rs192207391 Chr.16: 57213787-57213787 on Build GRCh38
rs550565184 Chr.16: 57213755-57213755 on Build GRCh38
rs766826496 Chr.16: 57213868-57213868 on Build GRCh38
rs771628498 Chr.16: 57213998-57213998 on Build GRCh38
rs768344573 Chr.16: 57213974-57213974 on Build GRCh38
rs754282738 Chr.16: 57213878-57213878 on Build GRCh38
rs1050467385 Chr.16: 57213747-57213747 on Build GRCh38
rs200599144 Chr.16: 57213978-57213978 on Build GRCh38
rs11859493 Chr.16: 57213742-57213742 on Build GRCh38
rs779143074 Chr.16: 57213975-57213975 on Build GRCh38
rs527894977 Chr.16: 57213962-57213962 on Build GRCh38
rs567963069 Chr.16: 57214034-57214034 on Build GRCh38
rs184466087 Chr.16: 57213949-57213949 on Build GRCh38
rs756740517 Chr.16: 57213915-57213915 on Build GRCh38
rs918711 Chr.16: 57213664-57213664 on Build GRCh38
rs774733900 Chr.16: 57213807-57213807 on Build GRCh38
rs763735470 Chr.16: 57213882-57213882 on Build GRCh38
rs373754710 Chr.16: 57213904-57213904 on Build GRCh38
rs932065005 Chr.16: 57214063-57214063 on Build GRCh38
rs760737747 Chr.16: 57214075-57214075 on Build GRCh38
rs201221888 Chr.16: 57213966-57213969 on Build GRCh38
rs901943195 Chr.16: 57213722-57213722 on Build GRCh38
rs980823410 Chr.16: 57213940-57213940 on Build GRCh38
rs374269118 Chr.16: 57213831-57213831 on Build GRCh38
rs369588365 Chr.16: 57213879-57213879 on Build GRCh38
rs946252816 Chr.16: 57213637-57213637 on Build GRCh38
rs556183896 Chr.16: 57214053-57214053 on Build GRCh38
rs946180569 Chr.16: 57214025-57214025 on Build GRCh38
rs924956688 Chr.16: 57213952-57213952 on Build GRCh38
rs1024698191 Chr.16: 57213891-57213891 on Build GRCh38
rs138413108 Chr.16: 57214069-57214069 on Build GRCh38
rs538583622 Chr.16: 57214042-57214042 on Build GRCh38
rs913525393 Chr.16: 57214024-57214024 on Build GRCh38
rs761248057 Chr.16: 57213995-57213995 on Build GRCh38
rs533458087 Chr.16: 57213789-57213789 on Build GRCh38
rs759793278 Chr.16: 57213920-57213920 on Build GRCh38
rs8048889 Chr.16: 57214082-57214082 on Build GRCh38
rs772241594 Chr.16: 57213985-57213985 on Build GRCh38

View Full Product Details