Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 41130164-41130664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD6
Entrez Gene ID
89890
Gene Name
kelch repeat and BTB domain containing 6
Gene Aliases
-
UniGene
Hs.534040
Gene Chromosome Location
Chr.13: 41127573-41132800 on Build GRCh38
Transcripts
NM_152903.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694124 c.1903_1905delGAA p.E635delE Chr.13: 41130607-41130609 on Build GRCh38
3813809 c.1891A>G p.T631A Chr.13: 41130621-41130621 on Build GRCh38
696570 c.2024G>A p.*675* Chr.13: 41130488-41130488 on Build GRCh38
3813808 c.1945T>A p.F649I Chr.13: 41130567-41130567 on Build GRCh38
1211380 c.2009G>A p.R670H Chr.13: 41130503-41130503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767305811 Chr.13: 41130550-41130551 on Build GRCh38
rs969247833 Chr.13: 41130464-41130464 on Build GRCh38
rs182018655 Chr.13: 41130331-41130331 on Build GRCh38
rs1013536137 Chr.13: 41130423-41130423 on Build GRCh38
rs556151277 Chr.13: 41130341-41130341 on Build GRCh38
rs900239611 Chr.13: 41130307-41130307 on Build GRCh38
rs778271886 Chr.13: 41130491-41130491 on Build GRCh38
rs576956897 Chr.13: 41130330-41130330 on Build GRCh38
rs747037725 Chr.13: 41130426-41130426 on Build GRCh38
rs752901799 Chr.13: 41130463-41130463 on Build GRCh38
rs968384718 Chr.13: 41130347-41130347 on Build GRCh38
rs1007679597 Chr.13: 41130275-41130275 on Build GRCh38
rs748881545 Chr.13: 41130565-41130565 on Build GRCh38
rs931723099 Chr.13: 41130604-41130604 on Build GRCh38
rs148227219 Chr.13: 41130351-41130351 on Build GRCh38
rs1051953376 Chr.13: 41130199-41130199 on Build GRCh38
rs186354848 Chr.13: 41130347-41130347 on Build GRCh38
rs554505812 Chr.13: 41130319-41130319 on Build GRCh38
rs886133065 Chr.13: 41130255-41130255 on Build GRCh38
rs202193213 Chr.13: 41130551-41130551 on Build GRCh38
rs752708701 Chr.13: 41130607-41130609 on Build GRCh38
rs925367335 Chr.13: 41130482-41130482 on Build GRCh38
rs980620056 Chr.13: 41130469-41130469 on Build GRCh38
rs995586049 Chr.13: 41130387-41130387 on Build GRCh38
rs773161485 Chr.13: 41130641-41130641 on Build GRCh38
rs1040136175 Chr.13: 41130283-41130283 on Build GRCh38
rs943543478 Chr.13: 41130547-41130547 on Build GRCh38
rs768496417 Chr.13: 41130591-41130591 on Build GRCh38
rs41286993 Chr.13: 41130523-41130523 on Build GRCh38
rs1040003164 Chr.13: 41130534-41130534 on Build GRCh38
rs747622286 Chr.13: 41130496-41130496 on Build GRCh38
rs778280667 Chr.13: 41130278-41130278 on Build GRCh38
rs541876155 Chr.13: 41130354-41130354 on Build GRCh38
rs779310444 Chr.13: 41130460-41130460 on Build GRCh38
rs960737761 Chr.13: 41130368-41130368 on Build GRCh38
rs889307166 Chr.13: 41130268-41130268 on Build GRCh38
rs951291151 Chr.13: 41130404-41130404 on Build GRCh38
rs767379342 Chr.13: 41130620-41130620 on Build GRCh38
rs1047395685 Chr.13: 41130246-41130249 on Build GRCh38
rs910797037 Chr.13: 41130542-41130542 on Build GRCh38
rs745388139 Chr.13: 41130527-41130527 on Build GRCh38
rs1024147758 Chr.13: 41130405-41130405 on Build GRCh38
rs1037836716 Chr.13: 41130635-41130635 on Build GRCh38
rs188355025 Chr.13: 41130386-41130386 on Build GRCh38
rs753840584 Chr.13: 41130437-41130437 on Build GRCh38
rs774128638 Chr.13: 41130603-41130603 on Build GRCh38
rs758849529 Chr.13: 41130475-41130475 on Build GRCh38
rs1052007254 Chr.13: 41130533-41130533 on Build GRCh38
rs572184493 Chr.13: 41130435-41130435 on Build GRCh38
rs755037173 Chr.13: 41130456-41130456 on Build GRCh38
rs1025287026 Chr.13: 41130402-41130402 on Build GRCh38
rs148761059 Chr.13: 41130529-41130529 on Build GRCh38
rs933549531 Chr.13: 41130187-41130187 on Build GRCh38
rs374738937 Chr.13: 41130521-41130521 on Build GRCh38

View Full Product Details