Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17580695-17581289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLGALT1
Entrez Gene ID
79709
Gene Name
collagen beta(1-O)galactosyltransferase 1
Gene Aliases
GLT25D1
UniGene
Hs.418795
Gene Chromosome Location
Chr.19: 17555594-17583162 on Build GRCh38
Transcripts
NM_024656.3 XM_005260080.4 XM_011528297.2 XM_011528298.2 XM_011528299.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079331 c.1447C>T p.R483C Chr.19: 17580751-17580751 on Build GRCh38
7106272 c.1629C>G p.L543L Chr.19: 17581204-17581204 on Build GRCh38
6633907 c.1670_1671insC p.T559fs*12 Chr.19: 17581245-17581246 on Build GRCh38
4929547 c.1444C>T p.P482S Chr.19: 17580748-17580748 on Build GRCh38
7129983 c.1654G>A p.E552K Chr.19: 17581229-17581229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992449032 Chr.19: 17581074-17581074 on Build GRCh38
rs769807437 Chr.19: 17581233-17581233 on Build GRCh38
rs561924033 Chr.19: 17581038-17581038 on Build GRCh38
rs781164776 Chr.19: 17580754-17580754 on Build GRCh38
rs200996348 Chr.19: 17580774-17580774 on Build GRCh38
rs74461907 Chr.19: 17580846-17580846 on Build GRCh38
rs377243848 Chr.19: 17581170-17581170 on Build GRCh38
rs145786853 Chr.19: 17580831-17580831 on Build GRCh38
rs202137060 Chr.19: 17580804-17580804 on Build GRCh38
rs778226768 Chr.19: 17581201-17581201 on Build GRCh38
rs149240687 Chr.19: 17581206-17581206 on Build GRCh38
rs539420031 Chr.19: 17580982-17580982 on Build GRCh38
rs137941571 Chr.19: 17580887-17580887 on Build GRCh38
rs1002320023 Chr.19: 17581248-17581248 on Build GRCh38
rs374537382 Chr.19: 17580943-17580943 on Build GRCh38
rs995061792 Chr.19: 17581019-17581019 on Build GRCh38
rs370199311 Chr.19: 17581172-17581172 on Build GRCh38
rs143412269 Chr.19: 17580798-17580798 on Build GRCh38
rs941514661 Chr.19: 17581116-17581116 on Build GRCh38
rs776878605 Chr.19: 17581068-17581068 on Build GRCh38
rs745389689 Chr.19: 17580786-17580786 on Build GRCh38
rs756336332 Chr.19: 17580751-17580751 on Build GRCh38
rs890486038 Chr.19: 17580926-17580926 on Build GRCh38
rs752557537 Chr.19: 17581160-17581162 on Build GRCh38
rs759507081 Chr.19: 17580724-17580725 on Build GRCh38
rs543254139 Chr.19: 17580892-17580892 on Build GRCh38
rs1017424368 Chr.19: 17581001-17581001 on Build GRCh38
rs778450738 Chr.19: 17581127-17581127 on Build GRCh38
rs758726918 Chr.19: 17581030-17581030 on Build GRCh38
rs377077306 Chr.19: 17580792-17580792 on Build GRCh38
rs60227951 Chr.19: 17581108-17581108 on Build GRCh38
rs765453006 Chr.19: 17581169-17581169 on Build GRCh38
rs762999806 Chr.19: 17581159-17581159 on Build GRCh38
rs747576813 Chr.19: 17581140-17581140 on Build GRCh38
rs368380442 Chr.19: 17580820-17580820 on Build GRCh38
rs1008827364 Chr.19: 17581000-17581002 on Build GRCh38
rs765383580 Chr.19: 17580897-17580897 on Build GRCh38
rs775767986 Chr.19: 17580730-17580730 on Build GRCh38
rs778596520 Chr.19: 17581180-17581180 on Build GRCh38
rs368315240 Chr.19: 17581012-17581012 on Build GRCh38
rs544290890 Chr.19: 17581090-17581090 on Build GRCh38
rs763410653 Chr.19: 17580734-17580734 on Build GRCh38
rs528025663 Chr.19: 17581095-17581097 on Build GRCh38
rs771357583 Chr.19: 17581155-17581155 on Build GRCh38
rs766255783 Chr.19: 17581174-17581174 on Build GRCh38
rs372863165 Chr.19: 17580891-17580891 on Build GRCh38
rs73524017 Chr.19: 17581055-17581055 on Build GRCh38
rs750227448 Chr.19: 17581173-17581173 on Build GRCh38
rs183198845 Chr.19: 17580823-17580823 on Build GRCh38
rs745941882 Chr.19: 17581215-17581215 on Build GRCh38
rs768313525 Chr.19: 17580954-17580954 on Build GRCh38
rs138362382 Chr.19: 17581205-17581205 on Build GRCh38
rs954603207 Chr.19: 17580985-17580985 on Build GRCh38
rs757122399 Chr.19: 17580841-17580841 on Build GRCh38
rs745950389 Chr.19: 17581085-17581085 on Build GRCh38
rs1034047904 Chr.19: 17581060-17581060 on Build GRCh38
rs199904709 Chr.19: 17580741-17580741 on Build GRCh38
rs149223760 Chr.19: 17580743-17580743 on Build GRCh38
rs759569654 Chr.19: 17581166-17581166 on Build GRCh38
rs140576945 Chr.19: 17580829-17580829 on Build GRCh38
rs200469379 Chr.19: 17580780-17580780 on Build GRCh38
rs927639392 Chr.19: 17581071-17581072 on Build GRCh38
rs939564653 Chr.19: 17581073-17581073 on Build GRCh38
rs764300831 Chr.19: 17581240-17581240 on Build GRCh38
rs537124242 Chr.19: 17580883-17580883 on Build GRCh38
rs377549471 Chr.19: 17580749-17580749 on Build GRCh38
rs779565570 Chr.19: 17580858-17580858 on Build GRCh38
rs73524018 Chr.19: 17581129-17581129 on Build GRCh38
rs534491656 Chr.19: 17580854-17580854 on Build GRCh38
rs989356896 Chr.19: 17581036-17581036 on Build GRCh38
rs758262050 Chr.19: 17580758-17580758 on Build GRCh38
rs200984516 Chr.19: 17580882-17580882 on Build GRCh38
rs750404892 Chr.19: 17580931-17580931 on Build GRCh38
rs954634522 Chr.19: 17580778-17580778 on Build GRCh38
rs770416761 Chr.19: 17580775-17580775 on Build GRCh38
rs753338332 Chr.19: 17581175-17581175 on Build GRCh38
rs370763953 Chr.19: 17580753-17580753 on Build GRCh38
rs751327244 Chr.19: 17580837-17580837 on Build GRCh38
rs944728387 Chr.19: 17581189-17581189 on Build GRCh38
rs776707399 Chr.19: 17581156-17581156 on Build GRCh38
rs565969520 Chr.19: 17580849-17580849 on Build GRCh38
rs1038559610 Chr.19: 17581118-17581118 on Build GRCh38
rs759748599 Chr.19: 17580896-17580896 on Build GRCh38
rs891184 Chr.19: 17580942-17580942 on Build GRCh38
rs199522734 Chr.19: 17581128-17581128 on Build GRCh38
rs748819009 Chr.19: 17580833-17580833 on Build GRCh38
rs897229097 Chr.19: 17581124-17581124 on Build GRCh38
rs895305181 Chr.19: 17580874-17580874 on Build GRCh38
rs201380344 Chr.19: 17580824-17580824 on Build GRCh38
rs370733358 Chr.19: 17581267-17581267 on Build GRCh38
rs771669705 Chr.19: 17581004-17581004 on Build GRCh38
rs1008789509 Chr.19: 17581167-17581167 on Build GRCh38
rs372306441 Chr.19: 17580801-17580801 on Build GRCh38
rs201797838 Chr.19: 17580766-17580766 on Build GRCh38
rs557619299 Chr.19: 17581021-17581021 on Build GRCh38
rs925798899 Chr.19: 17581134-17581134 on Build GRCh38
rs142410173 Chr.19: 17581196-17581196 on Build GRCh38
rs764463626 Chr.19: 17580735-17580735 on Build GRCh38
rs961695783 Chr.19: 17580720-17580720 on Build GRCh38
rs754493620 Chr.19: 17581179-17581179 on Build GRCh38
rs752144490 Chr.19: 17580736-17580736 on Build GRCh38
rs533405537 Chr.19: 17580835-17580835 on Build GRCh38
rs554821327 Chr.19: 17580870-17580870 on Build GRCh38
rs200075346 Chr.19: 17580827-17580827 on Build GRCh38
rs142585342 Chr.19: 17581234-17581234 on Build GRCh38
rs144502149 Chr.19: 17580988-17580988 on Build GRCh38
rs780452836 Chr.19: 17580752-17580752 on Build GRCh38
rs750628149 Chr.19: 17581024-17581024 on Build GRCh38
rs763026423 Chr.19: 17581162-17581165 on Build GRCh38
rs371051742 Chr.19: 17580947-17580947 on Build GRCh38
rs769398603 Chr.19: 17580793-17580793 on Build GRCh38
rs573495169 Chr.19: 17580996-17580996 on Build GRCh38
rs777542070 Chr.19: 17580772-17580772 on Build GRCh38
rs774707314 Chr.19: 17580881-17580881 on Build GRCh38
rs1008892446 Chr.19: 17580940-17580940 on Build GRCh38
rs757069012 Chr.19: 17580915-17580915 on Build GRCh38
rs781090013 Chr.19: 17580920-17580920 on Build GRCh38
rs777029758 Chr.19: 17581210-17581210 on Build GRCh38
rs751318338 Chr.19: 17580903-17580903 on Build GRCh38
rs374956374 Chr.19: 17580895-17580895 on Build GRCh38
rs201511712 Chr.19: 17581254-17581254 on Build GRCh38
rs768144953 Chr.19: 17580876-17580876 on Build GRCh38
rs960376578 Chr.19: 17581232-17581232 on Build GRCh38
rs758823019 Chr.19: 17581197-17581197 on Build GRCh38
rs764898652 Chr.19: 17580937-17580937 on Build GRCh38
rs781675245 Chr.19: 17581249-17581249 on Build GRCh38
rs145475968 Chr.19: 17581025-17581025 on Build GRCh38
rs748749387 Chr.19: 17580952-17580952 on Build GRCh38
rs759245476 Chr.19: 17580724-17580724 on Build GRCh38
rs766548989 Chr.19: 17580888-17580888 on Build GRCh38
rs151321150 Chr.19: 17580816-17580816 on Build GRCh38

View Full Product Details