Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 106652204-106652553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QRSL1
Entrez Gene ID
55278
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
GatA
UniGene
Hs.406917
Gene Chromosome Location
Chr.6: 106629566-106668417 on Build GRCh38
Transcripts
NM_018292.4 XM_011535924.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
200596 c.677C>T p.S226L Chr.6: 106652328-106652328 on Build GRCh38
6761011 c.766A>G p.T256A Chr.6: 106652499-106652499 on Build GRCh38
6761018 c.653A>G p.H218R Chr.6: 106652304-106652304 on Build GRCh38
6761019 c.653A>G p.H218R Chr.6: 106652304-106652304 on Build GRCh38
6761012 c.766A>G p.T256A Chr.6: 106652499-106652499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899120471 Chr.6: 106652320-106652320 on Build GRCh38
rs746900758 Chr.6: 106652418-106652418 on Build GRCh38
rs144191952 Chr.6: 106652345-106652345 on Build GRCh38
rs145214492 Chr.6: 106652466-106652466 on Build GRCh38
rs748164760 Chr.6: 106652267-106652267 on Build GRCh38
rs765013402 Chr.6: 106652390-106652391 on Build GRCh38
rs779838879 Chr.6: 106652393-106652393 on Build GRCh38
rs768131259 Chr.6: 106652302-106652302 on Build GRCh38
rs1023817489 Chr.6: 106652497-106652497 on Build GRCh38
rs146391577 Chr.6: 106652333-106652333 on Build GRCh38
rs774830851 Chr.6: 106652299-106652299 on Build GRCh38
rs780416940 Chr.6: 106652532-106652532 on Build GRCh38
rs776177177 Chr.6: 106652486-106652486 on Build GRCh38
rs754906372 Chr.6: 106652423-106652423 on Build GRCh38
rs766160251 Chr.6: 106652506-106652506 on Build GRCh38
rs758301427 Chr.6: 106652250-106652250 on Build GRCh38
rs112246044 Chr.6: 106652230-106652230 on Build GRCh38
rs752524551 Chr.6: 106652485-106652485 on Build GRCh38
rs201077509 Chr.6: 106652300-106652300 on Build GRCh38
rs779144840 Chr.6: 106652525-106652525 on Build GRCh38
rs929280958 Chr.6: 106652451-106652451 on Build GRCh38
rs199973725 Chr.6: 106652239-106652239 on Build GRCh38
rs773172962 Chr.6: 106652279-106652279 on Build GRCh38
rs750214959 Chr.6: 106652356-106652356 on Build GRCh38
rs1019736662 Chr.6: 106652452-106652452 on Build GRCh38
rs776120058 Chr.6: 106652309-106652309 on Build GRCh38
rs112965704 Chr.6: 106652329-106652329 on Build GRCh38
rs142361530 Chr.6: 106652510-106652510 on Build GRCh38
rs772505817 Chr.6: 106652464-106652464 on Build GRCh38
rs768184246 Chr.6: 106652236-106652236 on Build GRCh38
rs778638364 Chr.6: 106652241-106652241 on Build GRCh38
rs539588728 Chr.6: 106652280-106652280 on Build GRCh38
rs991490206 Chr.6: 106652496-106652496 on Build GRCh38
rs144555270 Chr.6: 106652337-106652337 on Build GRCh38
rs746018325 Chr.6: 106652531-106652531 on Build GRCh38
rs761379461 Chr.6: 106652489-106652489 on Build GRCh38
rs777865528 Chr.6: 106652426-106652426 on Build GRCh38
rs745712186 Chr.6: 106652247-106652247 on Build GRCh38
rs986259546 Chr.6: 106652246-106652246 on Build GRCh38
rs201592456 Chr.6: 106652478-106652478 on Build GRCh38
rs771303669 Chr.6: 106652295-106652295 on Build GRCh38
rs781434746 Chr.6: 106652507-106652507 on Build GRCh38
rs759025160 Chr.6: 106652235-106652235 on Build GRCh38
rs763639020 Chr.6: 106652238-106652238 on Build GRCh38
rs771244109 Chr.6: 106652434-106652434 on Build GRCh38
rs34221917 Chr.6: 106652521-106652521 on Build GRCh38
rs554306734 Chr.6: 106652468-106652468 on Build GRCh38
rs749380189 Chr.6: 106652430-106652430 on Build GRCh38
rs761098365 Chr.6: 106652328-106652328 on Build GRCh38
rs764858668 Chr.6: 106652492-106652492 on Build GRCh38
rs761204018 Chr.6: 106652477-106652477 on Build GRCh38
rs774777570 Chr.6: 106652440-106652440 on Build GRCh38
rs754924939 Chr.6: 106652509-106652509 on Build GRCh38
rs146630103 Chr.6: 106652522-106652522 on Build GRCh38
rs902589186 Chr.6: 106652469-106652469 on Build GRCh38
rs148734046 Chr.6: 106652494-106652494 on Build GRCh38
rs758250242 Chr.6: 106652360-106652360 on Build GRCh38
rs769857680 Chr.6: 106652269-106652269 on Build GRCh38
rs369873738 Chr.6: 106652237-106652237 on Build GRCh38
rs780034902 Chr.6: 106652261-106652261 on Build GRCh38
rs1037765160 Chr.6: 106652301-106652301 on Build GRCh38
rs746217672 Chr.6: 106652460-106652460 on Build GRCh38
rs542432643 Chr.6: 106652263-106652263 on Build GRCh38
rs754881125 Chr.6: 106652251-106652251 on Build GRCh38

View Full Product Details