Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27822082-27822676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PBK
Entrez Gene ID
55872
Gene Name
PDZ binding kinase
Gene Aliases
CT84, HEL164, Nori-3, SPK, TOPK
UniGene
Hs.104741
Gene Chromosome Location
Chr.8: 27809621-27838095 on Build GRCh38
Transcripts
NM_001278945.1 NM_018492.3 XM_006716368.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5880657 c.343A>T p.M115L Chr.8: 27822441-27822441 on Build GRCh38
3982494 c.320A>G p.N107S Chr.8: 27822464-27822464 on Build GRCh38
5874489 c.439G>A p.A147T Chr.8: 27822345-27822345 on Build GRCh38
6425931 c.465G>T p.K155N Chr.8: 27822319-27822319 on Build GRCh38
5899564 c.365C>A p.S122Y Chr.8: 27822419-27822419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746900679 Chr.8: 27822482-27822482 on Build GRCh38
rs1055482374 Chr.8: 27822378-27822378 on Build GRCh38
rs1001913357 Chr.8: 27822195-27822195 on Build GRCh38
rs756387355 Chr.8: 27822458-27822458 on Build GRCh38
rs112967316 Chr.8: 27822577-27822577 on Build GRCh38
rs779729707 Chr.8: 27822478-27822478 on Build GRCh38
rs776301878 Chr.8: 27822293-27822293 on Build GRCh38
rs1011738371 Chr.8: 27822154-27822154 on Build GRCh38
rs780348633 Chr.8: 27822138-27822138 on Build GRCh38
rs766921897 Chr.8: 27822348-27822348 on Build GRCh38
rs1675236 Chr.8: 27822433-27822433 on Build GRCh38
rs746944771 Chr.8: 27822423-27822423 on Build GRCh38
rs977297231 Chr.8: 27822221-27822221 on Build GRCh38
rs373581110 Chr.8: 27822538-27822538 on Build GRCh38
rs372560974 Chr.8: 27822288-27822288 on Build GRCh38
rs935626169 Chr.8: 27822349-27822349 on Build GRCh38
rs896129878 Chr.8: 27822213-27822213 on Build GRCh38
rs111722078 Chr.8: 27822559-27822559 on Build GRCh38
rs774590430 Chr.8: 27822287-27822288 on Build GRCh38
rs773544696 Chr.8: 27822438-27822438 on Build GRCh38
rs569862657 Chr.8: 27822141-27822141 on Build GRCh38
rs747834390 Chr.8: 27822430-27822430 on Build GRCh38
rs758222396 Chr.8: 27822465-27822465 on Build GRCh38
rs755045401 Chr.8: 27822360-27822360 on Build GRCh38
rs190358801 Chr.8: 27822209-27822209 on Build GRCh38
rs766760760 Chr.8: 27822441-27822441 on Build GRCh38
rs771226147 Chr.8: 27822523-27822523 on Build GRCh38
rs1018289758 Chr.8: 27822572-27822572 on Build GRCh38
rs761388469 Chr.8: 27822305-27822305 on Build GRCh38
rs556489077 Chr.8: 27822332-27822332 on Build GRCh38
rs950636684 Chr.8: 27822263-27822263 on Build GRCh38
rs1025959208 Chr.8: 27822621-27822621 on Build GRCh38
rs768085381 Chr.8: 27822292-27822292 on Build GRCh38
rs182671594 Chr.8: 27822244-27822244 on Build GRCh38
rs1013767934 Chr.8: 27822602-27822602 on Build GRCh38
rs906047817 Chr.8: 27822187-27822187 on Build GRCh38
rs367671591 Chr.8: 27822495-27822495 on Build GRCh38
rs185478648 Chr.8: 27822428-27822428 on Build GRCh38
rs532374162 Chr.8: 27822585-27822585 on Build GRCh38
rs769516368 Chr.8: 27822257-27822257 on Build GRCh38
rs892071445 Chr.8: 27822129-27822129 on Build GRCh38
rs780020966 Chr.8: 27822419-27822419 on Build GRCh38
rs768760254 Chr.8: 27822445-27822445 on Build GRCh38
rs772196242 Chr.8: 27822487-27822487 on Build GRCh38
rs752783878 Chr.8: 27822451-27822451 on Build GRCh38
rs3779620 Chr.8: 27822464-27822464 on Build GRCh38
rs911259120 Chr.8: 27822188-27822188 on Build GRCh38
rs756407655 Chr.8: 27822367-27822367 on Build GRCh38
rs769494518 Chr.8: 27822436-27822436 on Build GRCh38
rs369819863 Chr.8: 27822183-27822183 on Build GRCh38
rs763346850 Chr.8: 27822440-27822440 on Build GRCh38
rs371839165 Chr.8: 27822502-27822502 on Build GRCh38
rs767514634 Chr.8: 27822363-27822363 on Build GRCh38
rs765520589 Chr.8: 27822329-27822329 on Build GRCh38
rs748961466 Chr.8: 27822521-27822521 on Build GRCh38
rs778842443 Chr.8: 27822370-27822370 on Build GRCh38
rs764456498 Chr.8: 27822463-27822463 on Build GRCh38
rs373579758 Chr.8: 27822449-27822449 on Build GRCh38
rs368908110 Chr.8: 27822267-27822267 on Build GRCh38
rs142068654 Chr.8: 27822447-27822447 on Build GRCh38
rs896258381 Chr.8: 27822587-27822587 on Build GRCh38
rs201842147 Chr.8: 27822404-27822404 on Build GRCh38
rs767635561 Chr.8: 27822431-27822457 on Build GRCh38
rs368375216 Chr.8: 27822528-27822528 on Build GRCh38
rs942819788 Chr.8: 27822211-27822211 on Build GRCh38
rs952000990 Chr.8: 27822532-27822532 on Build GRCh38
rs769406505 Chr.8: 27822507-27822507 on Build GRCh38
rs371557868 Chr.8: 27822395-27822395 on Build GRCh38
rs761978341 Chr.8: 27822393-27822394 on Build GRCh38
rs922803162 Chr.8: 27822235-27822235 on Build GRCh38
rs777364218 Chr.8: 27822519-27822519 on Build GRCh38
rs988856138 Chr.8: 27822474-27822474 on Build GRCh38
rs552397122 Chr.8: 27822636-27822636 on Build GRCh38
rs984692566 Chr.8: 27822374-27822374 on Build GRCh38
rs1033434789 Chr.8: 27822201-27822201 on Build GRCh38
rs752904921 Chr.8: 27822366-27822366 on Build GRCh38
rs774582841 Chr.8: 27822526-27822526 on Build GRCh38
rs750474138 Chr.8: 27822505-27822506 on Build GRCh38
rs751562351 Chr.8: 27822356-27822356 on Build GRCh38
rs754938337 Chr.8: 27822483-27822483 on Build GRCh38
rs993409349 Chr.8: 27822486-27822486 on Build GRCh38
rs763438217 Chr.8: 27822344-27822344 on Build GRCh38
rs946284105 Chr.8: 27822240-27822240 on Build GRCh38
rs1016012680 Chr.8: 27822375-27822375 on Build GRCh38
rs553961952 Chr.8: 27822127-27822127 on Build GRCh38
rs984149048 Chr.8: 27822172-27822172 on Build GRCh38
rs745711431 Chr.8: 27822396-27822396 on Build GRCh38
rs768158056 Chr.8: 27822427-27822427 on Build GRCh38
rs189891155 Chr.8: 27822576-27822576 on Build GRCh38

View Full Product Details