Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36052074-36052587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RECK
Entrez Gene ID
8434
Gene Name
reversion inducing cysteine rich protein with kazal motifs
Gene Aliases
ST15
UniGene
Hs.388918
Gene Chromosome Location
Chr.9: 36036905-36124455 on Build GRCh38
Transcripts
NM_001316345.1 NM_001316346.1 NM_001316347.1 NM_001316348.1 NM_021111.2 XM_017015207.1 XM_017015208.1 XM_017015209.1 XM_017015210.1 XM_017015211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6395447 c.145G>T p.D49Y Chr.9: 36052309-36052309 on Build GRCh38
5624738 c.143G>C p.R48P Chr.9: 36052307-36052307 on Build GRCh38
6764656 c.142C>T p.R48C Chr.9: 36052306-36052306 on Build GRCh38
455938 c.147T>C p.D49D Chr.9: 36052311-36052311 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775570123 Chr.9: 36052271-36052271 on Build GRCh38
rs202091867 Chr.9: 36052264-36052264 on Build GRCh38
rs985382558 Chr.9: 36052414-36052414 on Build GRCh38
rs912393222 Chr.9: 36052161-36052166 on Build GRCh38
rs35451547 Chr.9: 36052105-36052106 on Build GRCh38
rs763846920 Chr.9: 36052350-36052350 on Build GRCh38
rs573511010 Chr.9: 36052298-36052298 on Build GRCh38
rs745807897 Chr.9: 36052370-36052370 on Build GRCh38
rs755557754 Chr.9: 36052334-36052334 on Build GRCh38
rs753427541 Chr.9: 36052352-36052352 on Build GRCh38
rs929403629 Chr.9: 36052534-36052534 on Build GRCh38
rs1039166931 Chr.9: 36052395-36052395 on Build GRCh38
rs768693771 Chr.9: 36052440-36052440 on Build GRCh38
rs746102152 Chr.9: 36052224-36052224 on Build GRCh38
rs368592958 Chr.9: 36052401-36052401 on Build GRCh38
rs1052011594 Chr.9: 36052431-36052431 on Build GRCh38
rs972568365 Chr.9: 36052430-36052430 on Build GRCh38
rs577911752 Chr.9: 36052209-36052209 on Build GRCh38
rs929265094 Chr.9: 36052196-36052196 on Build GRCh38
rs570225508 Chr.9: 36052549-36052549 on Build GRCh38
rs768877933 Chr.9: 36052292-36052292 on Build GRCh38
rs774265556 Chr.9: 36052563-36052563 on Build GRCh38
rs769867079 Chr.9: 36052307-36052307 on Build GRCh38
rs758330647 Chr.9: 36052250-36052251 on Build GRCh38
rs866520798 Chr.9: 36052324-36052324 on Build GRCh38
rs968833195 Chr.9: 36052172-36052172 on Build GRCh38
rs139435906 Chr.9: 36052452-36052452 on Build GRCh38
rs1019567811 Chr.9: 36052423-36052423 on Build GRCh38
rs909737446 Chr.9: 36052362-36052362 on Build GRCh38
rs948988747 Chr.9: 36052234-36052234 on Build GRCh38
rs186878673 Chr.9: 36052166-36052166 on Build GRCh38
rs562233477 Chr.9: 36052320-36052320 on Build GRCh38
rs570266152 Chr.9: 36052122-36052122 on Build GRCh38
rs747784529 Chr.9: 36052240-36052241 on Build GRCh38
rs919352857 Chr.9: 36052194-36052194 on Build GRCh38
rs557754331 Chr.9: 36052208-36052208 on Build GRCh38
rs535876977 Chr.9: 36052168-36052168 on Build GRCh38
rs527665158 Chr.9: 36052366-36052366 on Build GRCh38
rs946321795 Chr.9: 36052447-36052447 on Build GRCh38
rs1043594842 Chr.9: 36052525-36052525 on Build GRCh38
rs147362697 Chr.9: 36052272-36052272 on Build GRCh38
rs778457154 Chr.9: 36052233-36052235 on Build GRCh38
rs375145347 Chr.9: 36052136-36052136 on Build GRCh38
rs564621036 Chr.9: 36052441-36052441 on Build GRCh38
rs756739530 Chr.9: 36052150-36052155 on Build GRCh38
rs767141359 Chr.9: 36052313-36052313 on Build GRCh38
rs933345828 Chr.9: 36052407-36052407 on Build GRCh38
rs749305193 Chr.9: 36052333-36052333 on Build GRCh38
rs757013837 Chr.9: 36052235-36052235 on Build GRCh38
rs75502058 Chr.9: 36052145-36052145 on Build GRCh38
rs757099762 Chr.9: 36052361-36052361 on Build GRCh38
rs921911182 Chr.9: 36052396-36052396 on Build GRCh38
rs763497859 Chr.9: 36052309-36052309 on Build GRCh38
rs533618563 Chr.9: 36052445-36052445 on Build GRCh38
rs543212097 Chr.9: 36052243-36052243 on Build GRCh38
rs778887487 Chr.9: 36052241-36052241 on Build GRCh38
rs58611653 Chr.9: 36052308-36052308 on Build GRCh38
rs965178299 Chr.9: 36052427-36052427 on Build GRCh38
rs780016030 Chr.9: 36052375-36052375 on Build GRCh38
rs79379858 Chr.9: 36052163-36052163 on Build GRCh38
rs372895159 Chr.9: 36052372-36052372 on Build GRCh38
rs373725096 Chr.9: 36052305-36052305 on Build GRCh38

View Full Product Details