Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 84152351-84152949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHITM
Entrez Gene ID
27069
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
UniGene
Hs.352656
Gene Chromosome Location
Chr.10: 84139429-84153555 on Build GRCh38
Transcripts
NM_014394.2

SNP Detail

SNP ID Chromosome Location
rs780205061 Chr.10: 84152393-84152393 on Build GRCh38
rs927190823 Chr.10: 84152427-84152427 on Build GRCh38
rs11550183 Chr.10: 84152429-84152429 on Build GRCh38
rs778927694 Chr.10: 84152598-84152598 on Build GRCh38
rs538864298 Chr.10: 84152546-84152546 on Build GRCh38
rs373611275 Chr.10: 84152640-84152640 on Build GRCh38
rs768818352 Chr.10: 84152801-84152801 on Build GRCh38
rs878983134 Chr.10: 84152875-84152876 on Build GRCh38
rs773621818 Chr.10: 84152410-84152410 on Build GRCh38
rs7576 Chr.10: 84152850-84152850 on Build GRCh38
rs1005421484 Chr.10: 84152743-84152743 on Build GRCh38
rs964972526 Chr.10: 84152595-84152595 on Build GRCh38
rs1040716954 Chr.10: 84152544-84152544 on Build GRCh38
rs1017838053 Chr.10: 84152583-84152584 on Build GRCh38
rs753705673 Chr.10: 84152891-84152891 on Build GRCh38
rs992725480 Chr.10: 84152781-84152781 on Build GRCh38
rs1055558238 Chr.10: 84152776-84152776 on Build GRCh38
rs917941521 Chr.10: 84152788-84152788 on Build GRCh38
rs761187915 Chr.10: 84152386-84152387 on Build GRCh38
rs75526201 Chr.10: 84152771-84152771 on Build GRCh38
rs978218857 Chr.10: 84152707-84152707 on Build GRCh38
rs748425846 Chr.10: 84152897-84152897 on Build GRCh38
rs1007547967 Chr.10: 84152530-84152530 on Build GRCh38
rs572480398 Chr.10: 84152868-84152868 on Build GRCh38
rs185835541 Chr.10: 84152918-84152918 on Build GRCh38
rs1025312712 Chr.10: 84152649-84152649 on Build GRCh38
rs964896139 Chr.10: 84152923-84152924 on Build GRCh38
rs773854502 Chr.10: 84152910-84152910 on Build GRCh38
rs1141864 Chr.10: 84152515-84152515 on Build GRCh38
rs923447145 Chr.10: 84152641-84152641 on Build GRCh38
rs952653960 Chr.10: 84152664-84152664 on Build GRCh38
rs538085368 Chr.10: 84152725-84152725 on Build GRCh38
rs970688855 Chr.10: 84152839-84152839 on Build GRCh38
rs949471750 Chr.10: 84152459-84152459 on Build GRCh38
rs566797129 Chr.10: 84152484-84152484 on Build GRCh38
rs1018389275 Chr.10: 84152907-84152907 on Build GRCh38
rs1024849641 Chr.10: 84152489-84152489 on Build GRCh38
rs766376709 Chr.10: 84152851-84152851 on Build GRCh38
rs1006999654 Chr.10: 84152548-84152548 on Build GRCh38
rs879364939 Chr.10: 84152448-84152448 on Build GRCh38
rs749995304 Chr.10: 84152383-84152383 on Build GRCh38
rs578070366 Chr.10: 84152780-84152780 on Build GRCh38
rs181016088 Chr.10: 84152483-84152483 on Build GRCh38
rs753435761 Chr.10: 84152374-84152374 on Build GRCh38
rs984450808 Chr.10: 84152425-84152425 on Build GRCh38
rs757200203 Chr.10: 84152499-84152499 on Build GRCh38
rs1137057 Chr.10: 84152529-84152529 on Build GRCh38
rs749938140 Chr.10: 84152707-84152708 on Build GRCh38
rs924009953 Chr.10: 84152856-84152856 on Build GRCh38
rs952428144 Chr.10: 84152547-84152547 on Build GRCh38
rs1137056 Chr.10: 84152527-84152527 on Build GRCh38
rs374071585 Chr.10: 84152628-84152628 on Build GRCh38
rs191920969 Chr.10: 84152437-84152437 on Build GRCh38
rs756893798 Chr.10: 84152378-84152378 on Build GRCh38
rs888217306 Chr.10: 84152884-84152884 on Build GRCh38
rs1048132843 Chr.10: 84152867-84152867 on Build GRCh38
rs1141863 Chr.10: 84152512-84152512 on Build GRCh38
rs997723698 Chr.10: 84152925-84152925 on Build GRCh38
rs1027043283 Chr.10: 84152519-84152519 on Build GRCh38
rs764675575 Chr.10: 84152382-84152382 on Build GRCh38
rs568094249 Chr.10: 84152594-84152594 on Build GRCh38
rs1055063391 Chr.10: 84152418-84152418 on Build GRCh38
rs374249365 Chr.10: 84152862-84152862 on Build GRCh38
rs747018111 Chr.10: 84152636-84152636 on Build GRCh38
rs757946508 Chr.10: 84152384-84152384 on Build GRCh38
rs1064705 Chr.10: 84152509-84152509 on Build GRCh38
rs1042472933 Chr.10: 84152477-84152477 on Build GRCh38
rs139674609 Chr.10: 84152919-84152919 on Build GRCh38

View Full Product Details