Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 98208111-98208699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
UniGene
Hs.419800
Gene Chromosome Location
Chr.10: 98134608-98244897 on Build GRCh38
Transcripts
NM_001256619.1 NM_001256620.1 NM_001256621.1 NM_014472.4 NM_138469.2 XM_011539639.1 XM_011539640.1 XM_011539641.1 XM_011539642.1 XM_011539643.1 XM_011539644.1 XM_011539645.2 XM_011539646.1 XM_011539647.1 XM_011539648.1 XM_011539649.1 XM_017016074.1 XM_017016075.1 XM_017016076.1 XM_017016077.1 XM_017016078.1 XM_017016079.1 XM_017016080.1 XM_017016081.1 XM_017016082.1 XM_017016083.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3442463 c.205C>T p.R69* Chr.10: 98208319-98208319 on Build GRCh38
4666505 c.173delA p.E60fs*12 Chr.10: 98208287-98208287 on Build GRCh38
1188017 c.334G>T p.E112* Chr.10: 98208448-98208448 on Build GRCh38
3442461 c.167T>A p.L56H Chr.10: 98208281-98208281 on Build GRCh38
4484833 c.285C>A p.D95E Chr.10: 98208399-98208399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144063837 Chr.10: 98208595-98208595 on Build GRCh38
rs748035476 Chr.10: 98208499-98208499 on Build GRCh38
rs148941402 Chr.10: 98208178-98208178 on Build GRCh38
rs763902171 Chr.10: 98208197-98208197 on Build GRCh38
rs765832966 Chr.10: 98208300-98208300 on Build GRCh38
rs368848459 Chr.10: 98208505-98208505 on Build GRCh38
rs777615233 Chr.10: 98208583-98208583 on Build GRCh38
rs756463875 Chr.10: 98208231-98208231 on Build GRCh38
rs375521100 Chr.10: 98208221-98208221 on Build GRCh38
rs1053155493 Chr.10: 98208137-98208137 on Build GRCh38
rs775591805 Chr.10: 98208278-98208278 on Build GRCh38
rs757543615 Chr.10: 98208243-98208243 on Build GRCh38
rs769525398 Chr.10: 98208362-98208362 on Build GRCh38
rs778174796 Chr.10: 98208587-98208603 on Build GRCh38
rs372453818 Chr.10: 98208572-98208572 on Build GRCh38
rs764870906 Chr.10: 98208284-98208284 on Build GRCh38
rs764036501 Chr.10: 98208649-98208649 on Build GRCh38
rs746975266 Chr.10: 98208328-98208328 on Build GRCh38
rs776472833 Chr.10: 98208281-98208281 on Build GRCh38
rs755899792 Chr.10: 98208276-98208276 on Build GRCh38
rs751664482 Chr.10: 98208454-98208454 on Build GRCh38
rs770428557 Chr.10: 98208607-98208607 on Build GRCh38
rs773813387 Chr.10: 98208523-98208523 on Build GRCh38
rs749524971 Chr.10: 98208241-98208241 on Build GRCh38
rs996651942 Chr.10: 98208466-98208466 on Build GRCh38
rs766101897 Chr.10: 98208343-98208346 on Build GRCh38
rs756948139 Chr.10: 98208587-98208587 on Build GRCh38
rs770480067 Chr.10: 98208380-98208380 on Build GRCh38
rs748729724 Chr.10: 98208601-98208601 on Build GRCh38
rs778651453 Chr.10: 98208588-98208588 on Build GRCh38
rs780064819 Chr.10: 98208440-98208440 on Build GRCh38
rs537212901 Chr.10: 98208135-98208135 on Build GRCh38
rs867119474 Chr.10: 98208265-98208265 on Build GRCh38
rs775931614 Chr.10: 98208443-98208443 on Build GRCh38
rs112811238 Chr.10: 98208131-98208131 on Build GRCh38
rs770958922 Chr.10: 98208168-98208168 on Build GRCh38
rs926745859 Chr.10: 98208620-98208620 on Build GRCh38
rs763394042 Chr.10: 98208611-98208611 on Build GRCh38
rs761155634 Chr.10: 98208390-98208390 on Build GRCh38
rs1056657847 Chr.10: 98208631-98208631 on Build GRCh38
rs757108666 Chr.10: 98208661-98208661 on Build GRCh38
rs747483284 Chr.10: 98208614-98208615 on Build GRCh38
rs758583644 Chr.10: 98208324-98208324 on Build GRCh38
rs754242171 Chr.10: 98208398-98208398 on Build GRCh38
rs371809764 Chr.10: 98208638-98208638 on Build GRCh38
rs777877415 Chr.10: 98208154-98208154 on Build GRCh38
rs780159397 Chr.10: 98208327-98208327 on Build GRCh38
rs370973175 Chr.10: 98208522-98208522 on Build GRCh38
rs759651880 Chr.10: 98208626-98208626 on Build GRCh38
rs372164414 Chr.10: 98208385-98208385 on Build GRCh38
rs1003266861 Chr.10: 98208274-98208274 on Build GRCh38
rs774555023 Chr.10: 98208174-98208174 on Build GRCh38
rs772878995 Chr.10: 98208368-98208368 on Build GRCh38
rs777227073 Chr.10: 98208546-98208546 on Build GRCh38
rs753188303 Chr.10: 98208303-98208303 on Build GRCh38
rs752772544 Chr.10: 98208647-98208647 on Build GRCh38
rs781208585 Chr.10: 98208487-98208487 on Build GRCh38
rs762559219 Chr.10: 98208376-98208376 on Build GRCh38
rs747984030 Chr.10: 98208353-98208353 on Build GRCh38
rs772789148 Chr.10: 98208293-98208293 on Build GRCh38
rs761224574 Chr.10: 98208311-98208311 on Build GRCh38
rs763342003 Chr.10: 98208557-98208557 on Build GRCh38
rs12775148 Chr.10: 98208451-98208451 on Build GRCh38
rs749891945 Chr.10: 98208249-98208250 on Build GRCh38
rs771654574 Chr.10: 98208593-98208593 on Build GRCh38
rs781204722 Chr.10: 98208220-98208220 on Build GRCh38
rs766609732 Chr.10: 98208621-98208621 on Build GRCh38
rs753726893 Chr.10: 98208651-98208651 on Build GRCh38
rs867369065 Chr.10: 98208217-98208217 on Build GRCh38
rs762380439 Chr.10: 98208549-98208549 on Build GRCh38
rs777525173 Chr.10: 98208515-98208515 on Build GRCh38
rs555004057 Chr.10: 98208296-98208296 on Build GRCh38
rs146588639 Chr.10: 98208577-98208577 on Build GRCh38
rs771435285 Chr.10: 98208539-98208539 on Build GRCh38
rs373657280 Chr.10: 98208320-98208320 on Build GRCh38
rs749089389 Chr.10: 98208585-98208585 on Build GRCh38
rs766990785 Chr.10: 98208220-98208222 on Build GRCh38
rs749930292 Chr.10: 98208215-98208215 on Build GRCh38
rs758679632 Chr.10: 98208433-98208433 on Build GRCh38
rs762292516 Chr.10: 98208403-98208403 on Build GRCh38
rs752678332 Chr.10: 98208580-98208580 on Build GRCh38
rs147319818 Chr.10: 98208648-98208648 on Build GRCh38
rs749489715 Chr.10: 98208163-98208163 on Build GRCh38
rs754229437 Chr.10: 98208313-98208313 on Build GRCh38
rs757629684 Chr.10: 98208318-98208318 on Build GRCh38
rs753163228 Chr.10: 98208420-98208421 on Build GRCh38
rs370226522 Chr.10: 98208337-98208337 on Build GRCh38
rs374768883 Chr.10: 98208425-98208425 on Build GRCh38
rs779291841 Chr.10: 98208319-98208319 on Build GRCh38
rs776186337 Chr.10: 98208198-98208198 on Build GRCh38
rs764428816 Chr.10: 98208312-98208312 on Build GRCh38
rs779202001 Chr.10: 98208253-98208253 on Build GRCh38
rs7918729 Chr.10: 98208409-98208409 on Build GRCh38
rs764784738 Chr.10: 98208214-98208214 on Build GRCh38
rs761439395 Chr.10: 98208282-98208282 on Build GRCh38
rs745586104 Chr.10: 98208676-98208676 on Build GRCh38
rs768383441 Chr.10: 98208280-98208280 on Build GRCh38
rs759098965 Chr.10: 98208534-98208534 on Build GRCh38
rs760402020 Chr.10: 98208188-98208188 on Build GRCh38
rs143631828 Chr.10: 98208338-98208338 on Build GRCh38
rs1019720514 Chr.10: 98208377-98208377 on Build GRCh38
rs183340020 Chr.10: 98208592-98208592 on Build GRCh38
rs774670563 Chr.10: 98208624-98208624 on Build GRCh38
rs756440903 Chr.10: 98208150-98208150 on Build GRCh38
rs773706530 Chr.10: 98208552-98208552 on Build GRCh38
rs757833466 Chr.10: 98208218-98208218 on Build GRCh38
rs772223175 Chr.10: 98208270-98208270 on Build GRCh38
rs201437571 Chr.10: 98208172-98208172 on Build GRCh38
rs745946797 Chr.10: 98208260-98208260 on Build GRCh38
rs750722213 Chr.10: 98208428-98208428 on Build GRCh38
rs755112924 Chr.10: 98208575-98208575 on Build GRCh38
rs758788220 Chr.10: 98208519-98208520 on Build GRCh38
rs542901557 Chr.10: 98208608-98208608 on Build GRCh38
rs754950643 Chr.10: 98208482-98208482 on Build GRCh38
rs868106160 Chr.10: 98208452-98208452 on Build GRCh38
rs761502568 Chr.10: 98208205-98208205 on Build GRCh38
rs767695966 Chr.10: 98208627-98208627 on Build GRCh38
rs756035136 Chr.10: 98208582-98208582 on Build GRCh38
rs555948031 Chr.10: 98208573-98208573 on Build GRCh38
rs778171436 Chr.10: 98208233-98208233 on Build GRCh38
rs775501969 Chr.10: 98208187-98208187 on Build GRCh38
rs7903980 Chr.10: 98208408-98208408 on Build GRCh38
rs369258826 Chr.10: 98208671-98208671 on Build GRCh38
rs144192127 Chr.10: 98208550-98208550 on Build GRCh38
rs139280105 Chr.10: 98208674-98208674 on Build GRCh38
rs753197111 Chr.10: 98208222-98208222 on Build GRCh38
rs764667669 Chr.10: 98208393-98208393 on Build GRCh38

View Full Product Details