Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 758581-759177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TALDO1
Entrez Gene ID
6888
Gene Name
transaldolase 1
Gene Aliases
TAL, TAL-H, TALDOR, TALH
UniGene
Hs.438678
Gene Chromosome Location
Chr.11: 747432-765024 on Build GRCh38
Transcripts
NM_006755.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4036777 c.280G>T p.E94* Chr.11: 759008-759008 on Build GRCh38
3416242 c.299C>T p.P100L Chr.11: 759027-759027 on Build GRCh38
6796526 c.234C>T p.D78D Chr.11: 758962-758962 on Build GRCh38
3453402 c.239T>A p.I80N Chr.11: 758967-758967 on Build GRCh38
4485904 c.298C>T p.P100S Chr.11: 759026-759026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373523138 Chr.11: 758608-758610 on Build GRCh38
rs560903146 Chr.11: 758842-758842 on Build GRCh38
rs758959534 Chr.11: 759099-759099 on Build GRCh38
rs942358093 Chr.11: 758996-758996 on Build GRCh38
rs559528907 Chr.11: 758883-758883 on Build GRCh38
rs758756190 Chr.11: 759021-759021 on Build GRCh38
rs969377124 Chr.11: 758857-758857 on Build GRCh38
rs774215394 Chr.11: 759054-759054 on Build GRCh38
rs928479369 Chr.11: 758766-758766 on Build GRCh38
rs766745943 Chr.11: 759056-759056 on Build GRCh38
rs762947540 Chr.11: 759046-759046 on Build GRCh38
rs925451883 Chr.11: 758609-758611 on Build GRCh38
rs1000895296 Chr.11: 758879-758879 on Build GRCh38
rs751243397 Chr.11: 758978-758978 on Build GRCh38
rs565905349 Chr.11: 758659-758659 on Build GRCh38
rs748499989 Chr.11: 758903-758903 on Build GRCh38
rs761656906 Chr.11: 759055-759055 on Build GRCh38
rs979558672 Chr.11: 758609-758609 on Build GRCh38
rs756188925 Chr.11: 759083-759083 on Build GRCh38
rs767660413 Chr.11: 758934-758934 on Build GRCh38
rs1010038296 Chr.11: 758617-758617 on Build GRCh38
rs948510883 Chr.11: 758753-758753 on Build GRCh38
rs369934964 Chr.11: 759028-759028 on Build GRCh38
rs1050684252 Chr.11: 758793-758793 on Build GRCh38
rs796228029 Chr.11: 758934-758934 on Build GRCh38
rs1022386706 Chr.11: 758641-758641 on Build GRCh38
rs762710253 Chr.11: 758964-758964 on Build GRCh38
rs1006601504 Chr.11: 758658-758658 on Build GRCh38
rs398075669 Chr.11: 758933-758933 on Build GRCh38
rs923574002 Chr.11: 758636-758636 on Build GRCh38
rs541236541 Chr.11: 758960-758960 on Build GRCh38
rs11246304 Chr.11: 758674-758674 on Build GRCh38
rs919685768 Chr.11: 758750-758750 on Build GRCh38
rs929745406 Chr.11: 759136-759136 on Build GRCh38
rs189988418 Chr.11: 758897-758897 on Build GRCh38
rs988688570 Chr.11: 758975-758975 on Build GRCh38
rs766124800 Chr.11: 758706-758706 on Build GRCh38
rs185224249 Chr.11: 758847-758847 on Build GRCh38
rs1015853722 Chr.11: 758884-758884 on Build GRCh38
rs375636303 Chr.11: 758692-758692 on Build GRCh38
rs368967856 Chr.11: 759052-759052 on Build GRCh38
rs1023593067 Chr.11: 758853-758853 on Build GRCh38
rs56340568 Chr.11: 758806-758806 on Build GRCh38
rs948138883 Chr.11: 758608-758609 on Build GRCh38
rs979920364 Chr.11: 758761-758761 on Build GRCh38
rs759321138 Chr.11: 758808-758808 on Build GRCh38
rs545136709 Chr.11: 758807-758807 on Build GRCh38
rs891311340 Chr.11: 758860-758860 on Build GRCh38
rs563875373 Chr.11: 758608-758608 on Build GRCh38
rs557177693 Chr.11: 758712-758712 on Build GRCh38
rs903928202 Chr.11: 758838-758838 on Build GRCh38
rs542864521 Chr.11: 758865-758865 on Build GRCh38
rs1015649861 Chr.11: 758708-758708 on Build GRCh38
rs1028482102 Chr.11: 758686-758686 on Build GRCh38
rs528700868 Chr.11: 758894-758894 on Build GRCh38
rs567483503 Chr.11: 758733-758733 on Build GRCh38
rs1013518037 Chr.11: 758852-758852 on Build GRCh38
rs200086212 Chr.11: 758979-758979 on Build GRCh38
rs1053038297 Chr.11: 758778-758778 on Build GRCh38
rs754089425 Chr.11: 759075-759075 on Build GRCh38
rs202135397 Chr.11: 759032-759032 on Build GRCh38
rs961293099 Chr.11: 758887-758887 on Build GRCh38
rs779609987 Chr.11: 759000-759000 on Build GRCh38
rs903981544 Chr.11: 758660-758660 on Build GRCh38
rs536513945 Chr.11: 758751-758751 on Build GRCh38
rs34225557 Chr.11: 758928-758928 on Build GRCh38
rs896356193 Chr.11: 758851-758851 on Build GRCh38
rs755075874 Chr.11: 758995-758995 on Build GRCh38
rs1049811172 Chr.11: 759124-759124 on Build GRCh38
rs778205812 Chr.11: 759027-759027 on Build GRCh38
rs891258711 Chr.11: 759146-759146 on Build GRCh38
rs766657765 Chr.11: 758980-758980 on Build GRCh38
rs763940714 Chr.11: 758976-758976 on Build GRCh38
rs892486476 Chr.11: 758610-758610 on Build GRCh38
rs113906981 Chr.11: 758948-758948 on Build GRCh38
rs1008403125 Chr.11: 758881-758881 on Build GRCh38
rs758999901 Chr.11: 758947-758947 on Build GRCh38
rs957077240 Chr.11: 758752-758752 on Build GRCh38
rs772797515 Chr.11: 758904-758904 on Build GRCh38
rs765195164 Chr.11: 758809-758809 on Build GRCh38
rs553228250 Chr.11: 758772-758772 on Build GRCh38
rs750129546 Chr.11: 759096-759104 on Build GRCh38
rs567624141 Chr.11: 759112-759112 on Build GRCh38
rs1028429633 Chr.11: 758848-758848 on Build GRCh38
rs996967673 Chr.11: 758667-758667 on Build GRCh38
rs557301276 Chr.11: 758726-758726 on Build GRCh38
rs775352129 Chr.11: 759042-759042 on Build GRCh38
rs753999490 Chr.11: 758990-758990 on Build GRCh38
rs752604909 Chr.11: 758867-758867 on Build GRCh38
rs558728228 Chr.11: 758819-758819 on Build GRCh38
rs879205722 Chr.11: 759059-759059 on Build GRCh38
rs751345391 Chr.11: 758795-758795 on Build GRCh38
rs770594711 Chr.11: 759031-759031 on Build GRCh38
rs761744895 Chr.11: 758927-758928 on Build GRCh38
rs551420192 Chr.11: 758695-758695 on Build GRCh38
rs969427959 Chr.11: 758723-758723 on Build GRCh38
rs755165740 Chr.11: 759081-759081 on Build GRCh38
rs575296779 Chr.11: 758826-758826 on Build GRCh38
rs374927511 Chr.11: 758849-758849 on Build GRCh38
rs375716755 Chr.11: 758910-758910 on Build GRCh38
rs778095063 Chr.11: 759109-759109 on Build GRCh38
rs148461396 Chr.11: 758722-758722 on Build GRCh38
rs748674950 Chr.11: 759007-759007 on Build GRCh38
rs752653071 Chr.11: 759096-759096 on Build GRCh38
rs571396228 Chr.11: 758696-758696 on Build GRCh38
rs761605291 Chr.11: 759004-759006 on Build GRCh38
rs765355304 Chr.11: 759091-759091 on Build GRCh38
rs745426306 Chr.11: 759033-759033 on Build GRCh38
rs543897139 Chr.11: 758829-758829 on Build GRCh38
rs910828121 Chr.11: 758973-758973 on Build GRCh38
rs529571995 Chr.11: 758643-758643 on Build GRCh38
rs755633643 Chr.11: 759107-759108 on Build GRCh38
rs1046912658 Chr.11: 758841-758841 on Build GRCh38
rs371351132 Chr.11: 758714-758714 on Build GRCh38
rs913146515 Chr.11: 759018-759018 on Build GRCh38
rs1027097090 Chr.11: 758895-758895 on Build GRCh38
rs769273951 Chr.11: 759037-759037 on Build GRCh38
rs111967732 Chr.11: 758662-758662 on Build GRCh38
rs549179090 Chr.11: 758654-758654 on Build GRCh38

View Full Product Details