Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68744707-68745294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TESMIN
Entrez Gene ID
9633
Gene Name
testis expressed metallothionein like protein
Gene Aliases
CXCDC2, MTL5, MTLT
UniGene
Hs.145932
Gene Chromosome Location
Chr.11: 68706674-68751564 on Build GRCh38
Transcripts
NM_001039656.1 NM_004923.3 XM_011545402.1 XM_011545403.2 XM_017018588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5117759 c.740A>C p.Y247S Chr.11: 68745002-68745002 on Build GRCh38
78114 c.653G>C p.G218A Chr.11: 68745089-68745089 on Build GRCh38
4641548 c.641A>C p.Q214P Chr.11: 68745101-68745101 on Build GRCh38
6871976 c.652G>T p.G218C Chr.11: 68745090-68745090 on Build GRCh38
931301 c.666A>G p.L222L Chr.11: 68745076-68745076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995418455 Chr.11: 68744895-68744895 on Build GRCh38
rs200666522 Chr.11: 68745022-68745022 on Build GRCh38
rs766334730 Chr.11: 68745023-68745023 on Build GRCh38
rs1031600193 Chr.11: 68745227-68745227 on Build GRCh38
rs770307593 Chr.11: 68744963-68744963 on Build GRCh38
rs899153019 Chr.11: 68745199-68745199 on Build GRCh38
rs752773586 Chr.11: 68745027-68745027 on Build GRCh38
rs148917619 Chr.11: 68745252-68745252 on Build GRCh38
rs958170308 Chr.11: 68745230-68745230 on Build GRCh38
rs191862043 Chr.11: 68744794-68744794 on Build GRCh38
rs924240598 Chr.11: 68744859-68744859 on Build GRCh38
rs1046485714 Chr.11: 68744797-68744797 on Build GRCh38
rs565667752 Chr.11: 68745208-68745208 on Build GRCh38
rs774386439 Chr.11: 68745158-68745158 on Build GRCh38
rs767464303 Chr.11: 68744980-68744983 on Build GRCh38
rs949584329 Chr.11: 68745109-68745109 on Build GRCh38
rs751737906 Chr.11: 68745056-68745056 on Build GRCh38
rs763027489 Chr.11: 68745039-68745039 on Build GRCh38
rs974799914 Chr.11: 68744742-68744742 on Build GRCh38
rs954938580 Chr.11: 68745176-68745176 on Build GRCh38
rs386754556 Chr.11: 68744793-68744794 on Build GRCh38
rs978313959 Chr.11: 68745219-68745219 on Build GRCh38
rs773936122 Chr.11: 68744966-68744966 on Build GRCh38
rs748818142 Chr.11: 68744973-68744973 on Build GRCh38
rs1046992138 Chr.11: 68745218-68745219 on Build GRCh38
rs759387020 Chr.11: 68744965-68744966 on Build GRCh38
rs948330746 Chr.11: 68744913-68744913 on Build GRCh38
rs773989622 Chr.11: 68745015-68745015 on Build GRCh38
rs559629490 Chr.11: 68744896-68744896 on Build GRCh38
rs938097384 Chr.11: 68745136-68745136 on Build GRCh38
rs12365708 Chr.11: 68745075-68745075 on Build GRCh38
rs539170978 Chr.11: 68745225-68745225 on Build GRCh38
rs201348974 Chr.11: 68744945-68744945 on Build GRCh38
rs756931325 Chr.11: 68744740-68744740 on Build GRCh38
rs747850077 Chr.11: 68745097-68745097 on Build GRCh38
rs779500448 Chr.11: 68744967-68744967 on Build GRCh38
rs906939899 Chr.11: 68744920-68744920 on Build GRCh38
rs752642431 Chr.11: 68745126-68745129 on Build GRCh38
rs757450851 Chr.11: 68745065-68745065 on Build GRCh38
rs145715040 Chr.11: 68744901-68744901 on Build GRCh38
rs954932513 Chr.11: 68744806-68744806 on Build GRCh38
rs1025928772 Chr.11: 68744900-68744900 on Build GRCh38
rs756641364 Chr.11: 68745074-68745074 on Build GRCh38
rs146894157 Chr.11: 68745100-68745100 on Build GRCh38
rs951260739 Chr.11: 68744930-68744930 on Build GRCh38
rs375655320 Chr.11: 68744995-68744995 on Build GRCh38
rs552428433 Chr.11: 68744919-68744919 on Build GRCh38
rs1053069505 Chr.11: 68744775-68744777 on Build GRCh38
rs764285578 Chr.11: 68745148-68745148 on Build GRCh38
rs145721990 Chr.11: 68745111-68745111 on Build GRCh38
rs761732540 Chr.11: 68745016-68745016 on Build GRCh38
rs773207349 Chr.11: 68745106-68745106 on Build GRCh38
rs770783365 Chr.11: 68745122-68745122 on Build GRCh38
rs541509436 Chr.11: 68744864-68744864 on Build GRCh38
rs762971284 Chr.11: 68745135-68745135 on Build GRCh38
rs748753442 Chr.11: 68745088-68745088 on Build GRCh38
rs1019376143 Chr.11: 68744946-68744946 on Build GRCh38
rs1002171061 Chr.11: 68744826-68744826 on Build GRCh38
rs776561783 Chr.11: 68745128-68745128 on Build GRCh38
rs572556752 Chr.11: 68744791-68744792 on Build GRCh38
rs764224339 Chr.11: 68745053-68745053 on Build GRCh38
rs773117507 Chr.11: 68745021-68745021 on Build GRCh38
rs750786155 Chr.11: 68745070-68745070 on Build GRCh38
rs184029172 Chr.11: 68745164-68745164 on Build GRCh38
rs868772662 Chr.11: 68744868-68744868 on Build GRCh38
rs750393592 Chr.11: 68745232-68745232 on Build GRCh38
rs149843149 Chr.11: 68744813-68744813 on Build GRCh38
rs781682081 Chr.11: 68744952-68744952 on Build GRCh38
rs746425083 Chr.11: 68744953-68744953 on Build GRCh38
rs7948522 Chr.11: 68744861-68744861 on Build GRCh38
rs771943100 Chr.11: 68745019-68745019 on Build GRCh38
rs373770823 Chr.11: 68744955-68744955 on Build GRCh38
rs938913318 Chr.11: 68744985-68744985 on Build GRCh38
rs780885260 Chr.11: 68744843-68744843 on Build GRCh38
rs768172750 Chr.11: 68745092-68745092 on Build GRCh38
rs553817172 Chr.11: 68745175-68745175 on Build GRCh38
rs781580058 Chr.11: 68745069-68745069 on Build GRCh38
rs761671970 Chr.11: 68744977-68744977 on Build GRCh38

View Full Product Details