Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123725275-123725851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF202
Entrez Gene ID
7753
Gene Name
zinc finger protein 202
Gene Aliases
ZKSCAN10, ZSCAN42
UniGene
Hs.112556
Gene Chromosome Location
Chr.11: 123723927-123741705 on Build GRCh38
Transcripts
NM_001301779.1 NM_001301780.1 NM_001301819.1 NM_003455.3 XM_005271659.1 XM_005271660.1 XM_005271661.1 XM_005271664.1 XM_006718901.2 XM_011542972.1 XM_011542973.1 XM_011542975.1 XM_011542976.1 XM_017018268.1

SNP Detail

SNP ID Chromosome Location
rs1042419422 Chr.11: 123725303-123725303 on Build GRCh38
rs962379356 Chr.11: 123725375-123725375 on Build GRCh38
rs1033793168 Chr.11: 123725343-123725343 on Build GRCh38
rs984033005 Chr.11: 123725607-123725607 on Build GRCh38
rs1046941543 Chr.11: 123725800-123725800 on Build GRCh38
rs918505667 Chr.11: 123725337-123725337 on Build GRCh38
rs1042387337 Chr.11: 123725433-123725433 on Build GRCh38
rs548948273 Chr.11: 123725528-123725528 on Build GRCh38
rs534801963 Chr.11: 123725592-123725592 on Build GRCh38
rs1024155618 Chr.11: 123725519-123725521 on Build GRCh38
rs903206509 Chr.11: 123725729-123725729 on Build GRCh38
rs755120074 Chr.11: 123725403-123725403 on Build GRCh38
rs10904 Chr.11: 123725347-123725347 on Build GRCh38
rs945622292 Chr.11: 123725423-123725423 on Build GRCh38
rs912769631 Chr.11: 123725420-123725420 on Build GRCh38
rs540363219 Chr.11: 123725379-123725379 on Build GRCh38
rs781512221 Chr.11: 123725454-123725454 on Build GRCh38
rs1015231834 Chr.11: 123725373-123725373 on Build GRCh38
rs547025721 Chr.11: 123725681-123725681 on Build GRCh38
rs182582061 Chr.11: 123725814-123725814 on Build GRCh38
rs1025235044 Chr.11: 123725581-123725581 on Build GRCh38
rs909794603 Chr.11: 123725613-123725613 on Build GRCh38
rs1055610532 Chr.11: 123725485-123725485 on Build GRCh38
rs907106365 Chr.11: 123725523-123725523 on Build GRCh38
rs770159553 Chr.11: 123725671-123725671 on Build GRCh38
rs894662887 Chr.11: 123725488-123725488 on Build GRCh38
rs895235556 Chr.11: 123725513-123725513 on Build GRCh38
rs949514849 Chr.11: 123725745-123725745 on Build GRCh38
rs557214206 Chr.11: 123725719-123725719 on Build GRCh38
rs982384536 Chr.11: 123725365-123725365 on Build GRCh38
rs1041761349 Chr.11: 123725728-123725728 on Build GRCh38
rs898419251 Chr.11: 123725819-123725819 on Build GRCh38
rs764064245 Chr.11: 123725316-123725316 on Build GRCh38
rs987342792 Chr.11: 123725660-123725660 on Build GRCh38
rs7938875 Chr.11: 123725381-123725381 on Build GRCh38
rs907034381 Chr.11: 123725807-123725807 on Build GRCh38
rs1003270855 Chr.11: 123725824-123725824 on Build GRCh38
rs1023844063 Chr.11: 123725514-123725514 on Build GRCh38
rs953826592 Chr.11: 123725657-123725657 on Build GRCh38
rs975235356 Chr.11: 123725686-123725686 on Build GRCh38
rs546471995 Chr.11: 123725355-123725355 on Build GRCh38
rs981750439 Chr.11: 123725571-123725571 on Build GRCh38
rs569982554 Chr.11: 123725465-123725465 on Build GRCh38
rs111411025 Chr.11: 123725363-123725363 on Build GRCh38
rs191535439 Chr.11: 123725304-123725304 on Build GRCh38
rs531986088 Chr.11: 123725724-123725724 on Build GRCh38
rs962346914 Chr.11: 123725596-123725596 on Build GRCh38
rs981739135 Chr.11: 123725335-123725335 on Build GRCh38
rs970464019 Chr.11: 123725521-123725521 on Build GRCh38
rs1055148094 Chr.11: 123725764-123725764 on Build GRCh38
rs759684279 Chr.11: 123725492-123725492 on Build GRCh38
rs747158655 Chr.11: 123725374-123725374 on Build GRCh38
rs753828929 Chr.11: 123725327-123725327 on Build GRCh38
rs903452800 Chr.11: 123725406-123725406 on Build GRCh38
rs34067094 Chr.11: 123725589-123725589 on Build GRCh38
rs906305507 Chr.11: 123725816-123725816 on Build GRCh38
rs11826384 Chr.11: 123725348-123725348 on Build GRCh38
rs571658461 Chr.11: 123725684-123725684 on Build GRCh38
rs749793791 Chr.11: 123725741-123725741 on Build GRCh38
rs148310963 Chr.11: 123725696-123725696 on Build GRCh38
rs929861160 Chr.11: 123725340-123725340 on Build GRCh38
rs531135805 Chr.11: 123725479-123725479 on Build GRCh38
rs973806495 Chr.11: 123725602-123725602 on Build GRCh38
rs1024604360 Chr.11: 123725350-123725351 on Build GRCh38
rs187897245 Chr.11: 123725421-123725421 on Build GRCh38
rs894723120 Chr.11: 123725810-123725810 on Build GRCh38
rs1004069709 Chr.11: 123725588-123725588 on Build GRCh38
rs12223520 Chr.11: 123725813-123725813 on Build GRCh38
rs932808382 Chr.11: 123725396-123725397 on Build GRCh38
rs143232560 Chr.11: 123725434-123725434 on Build GRCh38
rs936652156 Chr.11: 123725722-123725722 on Build GRCh38
rs934037021 Chr.11: 123725723-123725723 on Build GRCh38
rs183841293 Chr.11: 123725389-123725389 on Build GRCh38
rs903837264 Chr.11: 123725443-123725443 on Build GRCh38

View Full Product Details