Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118315292-118315870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
UniGene
Hs.3003
Gene Chromosome Location
Chr.11: 118304580-118316175 on Build GRCh38
Transcripts
NM_000733.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282365 c.568-6C>A p.? Chr.11: 118315480-118315480 on Build GRCh38
77442 c.610C>G p.Q204E Chr.11: 118315528-118315528 on Build GRCh38
1351581 c.588G>A p.R196R Chr.11: 118315506-118315506 on Build GRCh38
4797843 c.569C>T p.P190L Chr.11: 118315487-118315487 on Build GRCh38
220482 c.589G>A p.D197N Chr.11: 118315507-118315507 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs555454200 Chr.11: 118315687-118315687 on Build GRCh38
rs199737067 Chr.11: 118315449-118315449 on Build GRCh38
rs905152901 Chr.11: 118315579-118315579 on Build GRCh38
rs765977907 Chr.11: 118315479-118315479 on Build GRCh38
rs546940917 Chr.11: 118315623-118315623 on Build GRCh38
rs773134010 Chr.11: 118315555-118315555 on Build GRCh38
rs769877796 Chr.11: 118315475-118315475 on Build GRCh38
rs780379117 Chr.11: 118315532-118315532 on Build GRCh38
rs764127460 Chr.11: 118315474-118315475 on Build GRCh38
rs767177871 Chr.11: 118315591-118315591 on Build GRCh38
rs1037136378 Chr.11: 118315830-118315830 on Build GRCh38
rs568938400 Chr.11: 118315401-118315401 on Build GRCh38
rs200452738 Chr.11: 118315657-118315657 on Build GRCh38
rs183482409 Chr.11: 118315821-118315821 on Build GRCh38
rs2231450 Chr.11: 118315597-118315597 on Build GRCh38
rs761666384 Chr.11: 118315595-118315595 on Build GRCh38
rs953753419 Chr.11: 118315416-118315416 on Build GRCh38
rs749302779 Chr.11: 118315557-118315557 on Build GRCh38
rs952822130 Chr.11: 118315341-118315341 on Build GRCh38
rs116370847 Chr.11: 118315738-118315738 on Build GRCh38
rs899664323 Chr.11: 118315677-118315677 on Build GRCh38
rs776569602 Chr.11: 118315562-118315577 on Build GRCh38
rs200463838 Chr.11: 118315453-118315453 on Build GRCh38
rs757693478 Chr.11: 118315516-118315516 on Build GRCh38
rs2231449 Chr.11: 118315590-118315590 on Build GRCh38
rs1044670959 Chr.11: 118315711-118315711 on Build GRCh38
rs201490654 Chr.11: 118315450-118315450 on Build GRCh38
rs940055607 Chr.11: 118315816-118315816 on Build GRCh38
rs201301103 Chr.11: 118315458-118315458 on Build GRCh38
rs201116390 Chr.11: 118315632-118315632 on Build GRCh38
rs202217457 Chr.11: 118315633-118315633 on Build GRCh38
rs770715556 Chr.11: 118315576-118315576 on Build GRCh38
rs754411015 Chr.11: 118315511-118315511 on Build GRCh38
rs998818732 Chr.11: 118315645-118315645 on Build GRCh38
rs758682387 Chr.11: 118315531-118315531 on Build GRCh38
rs745871914 Chr.11: 118315526-118315526 on Build GRCh38
rs981500571 Chr.11: 118315566-118315566 on Build GRCh38
rs112837142 Chr.11: 118315348-118315348 on Build GRCh38
rs764411267 Chr.11: 118315505-118315505 on Build GRCh38
rs551375526 Chr.11: 118315499-118315499 on Build GRCh38
rs560285795 Chr.11: 118315352-118315352 on Build GRCh38
rs201731562 Chr.11: 118315689-118315689 on Build GRCh38
rs764554208 Chr.11: 118315614-118315614 on Build GRCh38
rs949933709 Chr.11: 118315706-118315706 on Build GRCh38
rs200332418 Chr.11: 118315686-118315686 on Build GRCh38
rs1054017768 Chr.11: 118315744-118315744 on Build GRCh38
rs917042998 Chr.11: 118315692-118315692 on Build GRCh38
rs888475605 Chr.11: 118315792-118315792 on Build GRCh38
rs200926380 Chr.11: 118315668-118315668 on Build GRCh38
rs1047168765 Chr.11: 118315448-118315448 on Build GRCh38
rs934447043 Chr.11: 118315492-118315492 on Build GRCh38
rs201952371 Chr.11: 118315690-118315690 on Build GRCh38
rs199760495 Chr.11: 118315495-118315495 on Build GRCh38
rs1004322289 Chr.11: 118315624-118315624 on Build GRCh38
rs567060197 Chr.11: 118315658-118315658 on Build GRCh38
rs778211203 Chr.11: 118315452-118315452 on Build GRCh38
rs908201331 Chr.11: 118315651-118315651 on Build GRCh38
rs200726133 Chr.11: 118315742-118315742 on Build GRCh38
rs146149691 Chr.11: 118315400-118315400 on Build GRCh38
rs534344418 Chr.11: 118315829-118315829 on Build GRCh38
rs200083794 Chr.11: 118315535-118315535 on Build GRCh38
rs977058961 Chr.11: 118315392-118315392 on Build GRCh38
rs537874937 Chr.11: 118315770-118315770 on Build GRCh38
rs200968283 Chr.11: 118315556-118315556 on Build GRCh38
rs141553540 Chr.11: 118315498-118315498 on Build GRCh38
rs773328619 Chr.11: 118315476-118315476 on Build GRCh38
rs1028578929 Chr.11: 118315653-118315653 on Build GRCh38
rs778855337 Chr.11: 118315462-118315462 on Build GRCh38
rs202052957 Chr.11: 118315493-118315493 on Build GRCh38
rs201356888 Chr.11: 118315444-118315444 on Build GRCh38
rs73612255 Chr.11: 118315788-118315788 on Build GRCh38
rs200610341 Chr.11: 118315789-118315789 on Build GRCh38
rs1020283742 Chr.11: 118315520-118315520 on Build GRCh38
rs193922618 Chr.11: 118315643-118315644 on Build GRCh38
rs756421356 Chr.11: 118315504-118315504 on Build GRCh38
rs773914324 Chr.11: 118315585-118315585 on Build GRCh38
rs759238312 Chr.11: 118315497-118315497 on Build GRCh38
rs529037971 Chr.11: 118315369-118315369 on Build GRCh38
rs756615857 Chr.11: 118315451-118315451 on Build GRCh38
rs747915307 Chr.11: 118315473-118315473 on Build GRCh38
rs535622181 Chr.11: 118315680-118315680 on Build GRCh38
rs200750253 Chr.11: 118315470-118315470 on Build GRCh38
rs965994447 Chr.11: 118315374-118315374 on Build GRCh38
rs202081675 Chr.11: 118315669-118315669 on Build GRCh38
rs375023083 Chr.11: 118315325-118315325 on Build GRCh38
rs925663212 Chr.11: 118315679-118315679 on Build GRCh38
rs199582695 Chr.11: 118315467-118315467 on Build GRCh38
rs779066638 Chr.11: 118315525-118315525 on Build GRCh38
rs200314888 Chr.11: 118315701-118315701 on Build GRCh38
rs1040734829 Chr.11: 118315670-118315670 on Build GRCh38
rs201024718 Chr.11: 118315764-118315764 on Build GRCh38
rs982444878 Chr.11: 118315643-118315643 on Build GRCh38
rs376024825 Chr.11: 118315461-118315461 on Build GRCh38
rs201426171 Chr.11: 118315703-118315703 on Build GRCh38
rs201482209 Chr.11: 118315534-118315534 on Build GRCh38
rs762763746 Chr.11: 118315477-118315477 on Build GRCh38
rs187341356 Chr.11: 118315838-118315838 on Build GRCh38

View Full Product Details