Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 65170873-65171255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEMD3
Entrez Gene ID
23592
Gene Name
LEM domain containing 3
Gene Aliases
MAN1
UniGene
Hs.744150
Gene Chromosome Location
Chr.12: 65169571-65248361 on Build GRCh38
Transcripts
NM_001167614.1 NM_014319.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6700428 c.1363C>T p.Q455* Chr.12: 65170959-65170959 on Build GRCh38
1991498 c.1304C>A p.T435N Chr.12: 65170900-65170900 on Build GRCh38
1562496 c.1422G>A p.S474S Chr.12: 65171018-65171018 on Build GRCh38
302543 c.1347C>A p.P449P Chr.12: 65170943-65170943 on Build GRCh38
694775 c.1404C>A p.F468L Chr.12: 65171000-65171000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779934044 Chr.12: 65171039-65171039 on Build GRCh38
rs1022322698 Chr.12: 65171136-65171136 on Build GRCh38
rs755984431 Chr.12: 65170992-65170992 on Build GRCh38
rs763991535 Chr.12: 65171122-65171122 on Build GRCh38
rs371748000 Chr.12: 65170941-65170941 on Build GRCh38
rs766893227 Chr.12: 65170951-65170951 on Build GRCh38
rs769281226 Chr.12: 65171057-65171057 on Build GRCh38
rs1042013399 Chr.12: 65170958-65170958 on Build GRCh38
rs756668294 Chr.12: 65170951-65170951 on Build GRCh38
rs761761659 Chr.12: 65171138-65171138 on Build GRCh38
rs764079361 Chr.12: 65170993-65170993 on Build GRCh38
rs998078047 Chr.12: 65170990-65170990 on Build GRCh38
rs35470165 Chr.12: 65171221-65171221 on Build GRCh38
rs763178871 Chr.12: 65171148-65171148 on Build GRCh38
rs768302841 Chr.12: 65170900-65170900 on Build GRCh38
rs1026974518 Chr.12: 65171171-65171171 on Build GRCh38
rs745933801 Chr.12: 65170894-65170894 on Build GRCh38
rs866627066 Chr.12: 65170976-65170976 on Build GRCh38
rs770000098 Chr.12: 65171065-65171065 on Build GRCh38
rs200415494 Chr.12: 65171032-65171032 on Build GRCh38
rs773654384 Chr.12: 65171084-65171084 on Build GRCh38
rs145116568 Chr.12: 65170946-65170946 on Build GRCh38
rs199915804 Chr.12: 65170995-65170995 on Build GRCh38
rs948403045 Chr.12: 65171188-65171188 on Build GRCh38
rs747141497 Chr.12: 65171045-65171045 on Build GRCh38
rs985448420 Chr.12: 65171179-65171179 on Build GRCh38
rs774970317 Chr.12: 65171099-65171099 on Build GRCh38
rs978248116 Chr.12: 65171147-65171147 on Build GRCh38
rs991085420 Chr.12: 65171175-65171175 on Build GRCh38
rs376326383 Chr.12: 65171169-65171169 on Build GRCh38
rs760247162 Chr.12: 65171104-65171104 on Build GRCh38
rs753825412 Chr.12: 65171134-65171134 on Build GRCh38
rs954067307 Chr.12: 65171054-65171054 on Build GRCh38
rs993523845 Chr.12: 65171158-65171159 on Build GRCh38
rs547625447 Chr.12: 65171027-65171027 on Build GRCh38
rs1000436494 Chr.12: 65171001-65171001 on Build GRCh38
rs777316626 Chr.12: 65171061-65171061 on Build GRCh38
rs771107177 Chr.12: 65171157-65171159 on Build GRCh38
rs773555333 Chr.12: 65171124-65171125 on Build GRCh38
rs901104339 Chr.12: 65170978-65170978 on Build GRCh38
rs376067130 Chr.12: 65170989-65170989 on Build GRCh38
rs116829554 Chr.12: 65171156-65171156 on Build GRCh38
rs375079599 Chr.12: 65171110-65171110 on Build GRCh38
rs371901972 Chr.12: 65171023-65171023 on Build GRCh38
rs747480471 Chr.12: 65170915-65170915 on Build GRCh38
rs187683275 Chr.12: 65171183-65171183 on Build GRCh38
rs766097829 Chr.12: 65170937-65170937 on Build GRCh38
rs750238555 Chr.12: 65171154-65171154 on Build GRCh38
rs570092479 Chr.12: 65171233-65171233 on Build GRCh38
rs764902501 Chr.12: 65171140-65171140 on Build GRCh38
rs1012975927 Chr.12: 65171130-65171130 on Build GRCh38
rs762287082 Chr.12: 65171218-65171218 on Build GRCh38
rs754270566 Chr.12: 65171202-65171202 on Build GRCh38
rs758680061 Chr.12: 65171034-65171034 on Build GRCh38
rs758493364 Chr.12: 65171166-65171166 on Build GRCh38
rs759040154 Chr.12: 65171086-65171086 on Build GRCh38
rs144127577 Chr.12: 65170926-65170926 on Build GRCh38
rs775662292 Chr.12: 65170908-65170908 on Build GRCh38
rs149129862 Chr.12: 65171033-65171033 on Build GRCh38
rs569480939 Chr.12: 65170927-65170927 on Build GRCh38
rs762333334 Chr.12: 65170936-65170936 on Build GRCh38
rs142265837 Chr.12: 65171094-65171094 on Build GRCh38
rs764695092 Chr.12: 65171132-65171132 on Build GRCh38
rs868489650 Chr.12: 65170977-65170977 on Build GRCh38

View Full Product Details