Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238041549-238042124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2F
Entrez Gene ID
140739
Gene Name
ubiquitin conjugating enzyme E2 F (putative)
Gene Aliases
NCE2
UniGene
Hs.471785
Gene Chromosome Location
Chr.2: 237966945-238042782 on Build GRCh38
Transcripts
NM_001278305.1 NM_001278306.1 NM_001278307.1 NM_001278308.1 NM_080678.2 NR_103498.1 NR_103499.1 NR_103500.1 NR_037904.1

Gene Symbol
UBE2F-SCLY
Entrez Gene ID
100533179
Gene Name
UBE2F-SCLY readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.471785
Gene Chromosome Location
Chr.2: 237966945-238099413 on Build GRCh38
Transcripts
NM_001278305.1 NM_001278306.1 NM_001278307.1 NM_001278308.1 NM_080678.2 NR_103498.1 NR_103499.1 NR_103500.1 NR_037904.1

SNP Detail

SNP ID Chromosome Location
rs140762097 Chr.2: 238041760-238041760 on Build GRCh38
rs146423957 Chr.2: 238041707-238041707 on Build GRCh38
rs376232561 Chr.2: 238041719-238041719 on Build GRCh38
rs769176061 Chr.2: 238041737-238041737 on Build GRCh38
rs535141916 Chr.2: 238041709-238041709 on Build GRCh38
rs1041951807 Chr.2: 238041871-238041871 on Build GRCh38
rs559142390 Chr.2: 238042079-238042079 on Build GRCh38
rs894672437 Chr.2: 238041945-238041945 on Build GRCh38
rs550474911 Chr.2: 238041606-238041606 on Build GRCh38
rs988003329 Chr.2: 238041710-238041710 on Build GRCh38
rs973222134 Chr.2: 238041769-238041770 on Build GRCh38
rs963281892 Chr.2: 238042076-238042076 on Build GRCh38
rs1000367139 Chr.2: 238041656-238041656 on Build GRCh38
rs921676479 Chr.2: 238041819-238041819 on Build GRCh38
rs748228865 Chr.2: 238041872-238041872 on Build GRCh38
rs142873709 Chr.2: 238042100-238042101 on Build GRCh38
rs1047867 Chr.2: 238042004-238042004 on Build GRCh38
rs557562271 Chr.2: 238041666-238041666 on Build GRCh38
rs911539870 Chr.2: 238041854-238041855 on Build GRCh38
rs1051883850 Chr.2: 238041605-238041605 on Build GRCh38
rs578093825 Chr.2: 238041759-238041759 on Build GRCh38
rs926651864 Chr.2: 238041838-238041838 on Build GRCh38
rs946266967 Chr.2: 238041609-238041609 on Build GRCh38
rs1056587402 Chr.2: 238042051-238042051 on Build GRCh38
rs1014748782 Chr.2: 238041979-238041979 on Build GRCh38
rs896645416 Chr.2: 238042075-238042075 on Build GRCh38
rs979416684 Chr.2: 238041803-238041803 on Build GRCh38
rs987266770 Chr.2: 238041846-238041846 on Build GRCh38
rs963303031 Chr.2: 238041584-238041584 on Build GRCh38
rs985876434 Chr.2: 238041677-238041677 on Build GRCh38
rs573863968 Chr.2: 238041801-238041801 on Build GRCh38
rs1014763870 Chr.2: 238042018-238042018 on Build GRCh38
rs945733483 Chr.2: 238041867-238041867 on Build GRCh38
rs1014364910 Chr.2: 238041676-238041676 on Build GRCh38
rs557264200 Chr.2: 238041799-238041799 on Build GRCh38
rs936132551 Chr.2: 238041874-238041874 on Build GRCh38
rs536634238 Chr.2: 238041577-238041577 on Build GRCh38
rs1029227449 Chr.2: 238042102-238042102 on Build GRCh38
rs746675977 Chr.2: 238041654-238041654 on Build GRCh38
rs570247055 Chr.2: 238041642-238041642 on Build GRCh38
rs542861026 Chr.2: 238041811-238041811 on Build GRCh38
rs192911996 Chr.2: 238041938-238041938 on Build GRCh38
rs931703841 Chr.2: 238041829-238041829 on Build GRCh38
rs1004179301 Chr.2: 238042001-238042001 on Build GRCh38
rs1042025429 Chr.2: 238041624-238041624 on Build GRCh38
rs536084949 Chr.2: 238041660-238041660 on Build GRCh38
rs538101880 Chr.2: 238042095-238042096 on Build GRCh38
rs904789988 Chr.2: 238041873-238041873 on Build GRCh38
rs1024435404 Chr.2: 238041986-238041986 on Build GRCh38
rs555690401 Chr.2: 238041670-238041670 on Build GRCh38
rs938108297 Chr.2: 238042000-238042000 on Build GRCh38
rs994491246 Chr.2: 238041602-238041602 on Build GRCh38

View Full Product Details