Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34882635-34882922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSS2
Entrez Gene ID
55902
Gene Name
acyl-CoA synthetase short-chain family member 2
Gene Aliases
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1
UniGene
Hs.517034
Gene Chromosome Location
Chr.20: 34874942-34927966 on Build GRCh38
Transcripts
NM_001076552.2 NM_001242393.1 NM_018677.3 XM_005260455.1 XM_005260456.1 XM_006723826.1 XM_011528905.1 XM_011528906.1 XM_011528907.1 XM_011528908.1 XM_011528909.2 XM_011528911.1 XM_011528912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1166862 c.286A>T p.M96L Chr.20: 34882901-34882901 on Build GRCh38
4473264 c.183C>T p.F61F Chr.20: 34882798-34882798 on Build GRCh38
5425553 c.276C>T p.F92F Chr.20: 34882891-34882891 on Build GRCh38
1026125 c.248A>C p.N83T Chr.20: 34882863-34882863 on Build GRCh38
7156420 c.40G>T p.D14Y Chr.20: 34882805-34882805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368562342 Chr.20: 34882767-34882767 on Build GRCh38
rs577002078 Chr.20: 34882722-34882722 on Build GRCh38
rs969340970 Chr.20: 34882825-34882825 on Build GRCh38
rs139489319 Chr.20: 34882893-34882893 on Build GRCh38
rs746176589 Chr.20: 34882821-34882821 on Build GRCh38
rs774402427 Chr.20: 34882876-34882876 on Build GRCh38
rs776624293 Chr.20: 34882773-34882773 on Build GRCh38
rs754574068 Chr.20: 34882777-34882777 on Build GRCh38
rs768784749 Chr.20: 34882860-34882860 on Build GRCh38
rs888044565 Chr.20: 34882689-34882689 on Build GRCh38
rs1025297158 Chr.20: 34882677-34882677 on Build GRCh38
rs192729072 Chr.20: 34882746-34882746 on Build GRCh38
rs750128848 Chr.20: 34882764-34882764 on Build GRCh38
rs770080429 Chr.20: 34882852-34882852 on Build GRCh38
rs575177684 Chr.20: 34882714-34882717 on Build GRCh38
rs200746034 Chr.20: 34882768-34882768 on Build GRCh38
rs4911163 Chr.20: 34882891-34882891 on Build GRCh38
rs757644302 Chr.20: 34882815-34882815 on Build GRCh38
rs778562096 Chr.20: 34882787-34882787 on Build GRCh38
rs781621444 Chr.20: 34882817-34882817 on Build GRCh38
rs897908364 Chr.20: 34882714-34882714 on Build GRCh38
rs768795451 Chr.20: 34882871-34882871 on Build GRCh38
rs60379405 Chr.20: 34882856-34882856 on Build GRCh38
rs771840227 Chr.20: 34882887-34882887 on Build GRCh38
rs1027425106 Chr.20: 34882870-34882870 on Build GRCh38
rs145989248 Chr.20: 34882733-34882733 on Build GRCh38
rs374435624 Chr.20: 34882748-34882748 on Build GRCh38
rs1050597313 Chr.20: 34882676-34882676 on Build GRCh38
rs868388083 Chr.20: 34882813-34882813 on Build GRCh38
rs761652151 Chr.20: 34882747-34882747 on Build GRCh38
rs187831610 Chr.20: 34882715-34882715 on Build GRCh38
rs371597129 Chr.20: 34882774-34882774 on Build GRCh38
rs761827348 Chr.20: 34882877-34882877 on Build GRCh38
rs770915049 Chr.20: 34882755-34882757 on Build GRCh38
rs747572708 Chr.20: 34882793-34882793 on Build GRCh38
rs57031852 Chr.20: 34882857-34882857 on Build GRCh38
rs6088638 Chr.20: 34882711-34882711 on Build GRCh38
rs1006360303 Chr.20: 34882702-34882702 on Build GRCh38

View Full Product Details