Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10085677-10086157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCD2
Entrez Gene ID
2177
Gene Name
Fanconi anemia complementation group D2
Gene Aliases
FA-D2, FA4, FACD, FAD, FAD2, FANCD
UniGene
Hs.208388
Gene Chromosome Location
Chr.3: 10026387-10101937 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Gene Symbol
FANCD2OS
Entrez Gene ID
115795
Gene Name
FANCD2 opposite strand
Gene Aliases
C3orf24
UniGene
Hs.190413
Gene Chromosome Location
Chr.3: 10081320-10108603 on Build GRCh38
Transcripts
NM_001018115.2 NM_001319984.1 NM_033084.4 NM_173472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895677 c.3225-6C>T p.? Chr.3: 10085806-10085806 on Build GRCh38
7154913 c.3304G>A p.E1102K Chr.3: 10085891-10085891 on Build GRCh38
4777223 c.3298C>G p.Q1100E Chr.3: 10085885-10085885 on Build GRCh38
5895678 c.3225-6C>T p.? Chr.3: 10085806-10085806 on Build GRCh38
1308326 c.3333C>T p.L1111L Chr.3: 10085920-10085920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748717972 Chr.3: 10085936-10085936 on Build GRCh38
rs773833838 Chr.3: 10085863-10085863 on Build GRCh38
rs773636192 Chr.3: 10085940-10085940 on Build GRCh38
rs770003791 Chr.3: 10085938-10085938 on Build GRCh38
rs747078545 Chr.3: 10085930-10085930 on Build GRCh38
rs767075732 Chr.3: 10085873-10085873 on Build GRCh38
rs759252565 Chr.3: 10085872-10085872 on Build GRCh38
rs192843638 Chr.3: 10085839-10085839 on Build GRCh38
rs956067003 Chr.3: 10085920-10085920 on Build GRCh38
rs983213312 Chr.3: 10086026-10086026 on Build GRCh38
rs755748094 Chr.3: 10085877-10085877 on Build GRCh38
rs984990877 Chr.3: 10085715-10085715 on Build GRCh38
rs776972450 Chr.3: 10085933-10085933 on Build GRCh38
rs775336123 Chr.3: 10085953-10085953 on Build GRCh38
rs753672518 Chr.3: 10085963-10085963 on Build GRCh38
rs951909943 Chr.3: 10085711-10085711 on Build GRCh38
rs757127466 Chr.3: 10085788-10085788 on Build GRCh38
rs748843683 Chr.3: 10085886-10085889 on Build GRCh38
rs758625993 Chr.3: 10085908-10085908 on Build GRCh38
rs763730607 Chr.3: 10085962-10085962 on Build GRCh38
rs923309974 Chr.3: 10086092-10086092 on Build GRCh38
rs372015176 Chr.3: 10085969-10085969 on Build GRCh38
rs139929432 Chr.3: 10085921-10085921 on Build GRCh38
rs1026065640 Chr.3: 10085982-10085982 on Build GRCh38
rs759189761 Chr.3: 10085941-10085941 on Build GRCh38
rs55736797 Chr.3: 10086073-10086073 on Build GRCh38
rs779586699 Chr.3: 10086066-10086066 on Build GRCh38
rs746020754 Chr.3: 10085903-10085903 on Build GRCh38
rs778794561 Chr.3: 10085897-10085897 on Build GRCh38
rs944411499 Chr.3: 10086074-10086074 on Build GRCh38
rs554557253 Chr.3: 10086019-10086019 on Build GRCh38
rs765672800 Chr.3: 10085862-10085862 on Build GRCh38
rs772092834 Chr.3: 10085828-10085828 on Build GRCh38
rs975960824 Chr.3: 10086075-10086075 on Build GRCh38
rs980259409 Chr.3: 10085979-10085979 on Build GRCh38
rs777639581 Chr.3: 10085783-10085783 on Build GRCh38
rs775561668 Chr.3: 10085829-10085829 on Build GRCh38
rs1000254003 Chr.3: 10085865-10085865 on Build GRCh38
rs569070151 Chr.3: 10085875-10085875 on Build GRCh38
rs1040693550 Chr.3: 10086085-10086085 on Build GRCh38
rs947583281 Chr.3: 10085912-10085912 on Build GRCh38
rs747450518 Chr.3: 10085837-10085837 on Build GRCh38
rs764105093 Chr.3: 10085878-10085878 on Build GRCh38
rs551095369 Chr.3: 10085735-10085735 on Build GRCh38
rs143239014 Chr.3: 10085734-10085734 on Build GRCh38
rs199616966 Chr.3: 10085786-10085786 on Build GRCh38
rs756129956 Chr.3: 10085774-10085774 on Build GRCh38
rs770686014 Chr.3: 10085886-10085887 on Build GRCh38
rs1024599037 Chr.3: 10085960-10085960 on Build GRCh38
rs55861547 Chr.3: 10085889-10085889 on Build GRCh38
rs35770561 Chr.3: 10086132-10086132 on Build GRCh38
rs777514146 Chr.3: 10085877-10085880 on Build GRCh38
rs762060936 Chr.3: 10085848-10085848 on Build GRCh38
rs746135951 Chr.3: 10085809-10085809 on Build GRCh38
rs536262348 Chr.3: 10086011-10086011 on Build GRCh38
rs769147120 Chr.3: 10085838-10085838 on Build GRCh38
rs764876175 Chr.3: 10085971-10085971 on Build GRCh38
rs368121256 Chr.3: 10085958-10085958 on Build GRCh38
rs767270364 Chr.3: 10085946-10085946 on Build GRCh38
rs778974182 Chr.3: 10085796-10085796 on Build GRCh38
rs183138323 Chr.3: 10086005-10086005 on Build GRCh38
rs534751086 Chr.3: 10085696-10085696 on Build GRCh38
rs752256004 Chr.3: 10085874-10085874 on Build GRCh38
rs996065278 Chr.3: 10085799-10085799 on Build GRCh38

View Full Product Details