Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 134254286-134254839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT5
Entrez Gene ID
4249
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase
Gene Aliases
GNT-V, GNT-VA
UniGene
Hs.4988
Gene Chromosome Location
Chr.2: 134119922-134454621 on Build GRCh38
Transcripts
NM_002410.4 XM_005263668.4 XM_005263669.4 XM_005263670.3 XM_006712534.3 XM_011511199.2 XM_011511200.1 XM_011511201.2 XM_011511202.1 XM_011511204.2 XM_017004147.1 XM_017004148.1 XM_017004149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
715708 c.120T>G p.P40P Chr.2: 134254523-134254523 on Build GRCh38
715707 c.224G>T p.G75V Chr.2: 134254627-134254627 on Build GRCh38
325808 c.163A>C p.K55Q Chr.2: 134254566-134254566 on Build GRCh38
5641519 c.162C>A p.S54R Chr.2: 134254565-134254565 on Build GRCh38
1399459 c.2T>C p.M1T Chr.2: 134254405-134254405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755103763 Chr.2: 134254577-134254577 on Build GRCh38
rs921114881 Chr.2: 134254506-134254506 on Build GRCh38
rs138863444 Chr.2: 134254468-134254468 on Build GRCh38
rs375743556 Chr.2: 134254366-134254366 on Build GRCh38
rs920169938 Chr.2: 134254665-134254665 on Build GRCh38
rs750952781 Chr.2: 134254566-134254566 on Build GRCh38
rs1034767237 Chr.2: 134254718-134254718 on Build GRCh38
rs751689227 Chr.2: 134254574-134254574 on Build GRCh38
rs749108316 Chr.2: 134254382-134254382 on Build GRCh38
rs960360973 Chr.2: 134254729-134254729 on Build GRCh38
rs757666834 Chr.2: 134254421-134254421 on Build GRCh38
rs559878042 Chr.2: 134254420-134254420 on Build GRCh38
rs969076385 Chr.2: 134254335-134254335 on Build GRCh38
rs760178949 Chr.2: 134254533-134254533 on Build GRCh38
rs908495139 Chr.2: 134254401-134254401 on Build GRCh38
rs758987140 Chr.2: 134254570-134254570 on Build GRCh38
rs962631877 Chr.2: 134254774-134254774 on Build GRCh38
rs561212748 Chr.2: 134254516-134254516 on Build GRCh38
rs749127602 Chr.2: 134254461-134254461 on Build GRCh38
rs765762056 Chr.2: 134254562-134254562 on Build GRCh38
rs760320533 Chr.2: 134254544-134254544 on Build GRCh38
rs373932293 Chr.2: 134254365-134254365 on Build GRCh38
rs777389323 Chr.2: 134254455-134254455 on Build GRCh38
rs747167679 Chr.2: 134254433-134254433 on Build GRCh38
rs761429621 Chr.2: 134254669-134254669 on Build GRCh38
rs764308294 Chr.2: 134254418-134254418 on Build GRCh38
rs770683241 Chr.2: 134254475-134254475 on Build GRCh38
rs755229226 Chr.2: 134254445-134254445 on Build GRCh38
rs867025604 Chr.2: 134254394-134254394 on Build GRCh38
rs1009463234 Chr.2: 134254815-134254815 on Build GRCh38
rs748361192 Chr.2: 134254607-134254607 on Build GRCh38
rs980501914 Chr.2: 134254350-134254350 on Build GRCh38
rs532198105 Chr.2: 134254765-134254765 on Build GRCh38
rs781457066 Chr.2: 134254584-134254584 on Build GRCh38
rs770224440 Chr.2: 134254671-134254671 on Build GRCh38
rs567277206 Chr.2: 134254808-134254808 on Build GRCh38
rs768091051 Chr.2: 134254662-134254662 on Build GRCh38
rs750738147 Chr.2: 134254716-134254716 on Build GRCh38
rs893717813 Chr.2: 134254777-134254777 on Build GRCh38
rs1041134132 Chr.2: 134254707-134254707 on Build GRCh38
rs773069653 Chr.2: 134254375-134254375 on Build GRCh38
rs527500629 Chr.2: 134254427-134254427 on Build GRCh38
rs778972339 Chr.2: 134254622-134254622 on Build GRCh38
rs771561378 Chr.2: 134254521-134254521 on Build GRCh38
rs1023554602 Chr.2: 134254314-134254314 on Build GRCh38
rs750631332 Chr.2: 134254685-134254704 on Build GRCh38
rs941279048 Chr.2: 134254413-134254413 on Build GRCh38
rs996844775 Chr.2: 134254748-134254748 on Build GRCh38
rs531539882 Chr.2: 134254706-134254706 on Build GRCh38
rs116127374 Chr.2: 134254710-134254710 on Build GRCh38
rs751005506 Chr.2: 134254428-134254428 on Build GRCh38
rs759632867 Chr.2: 134254694-134254694 on Build GRCh38
rs775123588 Chr.2: 134254404-134254404 on Build GRCh38
rs548835362 Chr.2: 134254513-134254513 on Build GRCh38
rs766496355 Chr.2: 134254572-134254572 on Build GRCh38
rs397820430 Chr.2: 134254815-134254816 on Build GRCh38
rs759026906 Chr.2: 134254387-134254387 on Build GRCh38
rs545103804 Chr.2: 134254374-134254374 on Build GRCh38
rs772002787 Chr.2: 134254630-134254630 on Build GRCh38
rs1012250418 Chr.2: 134254657-134254657 on Build GRCh38
rs996792254 Chr.2: 134254340-134254340 on Build GRCh38
rs982429983 Chr.2: 134254736-134254736 on Build GRCh38
rs145506782 Chr.2: 134254759-134254759 on Build GRCh38
rs1023104855 Chr.2: 134254663-134254663 on Build GRCh38
rs974417363 Chr.2: 134254805-134254805 on Build GRCh38
rs746553155 Chr.2: 134254645-134254645 on Build GRCh38
rs757046400 Chr.2: 134254619-134254619 on Build GRCh38
rs376572635 Chr.2: 134254364-134254364 on Build GRCh38
rs566213116 Chr.2: 134254797-134254797 on Build GRCh38
rs774995015 Chr.2: 134254522-134254522 on Build GRCh38
rs773712933 Chr.2: 134254674-134254674 on Build GRCh38
rs770928798 Chr.2: 134254383-134254383 on Build GRCh38
rs138982519 Chr.2: 134254623-134254623 on Build GRCh38
rs370766400 Chr.2: 134254667-134254667 on Build GRCh38
rs760435313 Chr.2: 134254411-134254411 on Build GRCh38
rs773806086 Chr.2: 134254385-134254385 on Build GRCh38
rs990370760 Chr.2: 134254313-134254313 on Build GRCh38
rs754289087 Chr.2: 134254354-134254354 on Build GRCh38
rs767058940 Chr.2: 134254400-134254400 on Build GRCh38
rs887258707 Chr.2: 134254677-134254678 on Build GRCh38
rs758547800 Chr.2: 134254430-134254430 on Build GRCh38
rs763840693 Chr.2: 134254540-134254540 on Build GRCh38
rs998888459 Chr.2: 134254548-134254548 on Build GRCh38
rs763510053 Chr.2: 134254676-134254676 on Build GRCh38
rs752052217 Chr.2: 134254679-134254679 on Build GRCh38
rs779501051 Chr.2: 134254632-134254632 on Build GRCh38
rs3833551 Chr.2: 134254811-134254812 on Build GRCh38
rs772029188 Chr.2: 134254733-134254733 on Build GRCh38
rs200509590 Chr.2: 134254367-134254367 on Build GRCh38
rs141403771 Chr.2: 134254543-134254543 on Build GRCh38

View Full Product Details